Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 37413933-37414434 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF569
Entrez Gene ID
148266
Gene Name
zinc finger protein 569
Gene Aliases
ZAP1, ZNF
UniGene
Hs.511848
Gene Chromosome Location
Chr.19: 37411146-37469268 on Build GRCh38
Transcripts
NM_152484.2 XM_006723046.2 XM_006723047.3 XM_006723048.3 XM_011526538.2 XM_011526539.2 XM_017026376.1 XM_017026377.1 XM_017026378.1 XM_017026379.1 XM_017026380.1 XM_017026381.1 XM_017026382.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6831295 c.486G>T p.K162N Chr.19: 37414172-37414172 on Build GRCh38
7295541 c.261G>A p.E87E Chr.19: 37414397-37414397 on Build GRCh38
3533179 c.366A>G p.V122V Chr.19: 37414292-37414292 on Build GRCh38
4499972 c.554C>T p.P185L Chr.19: 37414104-37414104 on Build GRCh38
7403296 c.395C>G p.S132C Chr.19: 37414263-37414263 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757554172 Chr.19: 37414253-37414253 on Build GRCh38
rs900172658 Chr.19: 37414193-37414193 on Build GRCh38
rs745588907 Chr.19: 37414149-37414149 on Build GRCh38
rs775141489 Chr.19: 37414036-37414036 on Build GRCh38
rs777881861 Chr.19: 37414119-37414119 on Build GRCh38
rs760293971 Chr.19: 37414322-37414322 on Build GRCh38
rs751650319 Chr.19: 37414248-37414248 on Build GRCh38
rs758663915 Chr.19: 37413966-37413966 on Build GRCh38
rs773071917 Chr.19: 37414283-37414283 on Build GRCh38
rs1007476978 Chr.19: 37414011-37414011 on Build GRCh38
rs755466945 Chr.19: 37413959-37413959 on Build GRCh38
rs565886241 Chr.19: 37414168-37414168 on Build GRCh38
rs771647457 Chr.19: 37414184-37414184 on Build GRCh38
rs1004561129 Chr.19: 37414384-37414384 on Build GRCh38
rs764134579 Chr.19: 37414247-37414247 on Build GRCh38
rs570264602 Chr.19: 37414268-37414268 on Build GRCh38
rs750264422 Chr.19: 37414257-37414257 on Build GRCh38
rs762312182 Chr.19: 37414041-37414041 on Build GRCh38
rs757484800 Chr.19: 37414137-37414137 on Build GRCh38
rs565020205 Chr.19: 37414002-37414002 on Build GRCh38
rs571379430 Chr.19: 37414399-37414399 on Build GRCh38
rs755930620 Chr.19: 37414265-37414265 on Build GRCh38
rs370846456 Chr.19: 37414008-37414008 on Build GRCh38
rs140277147 Chr.19: 37414166-37414166 on Build GRCh38
rs532445392 Chr.19: 37414071-37414071 on Build GRCh38
rs752800971 Chr.19: 37414111-37414111 on Build GRCh38
rs776541895 Chr.19: 37414007-37414007 on Build GRCh38
rs146428389 Chr.19: 37414230-37414230 on Build GRCh38
rs759848319 Chr.19: 37414205-37414205 on Build GRCh38
rs781503560 Chr.19: 37414256-37414256 on Build GRCh38
rs77693590 Chr.19: 37414315-37414315 on Build GRCh38
rs770196275 Chr.19: 37414325-37414325 on Build GRCh38
rs367716862 Chr.19: 37414200-37414200 on Build GRCh38
rs778293381 Chr.19: 37414294-37414294 on Build GRCh38
rs762570930 Chr.19: 37413994-37413994 on Build GRCh38
rs748160273 Chr.19: 37413963-37413963 on Build GRCh38
rs751286253 Chr.19: 37414104-37414104 on Build GRCh38
rs754198051 Chr.19: 37414080-37414080 on Build GRCh38
rs763433392 Chr.19: 37414338-37414338 on Build GRCh38
rs758372127 Chr.19: 37414114-37414114 on Build GRCh38
rs765767600 Chr.19: 37414215-37414215 on Build GRCh38
rs751775417 Chr.19: 37414127-37414127 on Build GRCh38
rs372002839 Chr.19: 37414244-37414244 on Build GRCh38
rs760040745 Chr.19: 37414088-37414088 on Build GRCh38
rs374997645 Chr.19: 37414151-37414151 on Build GRCh38
rs763830347 Chr.19: 37414056-37414056 on Build GRCh38
rs763651392 Chr.19: 37414010-37414010 on Build GRCh38
rs779032580 Chr.19: 37413961-37413961 on Build GRCh38
rs947458080 Chr.19: 37414353-37414353 on Build GRCh38
rs201465276 Chr.19: 37414347-37414347 on Build GRCh38
rs761494365 Chr.19: 37414068-37414068 on Build GRCh38
rs775907398 Chr.19: 37414337-37414337 on Build GRCh38
rs765677984 Chr.19: 37414106-37414106 on Build GRCh38
rs761934677 Chr.19: 37414408-37414408 on Build GRCh38
rs148474590 Chr.19: 37414203-37414203 on Build GRCh38
rs768634649 Chr.19: 37414286-37414286 on Build GRCh38
rs377591054 Chr.19: 37414120-37414120 on Build GRCh38
rs780089041 Chr.19: 37414160-37414160 on Build GRCh38
rs77849131 Chr.19: 37414341-37414341 on Build GRCh38
rs773833899 Chr.19: 37414060-37414060 on Build GRCh38
rs764592188 Chr.19: 37414339-37414339 on Build GRCh38
rs140750555 Chr.19: 37414314-37414314 on Build GRCh38
rs374062239 Chr.19: 37414017-37414017 on Build GRCh38
rs201479981 Chr.19: 37414142-37414142 on Build GRCh38
rs144008113 Chr.19: 37414171-37414171 on Build GRCh38
rs914694812 Chr.19: 37414316-37414316 on Build GRCh38
rs752923644 Chr.19: 37414268-37414269 on Build GRCh38
rs891259957 Chr.19: 37414139-37414139 on Build GRCh38
rs868579479 Chr.19: 37414000-37414000 on Build GRCh38
rs777814000 Chr.19: 37413969-37413969 on Build GRCh38

View Full Product Details