Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 45718704-45719248 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IPP
Entrez Gene ID
3652
Gene Name
intracisternal A particle-promoted polypeptide
Gene Aliases
KLHL27
UniGene
Hs.744252
Gene Chromosome Location
Chr.1: 45694324-45750813 on Build GRCh38
Transcripts
NM_001145349.1 NM_005897.2 XM_005270839.4 XM_006710623.3 XM_011541394.2 XM_017001210.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4692857 c.1163_1164delGT p.C388fs*14 Chr.1: 45719225-45719226 on Build GRCh38
5532780 c.1176C>T p.I392I Chr.1: 45719213-45719213 on Build GRCh38
5532779 c.1176C>T p.I392I Chr.1: 45719213-45719213 on Build GRCh38
4692856 c.1163_1164delGT p.C388fs*14 Chr.1: 45719225-45719226 on Build GRCh38
1254899 c.1144G>A p.G382S Chr.1: 45719245-45719245 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751452108 Chr.1: 45719215-45719215 on Build GRCh38
rs1043131001 Chr.1: 45718883-45718883 on Build GRCh38
rs571731648 Chr.1: 45718898-45718898 on Build GRCh38
rs934517531 Chr.1: 45718918-45718919 on Build GRCh38
rs762177871 Chr.1: 45719194-45719194 on Build GRCh38
rs976991253 Chr.1: 45718875-45718875 on Build GRCh38
rs758517801 Chr.1: 45719222-45719222 on Build GRCh38
rs190061722 Chr.1: 45719020-45719020 on Build GRCh38
rs952906197 Chr.1: 45718762-45718762 on Build GRCh38
rs766428573 Chr.1: 45719214-45719214 on Build GRCh38
rs553599817 Chr.1: 45719147-45719147 on Build GRCh38
rs746830370 Chr.1: 45719111-45719114 on Build GRCh38
rs753031529 Chr.1: 45719153-45719153 on Build GRCh38
rs146332127 Chr.1: 45719103-45719103 on Build GRCh38
rs895612539 Chr.1: 45718936-45718936 on Build GRCh38
rs972268225 Chr.1: 45718918-45718918 on Build GRCh38
rs149362026 Chr.1: 45719081-45719081 on Build GRCh38
rs1046417066 Chr.1: 45719115-45719115 on Build GRCh38
rs908177488 Chr.1: 45718837-45718837 on Build GRCh38
rs557511639 Chr.1: 45719016-45719016 on Build GRCh38
rs953865733 Chr.1: 45719169-45719169 on Build GRCh38
rs544803641 Chr.1: 45719206-45719206 on Build GRCh38
rs59512208 Chr.1: 45718918-45718918 on Build GRCh38
rs111912657 Chr.1: 45718923-45718923 on Build GRCh38
rs1033559379 Chr.1: 45718745-45718745 on Build GRCh38
rs1002192082 Chr.1: 45719000-45719000 on Build GRCh38
rs925672404 Chr.1: 45718902-45718903 on Build GRCh38
rs961048470 Chr.1: 45718863-45718863 on Build GRCh38
rs144663569 Chr.1: 45719225-45719226 on Build GRCh38
rs950018930 Chr.1: 45718951-45718951 on Build GRCh38
rs765528080 Chr.1: 45719195-45719195 on Build GRCh38
rs905210063 Chr.1: 45718982-45718982 on Build GRCh38
rs978418056 Chr.1: 45719193-45719193 on Build GRCh38
rs535024888 Chr.1: 45718730-45718730 on Build GRCh38
rs553454831 Chr.1: 45718854-45718854 on Build GRCh38
rs983917843 Chr.1: 45718806-45718806 on Build GRCh38
rs372037221 Chr.1: 45719085-45719085 on Build GRCh38
rs920935426 Chr.1: 45718878-45718878 on Build GRCh38
rs933592209 Chr.1: 45718955-45718955 on Build GRCh38
rs780868379 Chr.1: 45719228-45719228 on Build GRCh38
rs1011790112 Chr.1: 45718868-45718868 on Build GRCh38
rs998465700 Chr.1: 45718902-45718902 on Build GRCh38
rs542905114 Chr.1: 45719042-45719042 on Build GRCh38
rs772702368 Chr.1: 45719161-45719161 on Build GRCh38
rs769511705 Chr.1: 45719166-45719166 on Build GRCh38
rs910742972 Chr.1: 45719006-45719006 on Build GRCh38
rs952552495 Chr.1: 45718792-45718792 on Build GRCh38
rs1050579243 Chr.1: 45718977-45718977 on Build GRCh38
rs902826113 Chr.1: 45718900-45718900 on Build GRCh38
rs559149190 Chr.1: 45719126-45719126 on Build GRCh38
rs777006275 Chr.1: 45719191-45719191 on Build GRCh38
rs998855969 Chr.1: 45719130-45719130 on Build GRCh38
rs906458228 Chr.1: 45719154-45719154 on Build GRCh38
rs199741535 Chr.1: 45719177-45719177 on Build GRCh38
rs761177512 Chr.1: 45719174-45719174 on Build GRCh38
rs754901252 Chr.1: 45719216-45719216 on Build GRCh38
rs750601627 Chr.1: 45719198-45719198 on Build GRCh38
rs1009531262 Chr.1: 45719090-45719090 on Build GRCh38

View Full Product Details