Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 51356109-51356633 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPS15
Entrez Gene ID
2060
Gene Name
epidermal growth factor receptor pathway substrate 15
Gene Aliases
AF-1P, AF1P, MLLT5
UniGene
Hs.83722
Gene Chromosome Location
Chr.1: 51354263-51519354 on Build GRCh38
Transcripts
NM_001159969.1 NM_001981.2 XM_005270618.2 XM_017000615.1 XM_017000616.1 XM_017000617.1 XM_017000618.1 XM_017000619.1 XM_017000620.1

SNP Detail

SNP ID Chromosome Location
rs934500567 Chr.1: 51356184-51356184 on Build GRCh38
rs1054554160 Chr.1: 51356197-51356197 on Build GRCh38
rs540976530 Chr.1: 51356392-51356392 on Build GRCh38
rs934864859 Chr.1: 51356193-51356193 on Build GRCh38
rs1054278153 Chr.1: 51356528-51356530 on Build GRCh38
rs1052924351 Chr.1: 51356185-51356185 on Build GRCh38
rs548871924 Chr.1: 51356302-51356302 on Build GRCh38
rs975598214 Chr.1: 51356325-51356325 on Build GRCh38
rs935391268 Chr.1: 51356498-51356498 on Build GRCh38
rs557071188 Chr.1: 51356510-51356510 on Build GRCh38
rs936348626 Chr.1: 51356513-51356513 on Build GRCh38
rs903444652 Chr.1: 51356172-51356172 on Build GRCh38
rs41292519 Chr.1: 51356274-51356274 on Build GRCh38
rs982658494 Chr.1: 51356289-51356289 on Build GRCh38
rs551448857 Chr.1: 51356201-51356201 on Build GRCh38
rs963529853 Chr.1: 51356369-51356369 on Build GRCh38
rs1044283370 Chr.1: 51356242-51356246 on Build GRCh38
rs1054607145 Chr.1: 51356470-51356470 on Build GRCh38
rs144646125 Chr.1: 51356296-51356296 on Build GRCh38
rs1044912663 Chr.1: 51356570-51356570 on Build GRCh38
rs924239837 Chr.1: 51356395-51356395 on Build GRCh38
rs1023036141 Chr.1: 51356211-51356211 on Build GRCh38
rs914440617 Chr.1: 51356548-51356549 on Build GRCh38
rs962510540 Chr.1: 51356305-51356305 on Build GRCh38
rs922160145 Chr.1: 51356391-51356391 on Build GRCh38
rs951286279 Chr.1: 51356284-51356284 on Build GRCh38
rs948895494 Chr.1: 51356512-51356512 on Build GRCh38
rs1002527255 Chr.1: 51356261-51356261 on Build GRCh38
rs1034966973 Chr.1: 51356283-51356283 on Build GRCh38
rs36048901 Chr.1: 51356293-51356294 on Build GRCh38
rs907219532 Chr.1: 51356224-51356224 on Build GRCh38
rs924024825 Chr.1: 51356166-51356166 on Build GRCh38
rs540177134 Chr.1: 51356372-51356372 on Build GRCh38
rs183866695 Chr.1: 51356505-51356505 on Build GRCh38
rs921004575 Chr.1: 51356370-51356370 on Build GRCh38
rs907275826 Chr.1: 51356596-51356596 on Build GRCh38
rs947211734 Chr.1: 51356560-51356560 on Build GRCh38
rs140695546 Chr.1: 51356252-51356252 on Build GRCh38
rs372886639 Chr.1: 51356516-51356516 on Build GRCh38
rs538683476 Chr.1: 51356506-51356506 on Build GRCh38
rs9436905 Chr.1: 51356458-51356458 on Build GRCh38
rs111272942 Chr.1: 51356548-51356548 on Build GRCh38
rs1013430979 Chr.1: 51356205-51356205 on Build GRCh38
rs878936060 Chr.1: 51356517-51356517 on Build GRCh38
rs905567818 Chr.1: 51356249-51356249 on Build GRCh38
rs1052305345 Chr.1: 51356138-51356138 on Build GRCh38
rs34069014 Chr.1: 51356409-51356410 on Build GRCh38
rs778140757 Chr.1: 51356547-51356548 on Build GRCh38

View Full Product Details