Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 85930921-85931419 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP24
Entrez Gene ID
83478
Gene Name
Rho GTPase activating protein 24
Gene Aliases
FILGAP, RC-GAP72, RCGAP72, p73, p73RhoGAP
UniGene
Hs.444229
Gene Chromosome Location
Chr.4: 85475131-86002670 on Build GRCh38
Transcripts
NM_001025616.2 NM_001042669.1 NM_001287805.1 NM_031305.2 XM_011532300.2 XM_017008679.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6603275 c.21_22insG p.C10fs*14 Chr.4: 85930951-85930952 on Build GRCh38
7260820 c.3G>A p.M1I Chr.4: 85930933-85930933 on Build GRCh38
1245336 c.93G>T p.R31R Chr.4: 85931023-85931023 on Build GRCh38
6603282 c.33G>A p.P11P Chr.4: 85930963-85930963 on Build GRCh38
4003179 c.14G>T p.R5L Chr.4: 85930944-85930944 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs536911580 Chr.4: 85931319-85931319 on Build GRCh38
rs758584424 Chr.4: 85930954-85930955 on Build GRCh38
rs916380264 Chr.4: 85931056-85931056 on Build GRCh38
rs770001554 Chr.4: 85931009-85931009 on Build GRCh38
rs200870957 Chr.4: 85931309-85931310 on Build GRCh38
rs755809169 Chr.4: 85930959-85930959 on Build GRCh38
rs346499 Chr.4: 85931386-85931386 on Build GRCh38
rs951439326 Chr.4: 85931348-85931348 on Build GRCh38
rs1037734989 Chr.4: 85931391-85931391 on Build GRCh38
rs1008112831 Chr.4: 85931220-85931235 on Build GRCh38
rs1026942895 Chr.4: 85931331-85931331 on Build GRCh38
rs926812394 Chr.4: 85931063-85931063 on Build GRCh38
rs374481748 Chr.4: 85931306-85931306 on Build GRCh38
rs371286845 Chr.4: 85931153-85931153 on Build GRCh38
rs770856962 Chr.4: 85931126-85931126 on Build GRCh38
rs551685168 Chr.4: 85931129-85931129 on Build GRCh38
rs60002814 Chr.4: 85931055-85931055 on Build GRCh38
rs1024946543 Chr.4: 85931367-85931367 on Build GRCh38
rs530095654 Chr.4: 85931177-85931177 on Build GRCh38
rs373242250 Chr.4: 85930970-85930970 on Build GRCh38
rs749996308 Chr.4: 85930951-85930951 on Build GRCh38
rs951294867 Chr.4: 85931369-85931369 on Build GRCh38
rs549121157 Chr.4: 85931258-85931258 on Build GRCh38
rs766986929 Chr.4: 85931048-85931048 on Build GRCh38
rs749923387 Chr.4: 85931066-85931066 on Build GRCh38
rs890688017 Chr.4: 85931265-85931265 on Build GRCh38
rs996327011 Chr.4: 85931223-85931223 on Build GRCh38
rs149218128 Chr.4: 85931283-85931283 on Build GRCh38
rs981966861 Chr.4: 85931347-85931347 on Build GRCh38
rs1010660586 Chr.4: 85931320-85931320 on Build GRCh38
rs767378971 Chr.4: 85931384-85931384 on Build GRCh38
rs554038753 Chr.4: 85931379-85931379 on Build GRCh38
rs79743688 Chr.4: 85931055-85931055 on Build GRCh38
rs899673146 Chr.4: 85931325-85931325 on Build GRCh38
rs113093434 Chr.4: 85931324-85931324 on Build GRCh38
rs754790734 Chr.4: 85931090-85931091 on Build GRCh38
rs138330074 Chr.4: 85931259-85931259 on Build GRCh38
rs778083136 Chr.4: 85930964-85930964 on Build GRCh38
rs1041473869 Chr.4: 85931237-85931237 on Build GRCh38
rs1000201109 Chr.4: 85931281-85931281 on Build GRCh38
rs758623399 Chr.4: 85931091-85931091 on Build GRCh38
rs1027366663 Chr.4: 85930958-85930958 on Build GRCh38
rs760468811 Chr.4: 85931069-85931069 on Build GRCh38
rs776695655 Chr.4: 85931149-85931149 on Build GRCh38
rs1047694784 Chr.4: 85931131-85931131 on Build GRCh38
rs527475078 Chr.4: 85931245-85931245 on Build GRCh38
rs751097836 Chr.4: 85930942-85930942 on Build GRCh38
rs776657293 Chr.4: 85931256-85931256 on Build GRCh38
rs768646933 Chr.4: 85931036-85931036 on Build GRCh38
rs879876325 Chr.4: 85931295-85931295 on Build GRCh38
rs150444643 Chr.4: 85931083-85931083 on Build GRCh38
rs62305513 Chr.4: 85931355-85931355 on Build GRCh38
rs914217162 Chr.4: 85931176-85931176 on Build GRCh38
rs377522479 Chr.4: 85931021-85931021 on Build GRCh38
rs776930756 Chr.4: 85930978-85930978 on Build GRCh38
rs779458518 Chr.4: 85930962-85930962 on Build GRCh38
rs750455147 Chr.4: 85930952-85930952 on Build GRCh38
rs770873001 Chr.4: 85930973-85930973 on Build GRCh38
rs140387210 Chr.4: 85931037-85931037 on Build GRCh38
rs970857601 Chr.4: 85931029-85931029 on Build GRCh38
rs560477517 Chr.4: 85931236-85931236 on Build GRCh38
rs766114418 Chr.4: 85931071-85931071 on Build GRCh38
rs942087101 Chr.4: 85931170-85931170 on Build GRCh38
rs61758879 Chr.4: 85930944-85930944 on Build GRCh38
rs907874390 Chr.4: 85931140-85931140 on Build GRCh38
rs746134846 Chr.4: 85930981-85930981 on Build GRCh38
rs762682623 Chr.4: 85931030-85931030 on Build GRCh38
rs532565956 Chr.4: 85931122-85931122 on Build GRCh38
rs145811610 Chr.4: 85931383-85931383 on Build GRCh38
rs779111587 Chr.4: 85931292-85931292 on Build GRCh38
rs761408648 Chr.4: 85931038-85931038 on Build GRCh38
rs151225111 Chr.4: 85930963-85930963 on Build GRCh38
rs918026294 Chr.4: 85931022-85931022 on Build GRCh38
rs571209504 Chr.4: 85931326-85931326 on Build GRCh38
rs1054052053 Chr.4: 85931093-85931093 on Build GRCh38

View Full Product Details