Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 128877347-128877836 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCP
Entrez Gene ID
375616
Gene Name
kielin/chordin-like protein
Gene Aliases
CRIM2, KCP1, NET67
UniGene
Hs.371746
Gene Chromosome Location
Chr.7: 128870645-128910719 on Build GRCh38
Transcripts
NM_001135914.1 XM_017012184.1 XM_017012185.1 XM_017012186.1 XM_017012187.1 XM_017012188.1 XM_017012189.1 XM_017012190.1 XM_017012191.1 XM_017012192.1

SNP Detail

SNP ID Chromosome Location
rs566344673 Chr.7: 128877449-128877449 on Build GRCh38
rs1039886502 Chr.7: 128877774-128877774 on Build GRCh38
rs867676266 Chr.7: 128877496-128877496 on Build GRCh38
rs977752094 Chr.7: 128877649-128877649 on Build GRCh38
rs547823300 Chr.7: 128877428-128877428 on Build GRCh38
rs375356208 Chr.7: 128877734-128877734 on Build GRCh38
rs951975548 Chr.7: 128877488-128877488 on Build GRCh38
rs992357912 Chr.7: 128877503-128877503 on Build GRCh38
rs548817334 Chr.7: 128877491-128877491 on Build GRCh38
rs556588962 Chr.7: 128877398-128877398 on Build GRCh38
rs773503937 Chr.7: 128877455-128877455 on Build GRCh38
rs916373105 Chr.7: 128877712-128877712 on Build GRCh38
rs899901389 Chr.7: 128877621-128877621 on Build GRCh38
rs921327505 Chr.7: 128877696-128877696 on Build GRCh38
rs960847604 Chr.7: 128877704-128877704 on Build GRCh38
rs189646984 Chr.7: 128877421-128877421 on Build GRCh38
rs760204461 Chr.7: 128877570-128877570 on Build GRCh38
rs1048526164 Chr.7: 128877706-128877706 on Build GRCh38
rs537190903 Chr.7: 128877397-128877397 on Build GRCh38
rs80104976 Chr.7: 128877371-128877371 on Build GRCh38
rs924919914 Chr.7: 128877542-128877542 on Build GRCh38
rs570236887 Chr.7: 128877492-128877492 on Build GRCh38
rs372362406 Chr.7: 128877716-128877716 on Build GRCh38
rs745521883 Chr.7: 128877459-128877459 on Build GRCh38
rs944232904 Chr.7: 128877769-128877770 on Build GRCh38
rs777301981 Chr.7: 128877809-128877809 on Build GRCh38
rs763484619 Chr.7: 128877501-128877501 on Build GRCh38
rs768239836 Chr.7: 128877723-128877723 on Build GRCh38
rs991355895 Chr.7: 128877416-128877416 on Build GRCh38
rs755971268 Chr.7: 128877555-128877555 on Build GRCh38
rs532126274 Chr.7: 128877422-128877422 on Build GRCh38
rs866881766 Chr.7: 128877619-128877619 on Build GRCh38
rs774995709 Chr.7: 128877468-128877468 on Build GRCh38
rs542483498 Chr.7: 128877735-128877735 on Build GRCh38
rs1048566455 Chr.7: 128877523-128877523 on Build GRCh38
rs773941574 Chr.7: 128877725-128877725 on Build GRCh38
rs374120437 Chr.7: 128877453-128877453 on Build GRCh38
rs761847040 Chr.7: 128877730-128877730 on Build GRCh38
rs899974745 Chr.7: 128877792-128877792 on Build GRCh38
rs968961863 Chr.7: 128877752-128877752 on Build GRCh38
rs374742200 Chr.7: 128877513-128877513 on Build GRCh38
rs1874333 Chr.7: 128877439-128877439 on Build GRCh38
rs981195608 Chr.7: 128877379-128877379 on Build GRCh38
rs368508165 Chr.7: 128877636-128877636 on Build GRCh38
rs891332196 Chr.7: 128877582-128877582 on Build GRCh38
rs951350744 Chr.7: 128877646-128877646 on Build GRCh38
rs754112791 Chr.7: 128877810-128877810 on Build GRCh38
rs747594062 Chr.7: 128877428-128877428 on Build GRCh38
rs575790424 Chr.7: 128877761-128877761 on Build GRCh38
rs368547608 Chr.7: 128877528-128877528 on Build GRCh38
rs773891258 Chr.7: 128877423-128877423 on Build GRCh38
rs947743057 Chr.7: 128877529-128877529 on Build GRCh38
rs1041052120 Chr.7: 128877689-128877689 on Build GRCh38
rs914410256 Chr.7: 128877635-128877635 on Build GRCh38
rs578110016 Chr.7: 128877658-128877658 on Build GRCh38
rs774465179 Chr.7: 128877509-128877509 on Build GRCh38
rs115155038 Chr.7: 128877466-128877466 on Build GRCh38
rs995967089 Chr.7: 128877627-128877627 on Build GRCh38
rs747429865 Chr.7: 128877691-128877691 on Build GRCh38
rs1004884227 Chr.7: 128877541-128877541 on Build GRCh38
rs958473901 Chr.7: 128877607-128877607 on Build GRCh38
rs1048456071 Chr.7: 128877815-128877815 on Build GRCh38
rs778609481 Chr.7: 128877389-128877389 on Build GRCh38
rs572318375 Chr.7: 128877657-128877657 on Build GRCh38
rs866511239 Chr.7: 128877661-128877661 on Build GRCh38
rs371147510 Chr.7: 128877578-128877578 on Build GRCh38
rs554345291 Chr.7: 128877747-128877747 on Build GRCh38
rs116177150 Chr.7: 128877814-128877814 on Build GRCh38
rs899959089 Chr.7: 128877506-128877506 on Build GRCh38
rs748855812 Chr.7: 128877715-128877715 on Build GRCh38
rs867308081 Chr.7: 128877729-128877729 on Build GRCh38
rs138150784 Chr.7: 128877605-128877605 on Build GRCh38
rs779798989 Chr.7: 128877714-128877714 on Build GRCh38
rs375312754 Chr.7: 128877651-128877651 on Build GRCh38
rs1021426580 Chr.7: 128877750-128877750 on Build GRCh38
rs750178478 Chr.7: 128877537-128877537 on Build GRCh38
rs771179921 Chr.7: 128877705-128877705 on Build GRCh38
rs755115129 Chr.7: 128877604-128877604 on Build GRCh38
rs142343595 Chr.7: 128877620-128877620 on Build GRCh38
rs766671561 Chr.7: 128877577-128877577 on Build GRCh38
rs370254255 Chr.7: 128877502-128877502 on Build GRCh38
rs766047357 Chr.7: 128877790-128877790 on Build GRCh38
rs760557906 Chr.7: 128877766-128877766 on Build GRCh38
rs907723158 Chr.7: 128877499-128877499 on Build GRCh38
rs1004429460 Chr.7: 128877719-128877719 on Build GRCh38

View Full Product Details