Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 842028-842562 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMRT1
Entrez Gene ID
1761
Gene Name
doublesex and mab-3 related transcription factor 1
Gene Aliases
CT154, DMT1
UniGene
Hs.98586
Gene Chromosome Location
Chr.9: 841647-969090 on Build GRCh38
Transcripts
NM_021951.2 XM_006716732.1 XM_011517770.1 XM_011517771.1 XM_011517772.1 XM_017014374.1 XM_017014375.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6377507 c.194C>T p.A65V Chr.9: 842032-842032 on Build GRCh38
6375828 c.284G>A p.C95Y Chr.9: 842122-842122 on Build GRCh38
5711350 c.233G>A p.R78H Chr.9: 842071-842071 on Build GRCh38
6375334 c.250T>C p.Y84H Chr.9: 842088-842088 on Build GRCh38
4678133 c.288G>A p.M96I Chr.9: 842126-842126 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747369731 Chr.9: 842165-842165 on Build GRCh38
rs755079068 Chr.9: 842090-842090 on Build GRCh38
rs533279401 Chr.9: 842461-842461 on Build GRCh38
rs948716176 Chr.9: 842320-842320 on Build GRCh38
rs534106583 Chr.9: 842317-842317 on Build GRCh38
rs201947617 Chr.9: 842146-842146 on Build GRCh38
rs1056297742 Chr.9: 842342-842342 on Build GRCh38
rs976067146 Chr.9: 842392-842392 on Build GRCh38
rs1028424156 Chr.9: 842393-842393 on Build GRCh38
rs781310087 Chr.9: 842236-842237 on Build GRCh38
rs750124106 Chr.9: 842433-842433 on Build GRCh38
rs974317911 Chr.9: 842298-842298 on Build GRCh38
rs1007959097 Chr.9: 842396-842396 on Build GRCh38
rs376518776 Chr.9: 842051-842051 on Build GRCh38
rs996486717 Chr.9: 842380-842380 on Build GRCh38
rs928714863 Chr.9: 842327-842327 on Build GRCh38
rs1014868245 Chr.9: 842243-842243 on Build GRCh38
rs370607838 Chr.9: 842076-842076 on Build GRCh38
rs914303157 Chr.9: 842414-842414 on Build GRCh38
rs757201575 Chr.9: 842474-842474 on Build GRCh38
rs1004337093 Chr.9: 842531-842531 on Build GRCh38
rs368022469 Chr.9: 842358-842358 on Build GRCh38
rs780064237 Chr.9: 842230-842231 on Build GRCh38
rs569156714 Chr.9: 842246-842246 on Build GRCh38
rs945826176 Chr.9: 842428-842428 on Build GRCh38
rs758517384 Chr.9: 842308-842308 on Build GRCh38
rs1034550404 Chr.9: 842108-842108 on Build GRCh38
rs945733040 Chr.9: 842306-842309 on Build GRCh38
rs966351098 Chr.9: 842391-842391 on Build GRCh38
rs1028038971 Chr.9: 842087-842087 on Build GRCh38
rs989834261 Chr.9: 842407-842407 on Build GRCh38
rs753665192 Chr.9: 842398-842398 on Build GRCh38
rs541946356 Chr.9: 842355-842355 on Build GRCh38
rs1004198204 Chr.9: 842381-842381 on Build GRCh38
rs148624819 Chr.9: 842395-842395 on Build GRCh38
rs1056057041 Chr.9: 842480-842480 on Build GRCh38
rs921824580 Chr.9: 842275-842275 on Build GRCh38
rs746172391 Chr.9: 842385-842385 on Build GRCh38
rs56130013 Chr.9: 842316-842316 on Build GRCh38
rs866146124 Chr.9: 842107-842107 on Build GRCh38
rs756302236 Chr.9: 842319-842319 on Build GRCh38
rs1019656101 Chr.9: 842384-842384 on Build GRCh38
rs986904108 Chr.9: 842117-842117 on Build GRCh38
rs781655856 Chr.9: 842200-842200 on Build GRCh38
rs980100485 Chr.9: 842495-842495 on Build GRCh38
rs548919882 Chr.9: 842239-842239 on Build GRCh38
rs373843416 Chr.9: 842321-842321 on Build GRCh38
rs767338325 Chr.9: 842255-842255 on Build GRCh38
rs778098130 Chr.9: 842153-842153 on Build GRCh38
rs756669929 Chr.9: 842186-842187 on Build GRCh38
rs775835389 Chr.9: 842318-842318 on Build GRCh38
rs770230827 Chr.9: 842209-842209 on Build GRCh38
rs368228276 Chr.9: 842132-842132 on Build GRCh38
rs995961933 Chr.9: 842339-842339 on Build GRCh38
rs746393828 Chr.9: 842201-842201 on Build GRCh38
rs959034753 Chr.9: 842150-842150 on Build GRCh38
rs796831808 Chr.9: 842317-842317 on Build GRCh38
rs989862363 Chr.9: 842295-842300 on Build GRCh38
rs938730037 Chr.9: 842535-842535 on Build GRCh38
rs764456336 Chr.9: 842316-842316 on Build GRCh38
rs903775848 Chr.9: 842379-842379 on Build GRCh38
rs938893042 Chr.9: 842467-842467 on Build GRCh38
rs752764799 Chr.9: 842093-842093 on Build GRCh38
rs761639194 Chr.9: 842048-842048 on Build GRCh38
rs544228278 Chr.9: 842442-842442 on Build GRCh38
rs549953533 Chr.9: 842329-842329 on Build GRCh38
rs766368473 Chr.9: 842286-842286 on Build GRCh38
rs894607519 Chr.9: 842483-842483 on Build GRCh38
rs558154854 Chr.9: 842265-842265 on Build GRCh38
rs868559576 Chr.9: 842147-842147 on Build GRCh38
rs928656859 Chr.9: 842506-842506 on Build GRCh38
rs535563425 Chr.9: 842226-842226 on Build GRCh38
rs566591888 Chr.9: 842186-842186 on Build GRCh38
rs752852716 Chr.9: 842295-842295 on Build GRCh38
rs1026947032 Chr.9: 842208-842208 on Build GRCh38
rs578219773 Chr.9: 842314-842314 on Build GRCh38
rs961561777 Chr.9: 842453-842453 on Build GRCh38
rs564203514 Chr.9: 842459-842459 on Build GRCh38
rs538280651 Chr.9: 842260-842260 on Build GRCh38
rs767227773 Chr.9: 842229-842230 on Build GRCh38
rs931488473 Chr.9: 842484-842484 on Build GRCh38
rs748757340 Chr.9: 842231-842231 on Build GRCh38
rs12340514 Chr.9: 842120-842120 on Build GRCh38
rs752939956 Chr.9: 842285-842285 on Build GRCh38
rs796589183 Chr.9: 842229-842230 on Build GRCh38
rs931723275 Chr.9: 842291-842291 on Build GRCh38
rs1043681952 Chr.9: 842362-842362 on Build GRCh38
rs1027086111 Chr.9: 842344-842344 on Build GRCh38
rs751805873 Chr.9: 842323-842323 on Build GRCh38
rs761570527 Chr.9: 842254-842254 on Build GRCh38
rs370876706 Chr.9: 842494-842494 on Build GRCh38
rs772814165 Chr.9: 842256-842256 on Build GRCh38
rs746133675 Chr.9: 842304-842304 on Build GRCh38
rs765410506 Chr.9: 842091-842091 on Build GRCh38

View Full Product Details