Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 114123791-114124304 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC186
Entrez Gene ID
55088
Gene Name
coiled-coil domain containing 186
Gene Aliases
C10orf118
UniGene
Hs.159066
Gene Chromosome Location
Chr.10: 114120862-114174312 on Build GRCh38
Transcripts
NM_001321829.1 NM_018017.3 NR_135815.1 XM_011539915.2

SNP Detail

SNP ID Chromosome Location
rs116059137 Chr.10: 114124195-114124195 on Build GRCh38
rs41292642 Chr.10: 114123878-114123878 on Build GRCh38
rs1030716651 Chr.10: 114123916-114123916 on Build GRCh38
rs769822379 Chr.10: 114123824-114123824 on Build GRCh38
rs927307704 Chr.10: 114123978-114123978 on Build GRCh38
rs972952349 Chr.10: 114123945-114123945 on Build GRCh38
rs569005367 Chr.10: 114123849-114123849 on Build GRCh38
rs41293044 Chr.10: 114123958-114123958 on Build GRCh38
rs537506482 Chr.10: 114123874-114123874 on Build GRCh38
rs1036599196 Chr.10: 114124047-114124047 on Build GRCh38
rs536775575 Chr.10: 114123959-114123959 on Build GRCh38
rs116731523 Chr.10: 114123832-114123832 on Build GRCh38
rs1028205019 Chr.10: 114124051-114124051 on Build GRCh38
rs891853868 Chr.10: 114124024-114124024 on Build GRCh38
rs750008653 Chr.10: 114124198-114124198 on Build GRCh38
rs529185242 Chr.10: 114123829-114123829 on Build GRCh38
rs911759261 Chr.10: 114123998-114123998 on Build GRCh38
rs983874012 Chr.10: 114123839-114123839 on Build GRCh38
rs953021935 Chr.10: 114123980-114123980 on Build GRCh38
rs113478153 Chr.10: 114124147-114124147 on Build GRCh38
rs950371193 Chr.10: 114123972-114123972 on Build GRCh38
rs980326877 Chr.10: 114124202-114124202 on Build GRCh38
rs953709194 Chr.10: 114124068-114124068 on Build GRCh38
rs561625756 Chr.10: 114123826-114123826 on Build GRCh38
rs558205499 Chr.10: 114123981-114123981 on Build GRCh38
rs553434168 Chr.10: 114124100-114124100 on Build GRCh38
rs970936251 Chr.10: 114123907-114123907 on Build GRCh38
rs888722033 Chr.10: 114123833-114123833 on Build GRCh38
rs534993252 Chr.10: 114124191-114124191 on Build GRCh38
rs900992323 Chr.10: 114124041-114124041 on Build GRCh38
rs1044622768 Chr.10: 114123977-114123977 on Build GRCh38
rs761392386 Chr.10: 114124126-114124126 on Build GRCh38
rs879210450 Chr.10: 114124224-114124224 on Build GRCh38
rs1010725221 Chr.10: 114124032-114124032 on Build GRCh38
rs574963259 Chr.10: 114124181-114124181 on Build GRCh38
rs1054488665 Chr.10: 114124016-114124016 on Build GRCh38
rs1004080018 Chr.10: 114123915-114123915 on Build GRCh38
rs535816092 Chr.10: 114124135-114124135 on Build GRCh38
rs575755846 Chr.10: 114124272-114124272 on Build GRCh38
rs961176072 Chr.10: 114124144-114124144 on Build GRCh38
rs935965044 Chr.10: 114123987-114123987 on Build GRCh38
rs927526557 Chr.10: 114124251-114124251 on Build GRCh38
rs1012485365 Chr.10: 114123848-114123848 on Build GRCh38
rs552742047 Chr.10: 114124053-114124053 on Build GRCh38
rs936017347 Chr.10: 114124274-114124274 on Build GRCh38
rs116820646 Chr.10: 114124040-114124040 on Build GRCh38
rs956447694 Chr.10: 114123936-114123936 on Build GRCh38
rs1005190844 Chr.10: 114124117-114124117 on Build GRCh38
rs41293046 Chr.10: 114124131-114124131 on Build GRCh38
rs917460923 Chr.10: 114124175-114124175 on Build GRCh38
rs375609520 Chr.10: 114124159-114124159 on Build GRCh38

View Full Product Details