Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 43647197-43647700 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CATSPER2
Entrez Gene ID
117155
Gene Name
cation channel sperm associated 2
Gene Aliases
-
UniGene
Hs.662284
Gene Chromosome Location
Chr.15: 43630562-43648845 on Build GRCh38
Transcripts
NM_001282309.1 NM_001282310.1 NM_054020.3 NM_172095.2 NR_110319.1 XM_017021904.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3999537 c.169G>A p.V57I Chr.15: 43647444-43647444 on Build GRCh38
6620762 c.168T>C p.L56L Chr.15: 43647445-43647445 on Build GRCh38
1199699 c.162G>T p.K54N Chr.15: 43647451-43647451 on Build GRCh38
6142097 c.195G>T p.V65V Chr.15: 43647418-43647418 on Build GRCh38
6620760 c.223G>A p.E75K Chr.15: 43647390-43647390 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755568758 Chr.15: 43647398-43647398 on Build GRCh38
rs536026719 Chr.15: 43647397-43647397 on Build GRCh38
rs544833343 Chr.15: 43647572-43647572 on Build GRCh38
rs574420233 Chr.15: 43647643-43647643 on Build GRCh38
rs750338105 Chr.15: 43647376-43647376 on Build GRCh38
rs755885533 Chr.15: 43647393-43647393 on Build GRCh38
rs771282534 Chr.15: 43647361-43647361 on Build GRCh38
rs749760832 Chr.15: 43647268-43647269 on Build GRCh38
rs746402010 Chr.15: 43647505-43647505 on Build GRCh38
rs968777909 Chr.15: 43647563-43647563 on Build GRCh38
rs140622661 Chr.15: 43647615-43647615 on Build GRCh38
rs1056862035 Chr.15: 43647650-43647650 on Build GRCh38
rs139742996 Chr.15: 43647419-43647419 on Build GRCh38
rs917064983 Chr.15: 43647669-43647669 on Build GRCh38
rs146592201 Chr.15: 43647379-43647379 on Build GRCh38
rs772518049 Chr.15: 43647512-43647512 on Build GRCh38
rs888140644 Chr.15: 43647431-43647431 on Build GRCh38
rs771587549 Chr.15: 43647285-43647285 on Build GRCh38
rs769296083 Chr.15: 43647339-43647339 on Build GRCh38
rs141594294 Chr.15: 43647346-43647346 on Build GRCh38
rs748630499 Chr.15: 43647243-43647243 on Build GRCh38
rs754652195 Chr.15: 43647395-43647395 on Build GRCh38
rs937431619 Chr.15: 43647612-43647612 on Build GRCh38
rs755799770 Chr.15: 43647482-43647482 on Build GRCh38
rs184419046 Chr.15: 43647607-43647607 on Build GRCh38
rs989095435 Chr.15: 43647554-43647554 on Build GRCh38
rs754634692 Chr.15: 43647329-43647329 on Build GRCh38
rs770341410 Chr.15: 43647248-43647248 on Build GRCh38
rs745336079 Chr.15: 43647371-43647371 on Build GRCh38
rs747202886 Chr.15: 43647518-43647518 on Build GRCh38
rs151235424 Chr.15: 43647579-43647579 on Build GRCh38
rs750391520 Chr.15: 43647308-43647308 on Build GRCh38
rs766292661 Chr.15: 43647454-43647454 on Build GRCh38
rs947435859 Chr.15: 43647222-43647222 on Build GRCh38
rs751517231 Chr.15: 43647458-43647458 on Build GRCh38
rs759785551 Chr.15: 43647373-43647373 on Build GRCh38
rs144999733 Chr.15: 43647488-43647488 on Build GRCh38
rs772571306 Chr.15: 43647423-43647423 on Build GRCh38
rs148292559 Chr.15: 43647327-43647327 on Build GRCh38
rs552779558 Chr.15: 43647430-43647430 on Build GRCh38
rs771771311 Chr.15: 43647413-43647414 on Build GRCh38
rs779736058 Chr.15: 43647320-43647320 on Build GRCh38
rs777247755 Chr.15: 43647483-43647483 on Build GRCh38
rs768817228 Chr.15: 43647442-43647442 on Build GRCh38
rs937210880 Chr.15: 43647324-43647324 on Build GRCh38
rs774622123 Chr.15: 43647364-43647364 on Build GRCh38
rs762933365 Chr.15: 43647449-43647449 on Build GRCh38
rs773559114 Chr.15: 43647250-43647250 on Build GRCh38
rs779223925 Chr.15: 43647490-43647490 on Build GRCh38
rs780754672 Chr.15: 43647396-43647396 on Build GRCh38
rs557984480 Chr.15: 43647567-43647567 on Build GRCh38
rs767884013 Chr.15: 43647663-43647663 on Build GRCh38
rs779611725 Chr.15: 43647334-43647337 on Build GRCh38
rs146109978 Chr.15: 43647461-43647461 on Build GRCh38
rs8042868 Chr.15: 43647444-43647444 on Build GRCh38
rs771394367 Chr.15: 43647417-43647417 on Build GRCh38
rs913206711 Chr.15: 43647556-43647556 on Build GRCh38
rs763234575 Chr.15: 43647253-43647253 on Build GRCh38
rs775191853 Chr.15: 43647417-43647418 on Build GRCh38
rs761000751 Chr.15: 43647441-43647441 on Build GRCh38
rs753350226 Chr.15: 43647487-43647487 on Build GRCh38
rs868607564 Chr.15: 43647282-43647282 on Build GRCh38
rs777292650 Chr.15: 43647409-43647409 on Build GRCh38
rs766663108 Chr.15: 43647261-43647261 on Build GRCh38
rs749775185 Chr.15: 43647416-43647416 on Build GRCh38
rs752877112 Chr.15: 43647283-43647283 on Build GRCh38
rs374455420 Chr.15: 43647277-43647277 on Build GRCh38
rs747722095 Chr.15: 43647336-43647336 on Build GRCh38
rs759713000 Chr.15: 43647272-43647272 on Build GRCh38
rs772244535 Chr.15: 43647374-43647374 on Build GRCh38
rs778002843 Chr.15: 43647261-43647262 on Build GRCh38
rs774580377 Chr.15: 43647263-43647263 on Build GRCh38
rs773648730 Chr.15: 43647345-43647345 on Build GRCh38
rs536658451 Chr.15: 43647307-43647307 on Build GRCh38
rs758155821 Chr.15: 43647310-43647310 on Build GRCh38
rs759192725 Chr.15: 43647459-43647459 on Build GRCh38
rs764284645 Chr.15: 43647375-43647375 on Build GRCh38
rs368049818 Chr.15: 43647513-43647513 on Build GRCh38
rs780665141 Chr.15: 43647334-43647334 on Build GRCh38
rs141294344 Chr.15: 43647385-43647385 on Build GRCh38
rs752302624 Chr.15: 43647463-43647463 on Build GRCh38

View Full Product Details