Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 25961753-25962248 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAFAH2
Entrez Gene ID
5051
Gene Name
platelet activating factor acetylhydrolase 2
Gene Aliases
HSD-PLA2
UniGene
Hs.477083
Gene Chromosome Location
Chr.1: 25959764-25998164 on Build GRCh38
Transcripts
NM_000437.3 XM_006710669.3 XM_006710670.3 XM_011541528.2 XM_011541529.2 XM_017001390.1 XM_017001391.1 XM_017001392.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4165138 c.1179G>T p.*393Y Chr.1: 25961989-25961989 on Build GRCh38
6736922 c.1159delC p.H387fs*>6 Chr.1: 25962009-25962009 on Build GRCh38
414510 c.1165C>G p.L389V Chr.1: 25962003-25962003 on Build GRCh38
4460216 c.1157C>T p.P386L Chr.1: 25962011-25962011 on Build GRCh38
6239150 c.1170C>A p.S390S Chr.1: 25961998-25961998 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780170272 Chr.1: 25962031-25962031 on Build GRCh38
rs773047561 Chr.1: 25962041-25962041 on Build GRCh38
rs751094564 Chr.1: 25961956-25961956 on Build GRCh38
rs767017184 Chr.1: 25962064-25962064 on Build GRCh38
rs770893083 Chr.1: 25962050-25962050 on Build GRCh38
rs1020116659 Chr.1: 25962153-25962153 on Build GRCh38
rs766782302 Chr.1: 25961933-25961933 on Build GRCh38
rs866180754 Chr.1: 25962022-25962022 on Build GRCh38
rs762729760 Chr.1: 25962045-25962045 on Build GRCh38
rs201672869 Chr.1: 25962026-25962026 on Build GRCh38
rs202124958 Chr.1: 25961953-25961953 on Build GRCh38
rs371059181 Chr.1: 25961831-25961831 on Build GRCh38
rs894996775 Chr.1: 25961913-25961913 on Build GRCh38
rs769587744 Chr.1: 25962038-25962038 on Build GRCh38
rs573554051 Chr.1: 25961895-25961895 on Build GRCh38
rs750894945 Chr.1: 25962100-25962100 on Build GRCh38
rs139778582 Chr.1: 25961844-25961844 on Build GRCh38
rs759051907 Chr.1: 25961958-25961958 on Build GRCh38
rs746640904 Chr.1: 25962092-25962093 on Build GRCh38
rs965526053 Chr.1: 25962181-25962181 on Build GRCh38
rs780123004 Chr.1: 25962111-25962111 on Build GRCh38
rs757245785 Chr.1: 25962057-25962057 on Build GRCh38
rs970989533 Chr.1: 25962220-25962220 on Build GRCh38
rs149392992 Chr.1: 25962107-25962107 on Build GRCh38
rs754412577 Chr.1: 25962003-25962003 on Build GRCh38
rs763828898 Chr.1: 25962074-25962074 on Build GRCh38
rs200452453 Chr.1: 25962032-25962032 on Build GRCh38
rs1012140498 Chr.1: 25962180-25962180 on Build GRCh38
rs779944624 Chr.1: 25962141-25962141 on Build GRCh38
rs755258582 Chr.1: 25962129-25962129 on Build GRCh38
rs367684862 Chr.1: 25962122-25962122 on Build GRCh38
rs1011972330 Chr.1: 25961954-25961954 on Build GRCh38
rs772171414 Chr.1: 25962029-25962029 on Build GRCh38
rs778220844 Chr.1: 25962000-25962000 on Build GRCh38
rs1023486045 Chr.1: 25962183-25962183 on Build GRCh38
rs35864830 Chr.1: 25962014-25962015 on Build GRCh38
rs757666574 Chr.1: 25962082-25962082 on Build GRCh38
rs775027716 Chr.1: 25962093-25962093 on Build GRCh38
rs375214843 Chr.1: 25961868-25961868 on Build GRCh38
rs758986960 Chr.1: 25962061-25962061 on Build GRCh38
rs199672043 Chr.1: 25962028-25962028 on Build GRCh38
rs368000628 Chr.1: 25962006-25962006 on Build GRCh38
rs760348294 Chr.1: 25962069-25962069 on Build GRCh38
rs754322407 Chr.1: 25962081-25962081 on Build GRCh38
rs546446095 Chr.1: 25962152-25962152 on Build GRCh38
rs762817871 Chr.1: 25961942-25961942 on Build GRCh38
rs538495657 Chr.1: 25961840-25961840 on Build GRCh38
rs577510074 Chr.1: 25962150-25962150 on Build GRCh38
rs745340726 Chr.1: 25962015-25962016 on Build GRCh38
rs1032297175 Chr.1: 25961914-25961914 on Build GRCh38
rs745975180 Chr.1: 25962016-25962016 on Build GRCh38
rs767121782 Chr.1: 25961959-25961959 on Build GRCh38
rs1032970930 Chr.1: 25962145-25962145 on Build GRCh38
rs769537562 Chr.1: 25961939-25961939 on Build GRCh38
rs778258088 Chr.1: 25962008-25962009 on Build GRCh38
rs1000238911 Chr.1: 25962121-25962121 on Build GRCh38
rs557233953 Chr.1: 25962087-25962087 on Build GRCh38
rs765712326 Chr.1: 25962086-25962086 on Build GRCh38
rs1037320648 Chr.1: 25962013-25962013 on Build GRCh38
rs181210727 Chr.1: 25961992-25961992 on Build GRCh38
rs752301352 Chr.1: 25962068-25962068 on Build GRCh38
rs754988391 Chr.1: 25961928-25961928 on Build GRCh38
rs200871432 Chr.1: 25962008-25962008 on Build GRCh38
rs3738455 Chr.1: 25961798-25961798 on Build GRCh38
rs375563101 Chr.1: 25961983-25961983 on Build GRCh38
rs773137480 Chr.1: 25961941-25961941 on Build GRCh38
rs936600257 Chr.1: 25961824-25961824 on Build GRCh38
rs187073312 Chr.1: 25962182-25962182 on Build GRCh38
rs774319261 Chr.1: 25962055-25962055 on Build GRCh38

View Full Product Details