Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 27355662-27356161 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP3K6
Entrez Gene ID
9064
Gene Name
mitogen-activated protein kinase kinase kinase 6
Gene Aliases
ASK2, MAPKKK6, MEKK6
UniGene
Hs.194694
Gene Chromosome Location
Chr.1: 27355179-27368037 on Build GRCh38
Transcripts
NM_001297609.1 NM_004672.4 XM_017002771.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
298470 c.3748A>T p.I1250F Chr.1: 27355685-27355685 on Build GRCh38
4751686 c.3684A>C p.Q1228H Chr.1: 27356029-27356029 on Build GRCh38
6363260 c.3782G>T p.R1261L Chr.1: 27355675-27355675 on Build GRCh38
7376285 c.3650C>T p.T1217M Chr.1: 27356087-27356087 on Build GRCh38
4872259 c.3754G>A p.A1252T Chr.1: 27355703-27355703 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs554613210 Chr.1: 27355895-27355895 on Build GRCh38
rs766148863 Chr.1: 27355706-27355706 on Build GRCh38
rs71638522 Chr.1: 27355722-27355722 on Build GRCh38
rs940459151 Chr.1: 27355922-27355922 on Build GRCh38
rs759303554 Chr.1: 27355979-27355979 on Build GRCh38
rs559281366 Chr.1: 27355991-27355991 on Build GRCh38
rs754225050 Chr.1: 27355735-27355735 on Build GRCh38
rs145180393 Chr.1: 27355844-27355844 on Build GRCh38
rs17162549 Chr.1: 27356039-27356039 on Build GRCh38
rs751036669 Chr.1: 27355707-27355707 on Build GRCh38
rs981810995 Chr.1: 27355896-27355896 on Build GRCh38
rs375091053 Chr.1: 27356019-27356019 on Build GRCh38
rs36035221 Chr.1: 27355949-27355950 on Build GRCh38
rs991153499 Chr.1: 27355817-27355817 on Build GRCh38
rs368729404 Chr.1: 27355995-27355995 on Build GRCh38
rs770886299 Chr.1: 27355791-27355791 on Build GRCh38
rs747106322 Chr.1: 27356030-27356030 on Build GRCh38
rs76570472 Chr.1: 27355710-27355710 on Build GRCh38
rs773144014 Chr.1: 27355688-27355688 on Build GRCh38
rs1051149593 Chr.1: 27356040-27356040 on Build GRCh38
rs767117230 Chr.1: 27355718-27355718 on Build GRCh38
rs756071813 Chr.1: 27356037-27356037 on Build GRCh38
rs907659533 Chr.1: 27355907-27355907 on Build GRCh38
rs184469734 Chr.1: 27356118-27356118 on Build GRCh38
rs1041809111 Chr.1: 27356117-27356117 on Build GRCh38
rs762373403 Chr.1: 27356088-27356088 on Build GRCh38
rs779406137 Chr.1: 27355775-27355775 on Build GRCh38
rs778055953 Chr.1: 27355734-27355734 on Build GRCh38
rs370126739 Chr.1: 27355765-27355765 on Build GRCh38
rs757432362 Chr.1: 27355746-27355746 on Build GRCh38
rs773090772 Chr.1: 27355781-27355781 on Build GRCh38
rs749122942 Chr.1: 27355790-27355790 on Build GRCh38
rs775081398 Chr.1: 27355989-27355989 on Build GRCh38
rs370697919 Chr.1: 27355695-27355695 on Build GRCh38
rs757110360 Chr.1: 27356115-27356115 on Build GRCh38
rs749283789 Chr.1: 27356046-27356046 on Build GRCh38
rs908874026 Chr.1: 27355805-27355805 on Build GRCh38
rs771781251 Chr.1: 27356063-27356063 on Build GRCh38
rs957942968 Chr.1: 27355799-27355799 on Build GRCh38
rs575195202 Chr.1: 27355825-27355825 on Build GRCh38
rs769707001 Chr.1: 27355685-27355685 on Build GRCh38
rs150923701 Chr.1: 27356082-27356082 on Build GRCh38
rs778003671 Chr.1: 27356129-27356129 on Build GRCh38
rs763814952 Chr.1: 27355782-27355784 on Build GRCh38
rs754384062 Chr.1: 27356002-27356002 on Build GRCh38
rs191299880 Chr.1: 27355898-27355898 on Build GRCh38
rs558947768 Chr.1: 27355773-27355773 on Build GRCh38
rs774824294 Chr.1: 27356071-27356071 on Build GRCh38
rs546863373 Chr.1: 27355684-27355684 on Build GRCh38
rs369311596 Chr.1: 27355948-27355949 on Build GRCh38
rs753581724 Chr.1: 27356065-27356065 on Build GRCh38
rs750811568 Chr.1: 27356122-27356122 on Build GRCh38
rs779938252 Chr.1: 27356027-27356027 on Build GRCh38
rs773866514 Chr.1: 27355976-27355976 on Build GRCh38
rs12569127 Chr.1: 27355990-27355990 on Build GRCh38
rs758394991 Chr.1: 27355766-27355766 on Build GRCh38
rs992364585 Chr.1: 27356064-27356064 on Build GRCh38
rs755608417 Chr.1: 27355761-27355761 on Build GRCh38
rs745456010 Chr.1: 27356061-27356061 on Build GRCh38
rs372060699 Chr.1: 27356018-27356018 on Build GRCh38
rs59974972 Chr.1: 27355952-27355953 on Build GRCh38
rs371594483 Chr.1: 27356128-27356128 on Build GRCh38
rs777110745 Chr.1: 27356087-27356087 on Build GRCh38
rs148477596 Chr.1: 27356058-27356058 on Build GRCh38
rs869184647 Chr.1: 27355950-27355951 on Build GRCh38
rs1051208866 Chr.1: 27355952-27355952 on Build GRCh38
rs140784694 Chr.1: 27356086-27356086 on Build GRCh38
rs757596262 Chr.1: 27356015-27356015 on Build GRCh38
rs377394094 Chr.1: 27355756-27355756 on Build GRCh38
rs1039012532 Chr.1: 27355881-27355881 on Build GRCh38
rs755523786 Chr.1: 27355950-27355950 on Build GRCh38
rs375004232 Chr.1: 27355724-27355724 on Build GRCh38
rs141031254 Chr.1: 27355726-27355726 on Build GRCh38
rs766874539 Chr.1: 27355983-27355983 on Build GRCh38
rs900095613 Chr.1: 27355914-27355914 on Build GRCh38
rs1033988352 Chr.1: 27356055-27356055 on Build GRCh38

View Full Product Details