Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 62807370-62807909 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADO
Entrez Gene ID
84890
Gene Name
2-aminoethanethiol dioxygenase
Gene Aliases
C10orf22
UniGene
Hs.99821
Gene Chromosome Location
Chr.10: 62804756-62808479 on Build GRCh38
Transcripts
NM_032804.5

SNP Detail

SNP ID Chromosome Location
rs959668992 Chr.10: 62807802-62807802 on Build GRCh38
rs1055963681 Chr.10: 62807595-62807595 on Build GRCh38
rs377593874 Chr.10: 62807717-62807717 on Build GRCh38
rs188386742 Chr.10: 62807643-62807643 on Build GRCh38
rs931118700 Chr.10: 62807503-62807503 on Build GRCh38
rs1049813324 Chr.10: 62807660-62807660 on Build GRCh38
rs180744896 Chr.10: 62807400-62807400 on Build GRCh38
rs976563563 Chr.10: 62807594-62807594 on Build GRCh38
rs952503958 Chr.10: 62807828-62807828 on Build GRCh38
rs796455913 Chr.10: 62807769-62807769 on Build GRCh38
rs1012557009 Chr.10: 62807817-62807817 on Build GRCh38
rs967515457 Chr.10: 62807576-62807576 on Build GRCh38
rs747529316 Chr.10: 62807647-62807647 on Build GRCh38
rs918240365 Chr.10: 62807740-62807740 on Build GRCh38
rs550198698 Chr.10: 62807777-62807777 on Build GRCh38
rs1035261391 Chr.10: 62807795-62807795 on Build GRCh38
rs982804869 Chr.10: 62807433-62807433 on Build GRCh38
rs909439227 Chr.10: 62807704-62807705 on Build GRCh38
rs1003731016 Chr.10: 62807762-62807762 on Build GRCh38
rs375555384 Chr.10: 62807486-62807486 on Build GRCh38
rs902717951 Chr.10: 62807737-62807737 on Build GRCh38
rs994310499 Chr.10: 62807439-62807439 on Build GRCh38
rs528298416 Chr.10: 62807587-62807587 on Build GRCh38
rs1038097272 Chr.10: 62807416-62807416 on Build GRCh38
rs1045853781 Chr.10: 62807816-62807817 on Build GRCh38
rs907028204 Chr.10: 62807470-62807470 on Build GRCh38
rs577951319 Chr.10: 62807691-62807691 on Build GRCh38
rs185407235 Chr.10: 62807412-62807412 on Build GRCh38
rs1041023641 Chr.10: 62807607-62807607 on Build GRCh38
rs545906921 Chr.10: 62807883-62807883 on Build GRCh38
rs1034236555 Chr.10: 62807485-62807486 on Build GRCh38
rs1023099756 Chr.10: 62807826-62807826 on Build GRCh38
rs908422923 Chr.10: 62807516-62807516 on Build GRCh38
rs776703758 Chr.10: 62807745-62807745 on Build GRCh38
rs1035315133 Chr.10: 62807446-62807446 on Build GRCh38
rs893893487 Chr.10: 62807701-62807701 on Build GRCh38
rs1509964 Chr.10: 62807604-62807604 on Build GRCh38
rs575643060 Chr.10: 62807493-62807493 on Build GRCh38
rs772363305 Chr.10: 62807605-62807605 on Build GRCh38
rs983975226 Chr.10: 62807510-62807510 on Build GRCh38
rs568493992 Chr.10: 62807832-62807832 on Build GRCh38
rs957373284 Chr.10: 62807490-62807490 on Build GRCh38
rs1020684834 Chr.10: 62807543-62807543 on Build GRCh38
rs78951580 Chr.10: 62807407-62807407 on Build GRCh38
rs907045250 Chr.10: 62807817-62807817 on Build GRCh38
rs959720985 Chr.10: 62807483-62807483 on Build GRCh38
rs769202824 Chr.10: 62807706-62807706 on Build GRCh38
rs564339739 Chr.10: 62807531-62807531 on Build GRCh38
rs896927919 Chr.10: 62807425-62807425 on Build GRCh38
rs986353127 Chr.10: 62807661-62807661 on Build GRCh38
rs369367430 Chr.10: 62807494-62807494 on Build GRCh38

View Full Product Details