Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 54636628-54637027 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LILRB1
Entrez Gene ID
10859
Gene Name
leukocyte immunoglobulin like receptor B1
Gene Aliases
CD85J, ILT-2, ILT2, LIR-1, LIR1, MIR-7, MIR7, PIR-B, PIRB
UniGene
Hs.667388
Gene Chromosome Location
Chr.19: 54616930-54637925 on Build GRCh38
Transcripts
NM_001081637.2 NM_001081638.3 NM_001081639.3 NM_001278398.2 NM_006669.6 NR_103518.2 XM_011526331.2 XM_011526332.2 XM_011526335.2 XM_011526336.2 XM_017026182.1 XM_017026183.1 XM_017026184.1 XM_017026185.1 XM_017026186.1 XM_017026187.1 XM_017026188.1 XM_017026189.1 XM_017026190.1 XM_017026191.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1481496 c.1791G>A p.R597R Chr.19: 54636637-54636637 on Build GRCh38
388101 c.1806+2T>A p.? Chr.19: 54636654-54636654 on Build GRCh38
1612799 c.1896C>A p.S632S Chr.19: 54636821-54636821 on Build GRCh38
124112 c.1816T>A p.S606T Chr.19: 54636741-54636741 on Build GRCh38
3423160 c.1842C>T p.Y614Y Chr.19: 54636767-54636767 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748534366 Chr.19: 54636886-54636886 on Build GRCh38
rs773631395 Chr.19: 54636889-54636889 on Build GRCh38
rs768540665 Chr.19: 54636695-54636695 on Build GRCh38
rs202014540 Chr.19: 54636884-54636884 on Build GRCh38
rs200208553 Chr.19: 54636844-54636844 on Build GRCh38
rs2104402 Chr.19: 54636708-54636708 on Build GRCh38
rs865783373 Chr.19: 54636875-54636875 on Build GRCh38
rs776521452 Chr.19: 54636830-54636831 on Build GRCh38
rs554496261 Chr.19: 54636881-54636881 on Build GRCh38
rs996099779 Chr.19: 54636828-54636828 on Build GRCh38
rs754062816 Chr.19: 54636687-54636687 on Build GRCh38
rs201250339 Chr.19: 54636654-54636654 on Build GRCh38
rs746914203 Chr.19: 54636772-54636772 on Build GRCh38
rs749861084 Chr.19: 54636685-54636685 on Build GRCh38
rs190988279 Chr.19: 54636689-54636689 on Build GRCh38
rs531820343 Chr.19: 54636788-54636788 on Build GRCh38
rs763488577 Chr.19: 54636907-54636908 on Build GRCh38
rs775609439 Chr.19: 54636837-54636837 on Build GRCh38
rs767213485 Chr.19: 54636917-54636918 on Build GRCh38
rs764525031 Chr.19: 54636723-54636723 on Build GRCh38
rs759415292 Chr.19: 54636832-54636834 on Build GRCh38
rs768398249 Chr.19: 54636662-54636662 on Build GRCh38
rs554111310 Chr.19: 54636892-54636892 on Build GRCh38
rs634222 Chr.19: 54636785-54636785 on Build GRCh38
rs755566487 Chr.19: 54636693-54636693 on Build GRCh38
rs766721159 Chr.19: 54636685-54636685 on Build GRCh38
rs751120567 Chr.19: 54636913-54636914 on Build GRCh38
rs887248063 Chr.19: 54636832-54636832 on Build GRCh38
rs761629909 Chr.19: 54636674-54636674 on Build GRCh38
rs761734409 Chr.19: 54636921-54636921 on Build GRCh38
rs753002642 Chr.19: 54636721-54636721 on Build GRCh38
rs1061684 Chr.19: 54636791-54636791 on Build GRCh38
rs774541702 Chr.19: 54636719-54636720 on Build GRCh38
rs774382935 Chr.19: 54636670-54636670 on Build GRCh38
rs16985478 Chr.19: 54636798-54636798 on Build GRCh38
rs753349590 Chr.19: 54636796-54636796 on Build GRCh38
rs758997220 Chr.19: 54636849-54636849 on Build GRCh38
rs748182472 Chr.19: 54636697-54636697 on Build GRCh38
rs371895769 Chr.19: 54636829-54636829 on Build GRCh38
rs760769335 Chr.19: 54636679-54636679 on Build GRCh38
rs765937694 Chr.19: 54636795-54636795 on Build GRCh38
rs8101240 Chr.19: 54636880-54636880 on Build GRCh38
rs41308748 Chr.19: 54636725-54636725 on Build GRCh38
rs574818482 Chr.19: 54636691-54636691 on Build GRCh38
rs767689783 Chr.19: 54636677-54636677 on Build GRCh38
rs1013357493 Chr.19: 54636969-54636969 on Build GRCh38
rs758057879 Chr.19: 54636863-54636863 on Build GRCh38
rs758169671 Chr.19: 54636802-54636802 on Build GRCh38
rs771848801 Chr.19: 54636767-54636767 on Build GRCh38
rs562284717 Chr.19: 54636825-54636825 on Build GRCh38
rs201166306 Chr.19: 54636763-54636763 on Build GRCh38
rs200679487 Chr.19: 54636891-54636891 on Build GRCh38
rs369673231 Chr.19: 54636787-54636787 on Build GRCh38
rs1057179255 Chr.19: 54636945-54636945 on Build GRCh38
rs750382211 Chr.19: 54636925-54636925 on Build GRCh38
rs8101262 Chr.19: 54636939-54636939 on Build GRCh38
rs777882631 Chr.19: 54636768-54636768 on Build GRCh38
rs200966826 Chr.19: 54636750-54636750 on Build GRCh38
rs771186716 Chr.19: 54636776-54636776 on Build GRCh38
rs758501058 Chr.19: 54636963-54636963 on Build GRCh38
rs746261569 Chr.19: 54636678-54636681 on Build GRCh38
rs763002667 Chr.19: 54636906-54636907 on Build GRCh38
rs780825726 Chr.19: 54636690-54636690 on Build GRCh38
rs762486622 Chr.19: 54636898-54636898 on Build GRCh38
rs756819197 Chr.19: 54636914-54636915 on Build GRCh38
rs781380241 Chr.19: 54636676-54636684 on Build GRCh38
rs200702196 Chr.19: 54636850-54636850 on Build GRCh38
rs763314536 Chr.19: 54636722-54636722 on Build GRCh38
rs938817342 Chr.19: 54636941-54636941 on Build GRCh38
rs777690584 Chr.19: 54636810-54636810 on Build GRCh38
rs765692940 Chr.19: 54636720-54636720 on Build GRCh38
rs752016180 Chr.19: 54636724-54636724 on Build GRCh38
rs759869507 Chr.19: 54636783-54636783 on Build GRCh38
rs370106064 Chr.19: 54636908-54636908 on Build GRCh38
rs757063636 Chr.19: 54636878-54636878 on Build GRCh38
rs755427263 Chr.19: 54636688-54636688 on Build GRCh38
rs373547731 Chr.19: 54636923-54636923 on Build GRCh38
rs756072447 Chr.19: 54636926-54636926 on Build GRCh38
rs752108032 Chr.19: 54636789-54636789 on Build GRCh38
rs774688543 Chr.19: 54636843-54636843 on Build GRCh38
rs748904486 Chr.19: 54636694-54636694 on Build GRCh38
rs748289578 Chr.19: 54636748-54636748 on Build GRCh38
rs768606488 Chr.19: 54636685-54636689 on Build GRCh38
rs41548213 Chr.19: 54636848-54636848 on Build GRCh38
rs756556488 Chr.19: 54636682-54636682 on Build GRCh38
rs67568554 Chr.19: 54636684-54636684 on Build GRCh38
rs199564534 Chr.19: 54636858-54636858 on Build GRCh38
rs879508695 Chr.19: 54636949-54636949 on Build GRCh38
rs764558492 Chr.19: 54636982-54636982 on Build GRCh38
rs769063443 Chr.19: 54636842-54636842 on Build GRCh38
rs781303190 Chr.19: 54636879-54636879 on Build GRCh38
rs1002930790 Chr.19: 54636833-54636833 on Build GRCh38
rs368877199 Chr.19: 54636705-54636705 on Build GRCh38
rs775561177 Chr.19: 54636883-54636883 on Build GRCh38
rs751211572 Chr.19: 54636909-54636909 on Build GRCh38
rs371743660 Chr.19: 54636661-54636661 on Build GRCh38
rs570772460 Chr.19: 54636845-54636845 on Build GRCh38
rs776837939 Chr.19: 54636779-54636779 on Build GRCh38
rs868783816 Chr.19: 54636907-54636907 on Build GRCh38
rs765194697 Chr.19: 54636882-54636882 on Build GRCh38
rs1035696164 Chr.19: 54636834-54636834 on Build GRCh38
rs8101469 Chr.19: 54636927-54636927 on Build GRCh38
rs774498158 Chr.19: 54636885-54636885 on Build GRCh38
rs756981570 Chr.19: 54636652-54636652 on Build GRCh38
rs759391990 Chr.19: 54636747-54636747 on Build GRCh38
rs201395386 Chr.19: 54636816-54636816 on Build GRCh38
rs1047213292 Chr.19: 54636831-54636831 on Build GRCh38
rs772374183 Chr.19: 54636825-54636826 on Build GRCh38
rs764047788 Chr.19: 54636865-54636865 on Build GRCh38
rs931593747 Chr.19: 54636938-54636938 on Build GRCh38
rs769871972 Chr.19: 54636835-54636835 on Build GRCh38
rs527847806 Chr.19: 54636838-54636838 on Build GRCh38
rs546237047 Chr.19: 54636814-54636814 on Build GRCh38
rs373606950 Chr.19: 54636700-54636700 on Build GRCh38
rs199786967 Chr.19: 54636678-54636678 on Build GRCh38
rs540511264 Chr.19: 54636696-54636696 on Build GRCh38
rs751405593 Chr.19: 54636871-54636871 on Build GRCh38
rs201258189 Chr.19: 54636732-54636732 on Build GRCh38
rs979286427 Chr.19: 54636748-54636748 on Build GRCh38
rs111542900 Chr.19: 54636952-54636952 on Build GRCh38
rs752353124 Chr.19: 54636800-54636800 on Build GRCh38
rs753858305 Chr.19: 54636681-54636681 on Build GRCh38
rs760859023 Chr.19: 54636701-54636701 on Build GRCh38

View Full Product Details