Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41869232-41869737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACLY
Entrez Gene ID
47
Gene Name
ATP citrate lyase
Gene Aliases
ACL, ATPCL, CLATP
UniGene
Hs.387567
Gene Chromosome Location
Chr.17: 41866916-41930542 on Build GRCh38
Transcripts
NM_001096.2 NM_001303274.1 NM_001303275.1 NM_198830.1 XM_005257395.1 XM_017024688.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5669299 c.2976T>C p.D992D Chr.17: 41869549-41869549 on Build GRCh38
436610 c.2996C>T p.P999L Chr.17: 41869529-41869529 on Build GRCh38
3188002 c.2979C>T p.Y993Y Chr.17: 41869546-41869546 on Build GRCh38
1302826 c.3030A>T p.V1010V Chr.17: 41869495-41869495 on Build GRCh38
3717363 c.2999C>T p.A1000V Chr.17: 41869526-41869526 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs556732165 Chr.17: 41869686-41869686 on Build GRCh38
rs782214778 Chr.17: 41869609-41869609 on Build GRCh38
rs971173504 Chr.17: 41869662-41869662 on Build GRCh38
rs782073119 Chr.17: 41869610-41869611 on Build GRCh38
rs566312834 Chr.17: 41869525-41869525 on Build GRCh38
rs3744780 Chr.17: 41869417-41869417 on Build GRCh38
rs995660871 Chr.17: 41869511-41869511 on Build GRCh38
rs781832836 Chr.17: 41869483-41869483 on Build GRCh38
rs754494996 Chr.17: 41869280-41869280 on Build GRCh38
rs782778188 Chr.17: 41869515-41869515 on Build GRCh38
rs144346203 Chr.17: 41869546-41869546 on Build GRCh38
rs782253764 Chr.17: 41869570-41869570 on Build GRCh38
rs891690282 Chr.17: 41869322-41869322 on Build GRCh38
rs946043546 Chr.17: 41869328-41869328 on Build GRCh38
rs782149851 Chr.17: 41869632-41869632 on Build GRCh38
rs782366209 Chr.17: 41869613-41869613 on Build GRCh38
rs927035536 Chr.17: 41869675-41869675 on Build GRCh38
rs200574187 Chr.17: 41869451-41869451 on Build GRCh38
rs371825708 Chr.17: 41869459-41869459 on Build GRCh38
rs782140647 Chr.17: 41869457-41869457 on Build GRCh38
rs782114788 Chr.17: 41869510-41869510 on Build GRCh38
rs781890934 Chr.17: 41869521-41869521 on Build GRCh38
rs200051153 Chr.17: 41869526-41869526 on Build GRCh38
rs368520691 Chr.17: 41869443-41869443 on Build GRCh38
rs1006927133 Chr.17: 41869424-41869424 on Build GRCh38
rs782365488 Chr.17: 41869450-41869450 on Build GRCh38
rs782554859 Chr.17: 41869454-41869454 on Build GRCh38
rs374368056 Chr.17: 41869441-41869441 on Build GRCh38
rs782811369 Chr.17: 41869639-41869639 on Build GRCh38
rs545147361 Chr.17: 41869690-41869690 on Build GRCh38
rs782528878 Chr.17: 41869599-41869599 on Build GRCh38
rs74873238 Chr.17: 41869532-41869532 on Build GRCh38
rs782299316 Chr.17: 41869429-41869429 on Build GRCh38
rs578035027 Chr.17: 41869687-41869687 on Build GRCh38
rs935954732 Chr.17: 41869309-41869309 on Build GRCh38
rs898646965 Chr.17: 41869500-41869500 on Build GRCh38
rs1026333389 Chr.17: 41869550-41869550 on Build GRCh38
rs781993667 Chr.17: 41869455-41869455 on Build GRCh38
rs556053302 Chr.17: 41869562-41869562 on Build GRCh38
rs568601775 Chr.17: 41869418-41869418 on Build GRCh38
rs782001029 Chr.17: 41869614-41869614 on Build GRCh38
rs376532911 Chr.17: 41869477-41869477 on Build GRCh38
rs1023967613 Chr.17: 41869624-41869624 on Build GRCh38
rs979827181 Chr.17: 41869672-41869672 on Build GRCh38
rs532823509 Chr.17: 41869442-41869442 on Build GRCh38
rs989961750 Chr.17: 41869712-41869712 on Build GRCh38
rs782732336 Chr.17: 41869482-41869482 on Build GRCh38
rs782711963 Chr.17: 41869260-41869260 on Build GRCh38
rs782281916 Chr.17: 41869426-41869426 on Build GRCh38
rs375423940 Chr.17: 41869316-41869316 on Build GRCh38
rs1054795916 Chr.17: 41869311-41869311 on Build GRCh38
rs781937318 Chr.17: 41869606-41869606 on Build GRCh38
rs782631078 Chr.17: 41869569-41869569 on Build GRCh38
rs538434361 Chr.17: 41869674-41869674 on Build GRCh38
rs781834281 Chr.17: 41869325-41869325 on Build GRCh38

View Full Product Details