Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219234687-219235276 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKZF1
Entrez Gene ID
55139
Gene Name
ankyrin repeat and zinc finger domain containing 1
Gene Aliases
ZNF744
UniGene
Hs.437647
Gene Chromosome Location
Chr.2: 219229757-219236669 on Build GRCh38
Transcripts
NM_001042410.1 NM_001282792.1 NM_018089.2 XM_005246663.2 XM_011511392.2 XM_017004420.1 XM_017004421.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5632576 c.1642G>T p.G548C Chr.2: 219235263-219235263 on Build GRCh38
4466118 c.1417C>T p.Q473* Chr.2: 219235038-219235038 on Build GRCh38
1192088 c.1370C>T p.T457I Chr.2: 219234991-219234991 on Build GRCh38
4637873 c.1415C>T p.S472F Chr.2: 219235036-219235036 on Build GRCh38
5861826 c.1654C>T p.R552C Chr.2: 219235275-219235275 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758181479 Chr.2: 219235031-219235031 on Build GRCh38
rs371881255 Chr.2: 219234764-219234764 on Build GRCh38
rs957573967 Chr.2: 219234912-219234912 on Build GRCh38
rs557678219 Chr.2: 219234807-219234808 on Build GRCh38
rs936994466 Chr.2: 219234755-219234755 on Build GRCh38
rs1054130197 Chr.2: 219235254-219235254 on Build GRCh38
rs751709088 Chr.2: 219234988-219234988 on Build GRCh38
rs376658598 Chr.2: 219235129-219235129 on Build GRCh38
rs749144113 Chr.2: 219234914-219234914 on Build GRCh38
rs751344224 Chr.2: 219235179-219235179 on Build GRCh38
rs762944656 Chr.2: 219235161-219235161 on Build GRCh38
rs768417817 Chr.2: 219234801-219234801 on Build GRCh38
rs371050271 Chr.2: 219234876-219234876 on Build GRCh38
rs764107165 Chr.2: 219235247-219235247 on Build GRCh38
rs765206323 Chr.2: 219235102-219235102 on Build GRCh38
rs770370575 Chr.2: 219235056-219235056 on Build GRCh38
rs369641231 Chr.2: 219235188-219235188 on Build GRCh38
rs889794776 Chr.2: 219234889-219234889 on Build GRCh38
rs747150991 Chr.2: 219235236-219235236 on Build GRCh38
rs981613108 Chr.2: 219234982-219234982 on Build GRCh38
rs755632549 Chr.2: 219235199-219235199 on Build GRCh38
rs747740100 Chr.2: 219234829-219234829 on Build GRCh38
rs756869180 Chr.2: 219235111-219235111 on Build GRCh38
rs536969364 Chr.2: 219234932-219234932 on Build GRCh38
rs765554137 Chr.2: 219234964-219234964 on Build GRCh38
rs994031957 Chr.2: 219234951-219234951 on Build GRCh38
rs773452240 Chr.2: 219235221-219235221 on Build GRCh38
rs752886051 Chr.2: 219234870-219234870 on Build GRCh38
rs761971891 Chr.2: 219235095-219235095 on Build GRCh38
rs752703809 Chr.2: 219235155-219235155 on Build GRCh38
rs780715354 Chr.2: 219235057-219235057 on Build GRCh38
rs1042525022 Chr.2: 219234836-219234836 on Build GRCh38
rs768901905 Chr.2: 219235084-219235084 on Build GRCh38
rs781695896 Chr.2: 219234840-219234842 on Build GRCh38
rs779486619 Chr.2: 219235200-219235200 on Build GRCh38
rs200249107 Chr.2: 219235036-219235036 on Build GRCh38
rs1047718251 Chr.2: 219234726-219234728 on Build GRCh38
rs765545483 Chr.2: 219234868-219234868 on Build GRCh38
rs915528031 Chr.2: 219235015-219235015 on Build GRCh38
rs756106712 Chr.2: 219234894-219234894 on Build GRCh38
rs772861491 Chr.2: 219234928-219234928 on Build GRCh38
rs771606097 Chr.2: 219234935-219234935 on Build GRCh38
rs140395841 Chr.2: 219234991-219234991 on Build GRCh38
rs145554881 Chr.2: 219235086-219235086 on Build GRCh38
rs763986651 Chr.2: 219235165-219235165 on Build GRCh38
rs776786551 Chr.2: 219235246-219235246 on Build GRCh38
rs199921422 Chr.2: 219235108-219235108 on Build GRCh38
rs200050507 Chr.2: 219234841-219234841 on Build GRCh38
rs868726854 Chr.2: 219234779-219234779 on Build GRCh38
rs763885416 Chr.2: 219234792-219234792 on Build GRCh38
rs747785980 Chr.2: 219234922-219234922 on Build GRCh38
rs1051621627 Chr.2: 219235092-219235092 on Build GRCh38
rs144970852 Chr.2: 219234762-219234762 on Build GRCh38
rs185752150 Chr.2: 219234864-219234864 on Build GRCh38
rs767522644 Chr.2: 219235096-219235096 on Build GRCh38
rs755408001 Chr.2: 219235208-219235208 on Build GRCh38
rs1045388904 Chr.2: 219235054-219235054 on Build GRCh38
rs748689495 Chr.2: 219234928-219234930 on Build GRCh38
rs528511639 Chr.2: 219234772-219234772 on Build GRCh38
rs754901479 Chr.2: 219235008-219235008 on Build GRCh38
rs777110647 Chr.2: 219234855-219234855 on Build GRCh38
rs771781981 Chr.2: 219234927-219234927 on Build GRCh38
rs756921185 Chr.2: 219235183-219235183 on Build GRCh38
rs986795948 Chr.2: 219234948-219234950 on Build GRCh38
rs746368114 Chr.2: 219235071-219235071 on Build GRCh38
rs1033162555 Chr.2: 219234874-219234874 on Build GRCh38
rs760804275 Chr.2: 219235145-219235145 on Build GRCh38
rs750448904 Chr.2: 219235099-219235099 on Build GRCh38
rs765127798 Chr.2: 219235150-219235150 on Build GRCh38
rs926970880 Chr.2: 219234739-219234739 on Build GRCh38
rs772277823 Chr.2: 219235132-219235132 on Build GRCh38
rs754704173 Chr.2: 219234915-219234915 on Build GRCh38
rs3172168 Chr.2: 219235164-219235164 on Build GRCh38
rs771728371 Chr.2: 219234830-219234830 on Build GRCh38
rs779848206 Chr.2: 219234911-219234911 on Build GRCh38
rs779840903 Chr.2: 219234791-219234791 on Build GRCh38
rs548641996 Chr.2: 219234824-219234824 on Build GRCh38
rs778575377 Chr.2: 219235014-219235014 on Build GRCh38
rs376576937 Chr.2: 219235241-219235241 on Build GRCh38
rs752605573 Chr.2: 219235103-219235103 on Build GRCh38
rs748531607 Chr.2: 219235212-219235212 on Build GRCh38
rs775860459 Chr.2: 219234972-219234972 on Build GRCh38
rs571484552 Chr.2: 219234863-219234863 on Build GRCh38
rs745372490 Chr.2: 219235116-219235116 on Build GRCh38
rs778718956 Chr.2: 219234917-219234917 on Build GRCh38
rs767197234 Chr.2: 219235187-219235187 on Build GRCh38
rs200734099 Chr.2: 219234835-219234835 on Build GRCh38
rs773222072 Chr.2: 219235135-219235135 on Build GRCh38
rs372235404 Chr.2: 219235202-219235202 on Build GRCh38
rs751584320 Chr.2: 219234884-219234884 on Build GRCh38
rs748884485 Chr.2: 219235046-219235047 on Build GRCh38
rs775779269 Chr.2: 219235077-219235077 on Build GRCh38
rs73077098 Chr.2: 219234763-219234763 on Build GRCh38
rs376794951 Chr.2: 219234960-219234960 on Build GRCh38
rs763178651 Chr.2: 219234975-219234975 on Build GRCh38
rs763307622 Chr.2: 219235078-219235078 on Build GRCh38
rs764165841 Chr.2: 219234883-219234883 on Build GRCh38
rs777337376 Chr.2: 219234952-219234954 on Build GRCh38
rs542260959 Chr.2: 219235220-219235220 on Build GRCh38
rs766385438 Chr.2: 219234992-219234992 on Build GRCh38
rs751289211 Chr.2: 219235109-219235109 on Build GRCh38
rs779260347 Chr.2: 219235210-219235210 on Build GRCh38
rs780584011 Chr.2: 219235114-219235114 on Build GRCh38
rs372230672 Chr.2: 219235128-219235128 on Build GRCh38
rs539759074 Chr.2: 219235076-219235076 on Build GRCh38
rs375313444 Chr.2: 219234946-219234946 on Build GRCh38
rs774352425 Chr.2: 219235251-219235251 on Build GRCh38
rs769249256 Chr.2: 219234933-219234951 on Build GRCh38
rs758245062 Chr.2: 219235106-219235106 on Build GRCh38
rs574591851 Chr.2: 219235240-219235240 on Build GRCh38
rs766934677 Chr.2: 219234849-219234849 on Build GRCh38
rs761635123 Chr.2: 219235255-219235255 on Build GRCh38
rs201647899 Chr.2: 219235026-219235026 on Build GRCh38
rs753737968 Chr.2: 219234994-219234994 on Build GRCh38
rs969803554 Chr.2: 219234771-219234771 on Build GRCh38
rs749697933 Chr.2: 219235126-219235126 on Build GRCh38
rs371028494 Chr.2: 219234985-219234985 on Build GRCh38
rs770671490 Chr.2: 219235061-219235061 on Build GRCh38
rs1020428855 Chr.2: 219235105-219235105 on Build GRCh38
rs769655144 Chr.2: 219234784-219234784 on Build GRCh38
rs759404205 Chr.2: 219235100-219235100 on Build GRCh38
rs974179618 Chr.2: 219235143-219235143 on Build GRCh38
rs368930399 Chr.2: 219235122-219235122 on Build GRCh38

View Full Product Details