Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 205087079-205087663 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBBP5
Entrez Gene ID
5929
Gene Name
RB binding protein 5, histone lysine methyltransferase complex subunit
Gene Aliases
RBQ3, SWD1
UniGene
Hs.519230
Gene Chromosome Location
Chr.1: 205086142-205122022 on Build GRCh38
Transcripts
NM_001193272.1 NM_001193273.1 NM_005057.3 XM_011509833.2

SNP Detail

SNP ID Chromosome Location
rs1038743638 Chr.1: 205087196-205087196 on Build GRCh38
rs549559853 Chr.1: 205087640-205087641 on Build GRCh38
rs550316725 Chr.1: 205087279-205087279 on Build GRCh38
rs973602717 Chr.1: 205087114-205087114 on Build GRCh38
rs149917398 Chr.1: 205087413-205087413 on Build GRCh38
rs775964290 Chr.1: 205087291-205087291 on Build GRCh38
rs545633667 Chr.1: 205087626-205087626 on Build GRCh38
rs77932648 Chr.1: 205087574-205087574 on Build GRCh38
rs748890988 Chr.1: 205087576-205087576 on Build GRCh38
rs757677149 Chr.1: 205087319-205087319 on Build GRCh38
rs899184526 Chr.1: 205087537-205087537 on Build GRCh38
rs77799390 Chr.1: 205087612-205087612 on Build GRCh38
rs35535472 Chr.1: 205087199-205087200 on Build GRCh38
rs904308461 Chr.1: 205087198-205087198 on Build GRCh38
rs188747185 Chr.1: 205087343-205087343 on Build GRCh38
rs559370413 Chr.1: 205087199-205087199 on Build GRCh38
rs182358453 Chr.1: 205087296-205087296 on Build GRCh38
rs935826430 Chr.1: 205087246-205087246 on Build GRCh38
rs891694031 Chr.1: 205087259-205087259 on Build GRCh38
rs376649038 Chr.1: 205087411-205087411 on Build GRCh38
rs920977764 Chr.1: 205087633-205087633 on Build GRCh38
rs950823783 Chr.1: 205087613-205087613 on Build GRCh38
rs994895542 Chr.1: 205087609-205087609 on Build GRCh38
rs557256309 Chr.1: 205087587-205087587 on Build GRCh38
rs905809821 Chr.1: 205087494-205087494 on Build GRCh38
rs949893694 Chr.1: 205087488-205087488 on Build GRCh38
rs547052034 Chr.1: 205087360-205087360 on Build GRCh38
rs11240353 Chr.1: 205087173-205087173 on Build GRCh38
rs529503594 Chr.1: 205087492-205087492 on Build GRCh38
rs1002618131 Chr.1: 205087347-205087347 on Build GRCh38
rs527670689 Chr.1: 205087113-205087113 on Build GRCh38
rs541076822 Chr.1: 205087170-205087170 on Build GRCh38
rs1002197073 Chr.1: 205087501-205087501 on Build GRCh38
rs1130310 Chr.1: 205087438-205087438 on Build GRCh38
rs1026462715 Chr.1: 205087395-205087395 on Build GRCh38
rs891600892 Chr.1: 205087476-205087476 on Build GRCh38
rs755852182 Chr.1: 205087198-205087199 on Build GRCh38
rs1033759830 Chr.1: 205087443-205087443 on Build GRCh38
rs140539676 Chr.1: 205087354-205087354 on Build GRCh38
rs397860389 Chr.1: 205087214-205087215 on Build GRCh38
rs971259105 Chr.1: 205087565-205087565 on Build GRCh38
rs958571356 Chr.1: 205087292-205087292 on Build GRCh38
rs144842069 Chr.1: 205087174-205087174 on Build GRCh38
rs973839465 Chr.1: 205087628-205087628 on Build GRCh38
rs193215290 Chr.1: 205087424-205087424 on Build GRCh38
rs12409915 Chr.1: 205087348-205087348 on Build GRCh38
rs993937126 Chr.1: 205087549-205087549 on Build GRCh38
rs932774430 Chr.1: 205087398-205087398 on Build GRCh38
rs935773806 Chr.1: 205087430-205087430 on Build GRCh38
rs1051383907 Chr.1: 205087126-205087126 on Build GRCh38
rs991358401 Chr.1: 205087318-205087318 on Build GRCh38
rs919496855 Chr.1: 205087134-205087134 on Build GRCh38
rs779712272 Chr.1: 205087451-205087451 on Build GRCh38
rs6660342 Chr.1: 205087119-205087119 on Build GRCh38
rs974250374 Chr.1: 205087375-205087375 on Build GRCh38
rs1008348483 Chr.1: 205087273-205087273 on Build GRCh38
rs533147508 Chr.1: 205087554-205087554 on Build GRCh38
rs1130311 Chr.1: 205087397-205087397 on Build GRCh38
rs973550590 Chr.1: 205087359-205087359 on Build GRCh38
rs1052910557 Chr.1: 205087255-205087255 on Build GRCh38
rs565818179 Chr.1: 205087353-205087353 on Build GRCh38
rs929569323 Chr.1: 205087160-205087160 on Build GRCh38
rs61068820 Chr.1: 205087575-205087575 on Build GRCh38
rs1042378870 Chr.1: 205087420-205087420 on Build GRCh38
rs1024141898 Chr.1: 205087327-205087327 on Build GRCh38
rs943039645 Chr.1: 205087195-205087195 on Build GRCh38
rs34533321 Chr.1: 205087526-205087526 on Build GRCh38
rs1052819549 Chr.1: 205087440-205087440 on Build GRCh38
rs541107950 Chr.1: 205087193-205087194 on Build GRCh38
rs945633595 Chr.1: 205087419-205087419 on Build GRCh38
rs925710282 Chr.1: 205087429-205087429 on Build GRCh38
rs755748071 Chr.1: 205087421-205087421 on Build GRCh38

View Full Product Details