Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 163443692-163444262 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNG1
Entrez Gene ID
900
Gene Name
cyclin G1
Gene Aliases
CCNG
UniGene
Hs.79101
Gene Chromosome Location
Chr.5: 163437567-163445016 on Build GRCh38
Transcripts
NM_004060.3 NM_199246.1

SNP Detail

SNP ID Chromosome Location
rs1055356076 Chr.5: 163443986-163443986 on Build GRCh38
rs925608643 Chr.5: 163443795-163443795 on Build GRCh38
rs1004527203 Chr.5: 163443909-163443909 on Build GRCh38
rs997966972 Chr.5: 163443847-163443847 on Build GRCh38
rs1039397240 Chr.5: 163443825-163443825 on Build GRCh38
rs751080292 Chr.5: 163443885-163443885 on Build GRCh38
rs754546531 Chr.5: 163444058-163444058 on Build GRCh38
rs759039482 Chr.5: 163443928-163443928 on Build GRCh38
rs987526549 Chr.5: 163443905-163443905 on Build GRCh38
rs547476702 Chr.5: 163443900-163443900 on Build GRCh38
rs1021533521 Chr.5: 163444157-163444157 on Build GRCh38
rs550314620 Chr.5: 163443827-163443827 on Build GRCh38
rs914091666 Chr.5: 163443750-163443750 on Build GRCh38
rs766545484 Chr.5: 163443729-163443729 on Build GRCh38
rs767105467 Chr.5: 163443969-163443969 on Build GRCh38
rs935614299 Chr.5: 163443807-163443807 on Build GRCh38
rs548708771 Chr.5: 163443724-163443724 on Build GRCh38
rs931164033 Chr.5: 163443982-163443982 on Build GRCh38
rs1032061398 Chr.5: 163443814-163443814 on Build GRCh38
rs1037754610 Chr.5: 163443943-163443943 on Build GRCh38
rs751747655 Chr.5: 163443742-163443742 on Build GRCh38
rs538874609 Chr.5: 163443853-163443853 on Build GRCh38
rs1002125349 Chr.5: 163443952-163443952 on Build GRCh38
rs940659906 Chr.5: 163443933-163443933 on Build GRCh38
rs943617302 Chr.5: 163444226-163444226 on Build GRCh38
rs565093529 Chr.5: 163443781-163443781 on Build GRCh38
rs565847062 Chr.5: 163444150-163444150 on Build GRCh38
rs766343551 Chr.5: 163443752-163443753 on Build GRCh38
rs898419201 Chr.5: 163443950-163443950 on Build GRCh38
rs1013918775 Chr.5: 163444089-163444090 on Build GRCh38
rs766423181 Chr.5: 163444050-163444050 on Build GRCh38
rs958210827 Chr.5: 163443815-163443815 on Build GRCh38
rs992278729 Chr.5: 163444027-163444027 on Build GRCh38
rs2291790 Chr.5: 163444179-163444179 on Build GRCh38
rs980013359 Chr.5: 163444192-163444192 on Build GRCh38
rs926670034 Chr.5: 163443718-163443718 on Build GRCh38
rs899555959 Chr.5: 163443837-163443837 on Build GRCh38
rs571492146 Chr.5: 163443833-163443833 on Build GRCh38
rs2069368 Chr.5: 163443720-163443720 on Build GRCh38
rs149348306 Chr.5: 163444204-163444204 on Build GRCh38
rs746775628 Chr.5: 163443922-163443922 on Build GRCh38
rs191618240 Chr.5: 163443762-163443762 on Build GRCh38
rs960754561 Chr.5: 163443974-163443974 on Build GRCh38
rs895087859 Chr.5: 163444062-163444062 on Build GRCh38
rs2069369 Chr.5: 163443979-163443979 on Build GRCh38
rs992706892 Chr.5: 163443730-163443730 on Build GRCh38

View Full Product Details