Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 43812860-43813440 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MFAP1
Entrez Gene ID
4236
Gene Name
microfibrillar associated protein 1
Gene Aliases
-
UniGene
Hs.61418
Gene Chromosome Location
Chr.15: 43804535-43824753 on Build GRCh38
Transcripts
NM_005926.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7117319 c.760G>A p.E254K Chr.15: 43813114-43813114 on Build GRCh38
433936 c.757G>C p.E253Q Chr.15: 43813117-43813117 on Build GRCh38
124423 c.700G>A p.E234K Chr.15: 43813275-43813275 on Build GRCh38
4186401 c.625C>T p.R209* Chr.15: 43813350-43813350 on Build GRCh38
4054888 c.691C>T p.R231C Chr.15: 43813284-43813284 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754648389 Chr.15: 43813340-43813345 on Build GRCh38
rs750166071 Chr.15: 43813088-43813088 on Build GRCh38
rs746455594 Chr.15: 43812998-43812998 on Build GRCh38
rs761691100 Chr.15: 43813171-43813171 on Build GRCh38
rs767634336 Chr.15: 43813081-43813081 on Build GRCh38
rs775277806 Chr.15: 43813387-43813387 on Build GRCh38
rs746121259 Chr.15: 43813241-43813241 on Build GRCh38
rs115417609 Chr.15: 43812993-43812993 on Build GRCh38
rs771230073 Chr.15: 43813068-43813068 on Build GRCh38
rs1003553457 Chr.15: 43812915-43812915 on Build GRCh38
rs1048502 Chr.15: 43813288-43813288 on Build GRCh38
rs765071360 Chr.15: 43812980-43812980 on Build GRCh38
rs754503743 Chr.15: 43813105-43813105 on Build GRCh38
rs755479126 Chr.15: 43813019-43813019 on Build GRCh38
rs1035012378 Chr.15: 43812924-43812924 on Build GRCh38
rs908677817 Chr.15: 43813131-43813131 on Build GRCh38
rs763742625 Chr.15: 43813392-43813392 on Build GRCh38
rs775672414 Chr.15: 43813107-43813107 on Build GRCh38
rs1020112057 Chr.15: 43813085-43813085 on Build GRCh38
rs1028172029 Chr.15: 43813237-43813237 on Build GRCh38
rs536461217 Chr.15: 43813240-43813240 on Build GRCh38
rs574154397 Chr.15: 43813345-43813345 on Build GRCh38
rs763882669 Chr.15: 43812947-43812947 on Build GRCh38
rs762059478 Chr.15: 43813332-43813332 on Build GRCh38
rs1023586077 Chr.15: 43812929-43812929 on Build GRCh38
rs376793426 Chr.15: 43813212-43813212 on Build GRCh38
rs768608251 Chr.15: 43813244-43813244 on Build GRCh38
rs548201833 Chr.15: 43813030-43813030 on Build GRCh38
rs751207702 Chr.15: 43813104-43813104 on Build GRCh38
rs372598196 Chr.15: 43813211-43813211 on Build GRCh38
rs114388454 Chr.15: 43813270-43813270 on Build GRCh38
rs748674863 Chr.15: 43813024-43813024 on Build GRCh38
rs769709479 Chr.15: 43813179-43813179 on Build GRCh38
rs757813387 Chr.15: 43813295-43813295 on Build GRCh38
rs940439274 Chr.15: 43813172-43813172 on Build GRCh38
rs778364172 Chr.15: 43813220-43813220 on Build GRCh38
rs985902763 Chr.15: 43813011-43813011 on Build GRCh38
rs758002968 Chr.15: 43812989-43812989 on Build GRCh38
rs1044688121 Chr.15: 43813320-43813320 on Build GRCh38
rs780384348 Chr.15: 43813323-43813323 on Build GRCh38
rs754381554 Chr.15: 43813232-43813232 on Build GRCh38
rs892338501 Chr.15: 43813402-43813402 on Build GRCh38
rs376271613 Chr.15: 43813331-43813331 on Build GRCh38
rs759205368 Chr.15: 43813208-43813208 on Build GRCh38
rs759625890 Chr.15: 43813077-43813077 on Build GRCh38
rs377106172 Chr.15: 43812987-43812987 on Build GRCh38
rs201564018 Chr.15: 43813199-43813199 on Build GRCh38
rs778023356 Chr.15: 43813115-43813115 on Build GRCh38
rs1130044 Chr.15: 43813129-43813129 on Build GRCh38
rs765784772 Chr.15: 43813407-43813407 on Build GRCh38
rs781615281 Chr.15: 43813022-43813022 on Build GRCh38
rs767129614 Chr.15: 43813266-43813266 on Build GRCh38
rs199924008 Chr.15: 43813046-43813046 on Build GRCh38
rs369786708 Chr.15: 43813242-43813242 on Build GRCh38
rs766158910 Chr.15: 43813195-43813195 on Build GRCh38
rs558861555 Chr.15: 43813246-43813246 on Build GRCh38
rs771748185 Chr.15: 43813377-43813377 on Build GRCh38
rs529757446 Chr.15: 43813181-43813181 on Build GRCh38
rs375134441 Chr.15: 43813170-43813170 on Build GRCh38
rs772097392 Chr.15: 43813159-43813159 on Build GRCh38
rs1000671894 Chr.15: 43812966-43812966 on Build GRCh38
rs749246546 Chr.15: 43813338-43813338 on Build GRCh38
rs756604625 Chr.15: 43813114-43813114 on Build GRCh38
rs138230004 Chr.15: 43813258-43813258 on Build GRCh38
rs758566526 Chr.15: 43813317-43813317 on Build GRCh38
rs763689659 Chr.15: 43813281-43813281 on Build GRCh38
rs373335300 Chr.15: 43813283-43813283 on Build GRCh38
rs147939605 Chr.15: 43813083-43813083 on Build GRCh38
rs765733336 Chr.15: 43813302-43813302 on Build GRCh38
rs771285123 Chr.15: 43813127-43813127 on Build GRCh38
rs779347296 Chr.15: 43813145-43813145 on Build GRCh38
rs752160310 Chr.15: 43813276-43813276 on Build GRCh38
rs780875647 Chr.15: 43813393-43813394 on Build GRCh38
rs750737501 Chr.15: 43813312-43813312 on Build GRCh38
rs774135063 Chr.15: 43813373-43813373 on Build GRCh38
rs368221373 Chr.15: 43813090-43813090 on Build GRCh38
rs746077647 Chr.15: 43813157-43813157 on Build GRCh38
rs764969883 Chr.15: 43813087-43813087 on Build GRCh38
rs756027647 Chr.15: 43812946-43812947 on Build GRCh38
rs749609306 Chr.15: 43813121-43813121 on Build GRCh38
rs200795331 Chr.15: 43812952-43812952 on Build GRCh38
rs150028567 Chr.15: 43813261-43813261 on Build GRCh38
rs751174921 Chr.15: 43813271-43813273 on Build GRCh38
rs753675249 Chr.15: 43812949-43812949 on Build GRCh38
rs780005858 Chr.15: 43813243-43813243 on Build GRCh38
rs754179458 Chr.15: 43813324-43813324 on Build GRCh38
rs370610836 Chr.15: 43813376-43813376 on Build GRCh38
rs897805822 Chr.15: 43813231-43813231 on Build GRCh38
rs372063834 Chr.15: 43813113-43813113 on Build GRCh38
rs1130041 Chr.15: 43813263-43813263 on Build GRCh38
rs116558850 Chr.15: 43813058-43813058 on Build GRCh38
rs1045361201 Chr.15: 43812896-43812896 on Build GRCh38
rs762797126 Chr.15: 43813190-43813190 on Build GRCh38
rs765946348 Chr.15: 43813100-43813100 on Build GRCh38
rs774632995 Chr.15: 43812904-43812904 on Build GRCh38
rs1012324212 Chr.15: 43812927-43812927 on Build GRCh38
rs779622474 Chr.15: 43813050-43813052 on Build GRCh38
rs761792659 Chr.15: 43813086-43813086 on Build GRCh38
rs1130043 Chr.15: 43813140-43813140 on Build GRCh38
rs776227954 Chr.15: 43813397-43813397 on Build GRCh38
rs760050583 Chr.15: 43813060-43813060 on Build GRCh38
rs993970602 Chr.15: 43812970-43812970 on Build GRCh38
rs774587854 Chr.15: 43813076-43813076 on Build GRCh38
rs757760290 Chr.15: 43813233-43813233 on Build GRCh38
rs759142088 Chr.15: 43813265-43813265 on Build GRCh38
rs553389201 Chr.15: 43813388-43813388 on Build GRCh38
rs774083697 Chr.15: 43813264-43813264 on Build GRCh38
rs756582466 Chr.15: 43813218-43813218 on Build GRCh38

View Full Product Details