Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 149198034-149198530 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABLIM3
Entrez Gene ID
22885
Gene Name
actin binding LIM protein family member 3
Gene Aliases
HMFN1661
UniGene
Hs.49688
Gene Chromosome Location
Chr.5: 149141483-149260439 on Build GRCh38
Transcripts
NM_001301015.1 NM_001301018.1 NM_001301027.1 NM_001301028.1 NM_014945.3 XM_005268392.1 XM_005268393.1 XM_005268394.1 XM_005268395.1 XM_005268396.1 XM_011537609.2 XM_017009238.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5060830 c.225G>T p.Q75H Chr.5: 149198292-149198292 on Build GRCh38
6591249 c.285G>A p.S95S Chr.5: 149198352-149198352 on Build GRCh38
299039 c.335+6T>G p.? Chr.5: 149198408-149198408 on Build GRCh38
736432 c.294C>A p.G98G Chr.5: 149198361-149198361 on Build GRCh38
1672028 c.221A>G p.Y74C Chr.5: 149198288-149198288 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199548691 Chr.5: 149198324-149198324 on Build GRCh38
rs768344927 Chr.5: 149198177-149198177 on Build GRCh38
rs757182651 Chr.5: 149198432-149198432 on Build GRCh38
rs754406235 Chr.5: 149198378-149198378 on Build GRCh38
rs943965898 Chr.5: 149198315-149198315 on Build GRCh38
rs748022848 Chr.5: 149198340-149198340 on Build GRCh38
rs769545470 Chr.5: 149198341-149198341 on Build GRCh38
rs765263007 Chr.5: 149198385-149198385 on Build GRCh38
rs758384739 Chr.5: 149198400-149198400 on Build GRCh38
rs770441625 Chr.5: 149198352-149198352 on Build GRCh38
rs762491848 Chr.5: 149198225-149198225 on Build GRCh38
rs117986539 Chr.5: 149198145-149198145 on Build GRCh38
rs928551949 Chr.5: 149198489-149198489 on Build GRCh38
rs530741758 Chr.5: 149198362-149198362 on Build GRCh38
rs757200798 Chr.5: 149198481-149198481 on Build GRCh38
rs780973202 Chr.5: 149198190-149198190 on Build GRCh38
rs753323555 Chr.5: 149198212-149198212 on Build GRCh38
rs764536483 Chr.5: 149198377-149198377 on Build GRCh38
rs746086610 Chr.5: 149198172-149198172 on Build GRCh38
rs761289040 Chr.5: 149198304-149198304 on Build GRCh38
rs777486718 Chr.5: 149198416-149198416 on Build GRCh38
rs377045773 Chr.5: 149198152-149198152 on Build GRCh38
rs771453867 Chr.5: 149198448-149198448 on Build GRCh38
rs773109048 Chr.5: 149198214-149198214 on Build GRCh38
rs778311975 Chr.5: 149198437-149198437 on Build GRCh38
rs764847515 Chr.5: 149198444-149198444 on Build GRCh38
rs145134862 Chr.5: 149198299-149198299 on Build GRCh38
rs759199343 Chr.5: 149198431-149198431 on Build GRCh38
rs558905586 Chr.5: 149198139-149198139 on Build GRCh38
rs540651699 Chr.5: 149198080-149198080 on Build GRCh38
rs375384469 Chr.5: 149198197-149198197 on Build GRCh38
rs6872559 Chr.5: 149198463-149198463 on Build GRCh38
rs769669191 Chr.5: 149198212-149198212 on Build GRCh38
rs373118040 Chr.5: 149198338-149198338 on Build GRCh38
rs749211850 Chr.5: 149198351-149198351 on Build GRCh38
rs199957988 Chr.5: 149198449-149198449 on Build GRCh38
rs977575822 Chr.5: 149198181-149198181 on Build GRCh38
rs746599622 Chr.5: 149198453-149198453 on Build GRCh38
rs755961832 Chr.5: 149198413-149198413 on Build GRCh38
rs896754072 Chr.5: 149198120-149198120 on Build GRCh38
rs750466880 Chr.5: 149198386-149198386 on Build GRCh38
rs757993871 Chr.5: 149198307-149198307 on Build GRCh38
rs377176079 Chr.5: 149198226-149198226 on Build GRCh38
rs1054297397 Chr.5: 149198096-149198096 on Build GRCh38
rs780870461 Chr.5: 149198332-149198332 on Build GRCh38
rs754389834 Chr.5: 149198308-149198308 on Build GRCh38
rs765829047 Chr.5: 149198195-149198195 on Build GRCh38
rs759146275 Chr.5: 149198359-149198359 on Build GRCh38
rs766661683 Chr.5: 149198405-149198405 on Build GRCh38
rs750610592 Chr.5: 149198318-149198318 on Build GRCh38
rs953866469 Chr.5: 149198266-149198266 on Build GRCh38
rs868430620 Chr.5: 149198265-149198265 on Build GRCh38
rs896722980 Chr.5: 149198427-149198427 on Build GRCh38
rs778989066 Chr.5: 149198309-149198309 on Build GRCh38
rs111421465 Chr.5: 149198151-149198151 on Build GRCh38
rs879521478 Chr.5: 149198499-149198499 on Build GRCh38
rs142971991 Chr.5: 149198070-149198070 on Build GRCh38
rs945928362 Chr.5: 149198474-149198474 on Build GRCh38
rs375798677 Chr.5: 149198373-149198373 on Build GRCh38
rs377666476 Chr.5: 149198058-149198058 on Build GRCh38
rs752286038 Chr.5: 149198191-149198191 on Build GRCh38
rs773934073 Chr.5: 149198238-149198238 on Build GRCh38
rs985161257 Chr.5: 149198446-149198446 on Build GRCh38
rs375779902 Chr.5: 149198056-149198056 on Build GRCh38
rs764209198 Chr.5: 149198375-149198375 on Build GRCh38
rs762339504 Chr.5: 149198383-149198383 on Build GRCh38
rs990382815 Chr.5: 149198458-149198458 on Build GRCh38
rs745435669 Chr.5: 149198444-149198444 on Build GRCh38
rs891683611 Chr.5: 149198100-149198100 on Build GRCh38
rs767782062 Chr.5: 149198296-149198296 on Build GRCh38
rs548869504 Chr.5: 149198363-149198363 on Build GRCh38
rs749228275 Chr.5: 149198422-149198422 on Build GRCh38
rs867501685 Chr.5: 149198452-149198452 on Build GRCh38
rs974220651 Chr.5: 149198189-149198189 on Build GRCh38
rs759170287 Chr.5: 149198256-149198256 on Build GRCh38
rs112480753 Chr.5: 149198412-149198412 on Build GRCh38

View Full Product Details