Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 67677127-67677694 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPHN
Entrez Gene ID
10243
Gene Name
gephyrin
Gene Aliases
GEPH, GPH, GPHRYN, HKPX1, MOCODC
UniGene
Hs.208765
Gene Chromosome Location
Chr.14: 66507407-67735831 on Build GRCh38
Transcripts
XM_017020925.1 NM_001252650.1 NM_016026.3

Gene Symbol
RDH11
Entrez Gene ID
51109
Gene Name
retinol dehydrogenase 11 (all-trans/9-cis/11-cis)
Gene Aliases
ARSDR1, CGI82, HCBP12, MDT1, PSDR1, RALR1, RDJCSS, SCALD, SDR7C1
UniGene
Hs.719925
Gene Chromosome Location
Chr.14: 67676800-67695793 on Build GRCh38
Transcripts
XM_017020925.1 NM_001252650.1 NM_016026.3

SNP Detail

SNP ID Chromosome Location
rs896879203 Chr.14: 67677307-67677307 on Build GRCh38
rs768377756 Chr.14: 67677363-67677364 on Build GRCh38
rs547443174 Chr.14: 67677628-67677628 on Build GRCh38
rs1023739202 Chr.14: 67677238-67677238 on Build GRCh38
rs772996305 Chr.14: 67677173-67677173 on Build GRCh38
rs972879156 Chr.14: 67677299-67677299 on Build GRCh38
rs929665114 Chr.14: 67677335-67677336 on Build GRCh38
rs760549319 Chr.14: 67677233-67677233 on Build GRCh38
rs7765 Chr.14: 67677612-67677612 on Build GRCh38
rs578049675 Chr.14: 67677179-67677179 on Build GRCh38
rs71129846 Chr.14: 67677392-67677393 on Build GRCh38
rs776699207 Chr.14: 67677536-67677536 on Build GRCh38
rs554367375 Chr.14: 67677354-67677357 on Build GRCh38
rs956968338 Chr.14: 67677558-67677558 on Build GRCh38
rs747016215 Chr.14: 67677197-67677197 on Build GRCh38
rs888116194 Chr.14: 67677355-67677355 on Build GRCh38
rs1003149614 Chr.14: 67677262-67677262 on Build GRCh38
rs984199441 Chr.14: 67677406-67677406 on Build GRCh38
rs199566697 Chr.14: 67677196-67677198 on Build GRCh38
rs960279086 Chr.14: 67677287-67677287 on Build GRCh38
rs1048495271 Chr.14: 67677342-67677342 on Build GRCh38
rs370783174 Chr.14: 67677339-67677339 on Build GRCh38
rs1011136541 Chr.14: 67677629-67677629 on Build GRCh38
rs935199197 Chr.14: 67677571-67677571 on Build GRCh38
rs865866963 Chr.14: 67677193-67677195 on Build GRCh38
rs967110293 Chr.14: 67677496-67677514 on Build GRCh38
rs370517528 Chr.14: 67677439-67677439 on Build GRCh38
rs371030190 Chr.14: 67677516-67677516 on Build GRCh38
rs969104489 Chr.14: 67677244-67677244 on Build GRCh38
rs775490684 Chr.14: 67677362-67677362 on Build GRCh38
rs989580377 Chr.14: 67677562-67677562 on Build GRCh38
rs199532098 Chr.14: 67677346-67677346 on Build GRCh38
rs141648914 Chr.14: 67677413-67677413 on Build GRCh38
rs1012607405 Chr.14: 67677426-67677427 on Build GRCh38
rs374553025 Chr.14: 67677442-67677442 on Build GRCh38
rs768799827 Chr.14: 67677253-67677253 on Build GRCh38
rs376098006 Chr.14: 67677210-67677210 on Build GRCh38
rs759213478 Chr.14: 67677273-67677275 on Build GRCh38
rs1059238 Chr.14: 67677474-67677474 on Build GRCh38
rs904933919 Chr.14: 67677674-67677674 on Build GRCh38
rs879204625 Chr.14: 67677350-67677350 on Build GRCh38
rs986389468 Chr.14: 67677188-67677188 on Build GRCh38
rs542992239 Chr.14: 67677588-67677588 on Build GRCh38
rs1059226 Chr.14: 67677547-67677547 on Build GRCh38
rs1044848674 Chr.14: 67677659-67677659 on Build GRCh38
rs186380369 Chr.14: 67677605-67677605 on Build GRCh38
rs145780940 Chr.14: 67677343-67677346 on Build GRCh38
rs866767995 Chr.14: 67677177-67677177 on Build GRCh38
rs377025166 Chr.14: 67677354-67677354 on Build GRCh38
rs1035937925 Chr.14: 67677267-67677267 on Build GRCh38
rs911280334 Chr.14: 67677470-67677470 on Build GRCh38
rs1023490061 Chr.14: 67677459-67677459 on Build GRCh38
rs918732129 Chr.14: 67677335-67677335 on Build GRCh38
rs776086010 Chr.14: 67677170-67677171 on Build GRCh38
rs796331583 Chr.14: 67677345-67677346 on Build GRCh38
rs575892361 Chr.14: 67677533-67677533 on Build GRCh38

View Full Product Details