Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 106323364-106323952 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TCP11L2
Entrez Gene ID
255394
Gene Name
t-complex 11 like 2
Gene Aliases
-
UniGene
Hs.696047
Gene Chromosome Location
Chr.12: 106302026-106347455 on Build GRCh38
Transcripts
NM_001286262.1 NM_152772.2 XM_005268767.4 XM_005268768.4 XM_011538129.2 XM_011538130.2 XM_011538131.2 XM_011538132.2 XM_017019127.1 XM_017019128.1 XM_017019129.1 XM_017019130.1 XM_017019131.1 XM_017019132.1 XM_017019133.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5813226 c.737G>A p.R246K Chr.12: 106323611-106323611 on Build GRCh38
6798906 c.721T>C p.L241L Chr.12: 106323595-106323595 on Build GRCh38
1358423 c.646C>A p.H216N Chr.12: 106323520-106323520 on Build GRCh38
6798909 c.674T>C p.M225T Chr.12: 106323548-106323548 on Build GRCh38
4622416 c.688A>G p.I230V Chr.12: 106323562-106323562 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766458271 Chr.12: 106323575-106323575 on Build GRCh38
rs763026516 Chr.12: 106323479-106323479 on Build GRCh38
rs957985329 Chr.12: 106323818-106323818 on Build GRCh38
rs752325422 Chr.12: 106323485-106323485 on Build GRCh38
rs1005799110 Chr.12: 106323697-106323697 on Build GRCh38
rs368148424 Chr.12: 106323497-106323497 on Build GRCh38
rs200010594 Chr.12: 106323713-106323717 on Build GRCh38
rs549938099 Chr.12: 106323821-106323821 on Build GRCh38
rs138241328 Chr.12: 106323582-106323582 on Build GRCh38
rs898638491 Chr.12: 106323712-106323712 on Build GRCh38
rs72143923 Chr.12: 106323748-106323748 on Build GRCh38
rs200123683 Chr.12: 106323657-106323657 on Build GRCh38
rs985987111 Chr.12: 106323794-106323794 on Build GRCh38
rs936556689 Chr.12: 106323543-106323543 on Build GRCh38
rs753271990 Chr.12: 106323589-106323589 on Build GRCh38
rs980626371 Chr.12: 106323770-106323770 on Build GRCh38
rs745728340 Chr.12: 106323614-106323614 on Build GRCh38
rs753864504 Chr.12: 106323532-106323532 on Build GRCh38
rs946079628 Chr.12: 106323820-106323820 on Build GRCh38
rs386377664 Chr.12: 106323751-106323752 on Build GRCh38
rs773360079 Chr.12: 106323641-106323641 on Build GRCh38
rs115338323 Chr.12: 106323814-106323814 on Build GRCh38
rs768434623 Chr.12: 106323637-106323637 on Build GRCh38
rs542279329 Chr.12: 106323703-106323703 on Build GRCh38
rs891318549 Chr.12: 106323396-106323396 on Build GRCh38
rs144695471 Chr.12: 106323578-106323578 on Build GRCh38
rs769158061 Chr.12: 106323696-106323697 on Build GRCh38
rs753239372 Chr.12: 106323598-106323598 on Build GRCh38
rs980572423 Chr.12: 106323583-106323583 on Build GRCh38
rs201078311 Chr.12: 106323737-106323738 on Build GRCh38
rs761226883 Chr.12: 106323695-106323695 on Build GRCh38
rs749209106 Chr.12: 106323712-106323713 on Build GRCh38
rs945412417 Chr.12: 106323894-106323894 on Build GRCh38
rs971838709 Chr.12: 106323450-106323450 on Build GRCh38
rs1007057315 Chr.12: 106323399-106323399 on Build GRCh38
rs747724494 Chr.12: 106323683-106323683 on Build GRCh38
rs572259716 Chr.12: 106323763-106323763 on Build GRCh38
rs371393637 Chr.12: 106323664-106323664 on Build GRCh38
rs984459208 Chr.12: 106323433-106323433 on Build GRCh38
rs369338544 Chr.12: 106323554-106323554 on Build GRCh38
rs756515281 Chr.12: 106323590-106323590 on Build GRCh38
rs770865478 Chr.12: 106323713-106323722 on Build GRCh38
rs573435623 Chr.12: 106323691-106323691 on Build GRCh38
rs772039578 Chr.12: 106323525-106323525 on Build GRCh38
rs772573374 Chr.12: 106323667-106323667 on Build GRCh38
rs775959889 Chr.12: 106323683-106323683 on Build GRCh38
rs761267454 Chr.12: 106323690-106323690 on Build GRCh38
rs990347874 Chr.12: 106323735-106323748 on Build GRCh38
rs745642540 Chr.12: 106323524-106323524 on Build GRCh38
rs12424952 Chr.12: 106323446-106323446 on Build GRCh38
rs565846323 Chr.12: 106323419-106323419 on Build GRCh38
rs561029741 Chr.12: 106323909-106323909 on Build GRCh38
rs754638364 Chr.12: 106323554-106323555 on Build GRCh38
rs557725793 Chr.12: 106323481-106323481 on Build GRCh38
rs891097914 Chr.12: 106323871-106323871 on Build GRCh38
rs759776392 Chr.12: 106323658-106323658 on Build GRCh38
rs370191573 Chr.12: 106323711-106323711 on Build GRCh38
rs749303317 Chr.12: 106323646-106323646 on Build GRCh38
rs201211620 Chr.12: 106323747-106323747 on Build GRCh38
rs375924731 Chr.12: 106323692-106323692 on Build GRCh38
rs144794557 Chr.12: 106323748-106323749 on Build GRCh38
rs149344079 Chr.12: 106323538-106323538 on Build GRCh38
rs764926168 Chr.12: 106323696-106323696 on Build GRCh38
rs753430285 Chr.12: 106323693-106323693 on Build GRCh38
rs1051105343 Chr.12: 106323831-106323831 on Build GRCh38
rs201802128 Chr.12: 106323742-106323742 on Build GRCh38
rs1054521807 Chr.12: 106323623-106323623 on Build GRCh38
rs746952367 Chr.12: 106323621-106323621 on Build GRCh38
rs151203894 Chr.12: 106323779-106323779 on Build GRCh38
rs563660101 Chr.12: 106323873-106323873 on Build GRCh38
rs143570657 Chr.12: 106323742-106323743 on Build GRCh38
rs780810107 Chr.12: 106323567-106323568 on Build GRCh38
rs201835036 Chr.12: 106323748-106323748 on Build GRCh38
rs879081645 Chr.12: 106323744-106323744 on Build GRCh38
rs60918030 Chr.12: 106323753-106323754 on Build GRCh38
rs763264237 Chr.12: 106323574-106323574 on Build GRCh38
rs187500023 Chr.12: 106323775-106323775 on Build GRCh38
rs778507581 Chr.12: 106323602-106323602 on Build GRCh38
rs142823625 Chr.12: 106323601-106323601 on Build GRCh38
rs993541064 Chr.12: 106323752-106323752 on Build GRCh38
rs759945434 Chr.12: 106323579-106323579 on Build GRCh38
rs760210066 Chr.12: 106323424-106323424 on Build GRCh38
rs990667604 Chr.12: 106323780-106323780 on Build GRCh38
rs1015903694 Chr.12: 106323434-106323434 on Build GRCh38
rs765443596 Chr.12: 106323459-106323459 on Build GRCh38
rs962455757 Chr.12: 106323440-106323440 on Build GRCh38
rs749449008 Chr.12: 106323736-106323736 on Build GRCh38
rs72028057 Chr.12: 106323749-106323750 on Build GRCh38
rs906583500 Chr.12: 106323605-106323605 on Build GRCh38
rs773470506 Chr.12: 106323476-106323476 on Build GRCh38
rs200198315 Chr.12: 106323743-106323743 on Build GRCh38
rs201390267 Chr.12: 106323744-106323748 on Build GRCh38
rs374835882 Chr.12: 106323593-106323593 on Build GRCh38
rs771221283 Chr.12: 106323651-106323651 on Build GRCh38
rs779507864 Chr.12: 106323742-106323742 on Build GRCh38
rs751260037 Chr.12: 106323714-106323714 on Build GRCh38
rs769929582 Chr.12: 106323550-106323550 on Build GRCh38
rs1039957500 Chr.12: 106323876-106323876 on Build GRCh38
rs779855054 Chr.12: 106323707-106323707 on Build GRCh38
rs774297233 Chr.12: 106323713-106323727 on Build GRCh38
rs933181249 Chr.12: 106323468-106323468 on Build GRCh38
rs766767729 Chr.12: 106323480-106323480 on Build GRCh38
rs989337205 Chr.12: 106323866-106323866 on Build GRCh38
rs374018727 Chr.12: 106323484-106323484 on Build GRCh38
rs756821064 Chr.12: 106323513-106323513 on Build GRCh38
rs748979164 Chr.12: 106323510-106323510 on Build GRCh38
rs71843294 Chr.12: 106323740-106323740 on Build GRCh38
rs200709273 Chr.12: 106323564-106323564 on Build GRCh38
rs771879139 Chr.12: 106323740-106323748 on Build GRCh38
rs879502423 Chr.12: 106323746-106323747 on Build GRCh38
rs371008886 Chr.12: 106323717-106323718 on Build GRCh38
rs755674671 Chr.12: 106323488-106323488 on Build GRCh38
rs757978762 Chr.12: 106323705-106323705 on Build GRCh38
rs140996881 Chr.12: 106323742-106323746 on Build GRCh38
rs891813780 Chr.12: 106323652-106323652 on Build GRCh38
rs754988150 Chr.12: 106323723-106323723 on Build GRCh38
rs951153016 Chr.12: 106323737-106323737 on Build GRCh38
rs780958246 Chr.12: 106323728-106323728 on Build GRCh38
rs995678938 Chr.12: 106323925-106323925 on Build GRCh38
rs759342169 Chr.12: 106323719-106323722 on Build GRCh38
rs778790808 Chr.12: 106323517-106323517 on Build GRCh38
rs758174695 Chr.12: 106323615-106323615 on Build GRCh38
rs536692790 Chr.12: 106323585-106323585 on Build GRCh38

View Full Product Details