Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 129316203-129316514 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
H1FX
Entrez Gene ID
8971
Gene Name
H1 histone family member X
Gene Aliases
H1.10, H1X
UniGene
Hs.75307
Gene Chromosome Location
Chr.3: 129314771-129316277 on Build GRCh38
Transcripts
NM_006026.3 NR_026991.1

Gene Symbol
H1FX-AS1
Entrez Gene ID
339942
Gene Name
H1FX antisense RNA 1
Gene Aliases
C3orf47
UniGene
Hs.450096
Gene Chromosome Location
Chr.3: 129316271-129324569 on Build GRCh38
Transcripts
NM_006026.3 NR_026991.1

SNP Detail

SNP ID Chromosome Location
rs562070518 Chr.3: 129316478-129316478 on Build GRCh38
rs907321293 Chr.3: 129316441-129316441 on Build GRCh38
rs906266986 Chr.3: 129316309-129316309 on Build GRCh38
rs542080815 Chr.3: 129316260-129316260 on Build GRCh38
rs892247435 Chr.3: 129316251-129316254 on Build GRCh38
rs778168417 Chr.3: 129316294-129316294 on Build GRCh38
rs986180137 Chr.3: 129316459-129316459 on Build GRCh38
rs933581647 Chr.3: 129316237-129316237 on Build GRCh38
rs1004862631 Chr.3: 129316278-129316278 on Build GRCh38
rs1030396263 Chr.3: 129316402-129316402 on Build GRCh38
rs1036857689 Chr.3: 129316290-129316290 on Build GRCh38
rs372867471 Chr.3: 129316233-129316233 on Build GRCh38
rs574664871 Chr.3: 129316253-129316253 on Build GRCh38
rs967448687 Chr.3: 129316323-129316323 on Build GRCh38
rs1015436583 Chr.3: 129316345-129316345 on Build GRCh38
rs557083555 Chr.3: 129316279-129316279 on Build GRCh38
rs1009314901 Chr.3: 129316251-129316251 on Build GRCh38
rs931211376 Chr.3: 129316258-129316258 on Build GRCh38
rs563885178 Chr.3: 129316300-129316300 on Build GRCh38
rs1055858373 Chr.3: 129316448-129316448 on Build GRCh38
rs972090248 Chr.3: 129316270-129316270 on Build GRCh38
rs1036359501 Chr.3: 129316439-129316439 on Build GRCh38
rs1011846602 Chr.3: 129316304-129316304 on Build GRCh38
rs113644266 Chr.3: 129316359-129316359 on Build GRCh38
rs887788945 Chr.3: 129316267-129316267 on Build GRCh38
rs13067185 Chr.3: 129316462-129316462 on Build GRCh38
rs975239001 Chr.3: 129316353-129316353 on Build GRCh38
rs907252928 Chr.3: 129316292-129316292 on Build GRCh38
rs940697221 Chr.3: 129316436-129316436 on Build GRCh38
rs1021852137 Chr.3: 129316319-129316319 on Build GRCh38
rs528104409 Chr.3: 129316348-129316348 on Build GRCh38
rs936908781 Chr.3: 129316283-129316283 on Build GRCh38
rs965095401 Chr.3: 129316350-129316350 on Build GRCh38
rs1039503584 Chr.3: 129316252-129316252 on Build GRCh38
rs910539056 Chr.3: 129316474-129316474 on Build GRCh38
rs906170731 Chr.3: 129316255-129316255 on Build GRCh38
rs1002941395 Chr.3: 129316446-129316446 on Build GRCh38
rs757620745 Chr.3: 129316351-129316351 on Build GRCh38
rs756611565 Chr.3: 129316289-129316289 on Build GRCh38
rs552717915 Chr.3: 129316239-129316239 on Build GRCh38
rs908416808 Chr.3: 129316427-129316427 on Build GRCh38
rs977590245 Chr.3: 129316228-129316228 on Build GRCh38
rs779086621 Chr.3: 129316380-129316380 on Build GRCh38
rs553326631 Chr.3: 129316249-129316249 on Build GRCh38
rs1046157203 Chr.3: 129316308-129316308 on Build GRCh38
rs931121525 Chr.3: 129316382-129316382 on Build GRCh38
rs575291958 Chr.3: 129316281-129316281 on Build GRCh38
rs545885996 Chr.3: 129316295-129316295 on Build GRCh38
rs945122861 Chr.3: 129316302-129316302 on Build GRCh38
rs910728935 Chr.3: 129316247-129316247 on Build GRCh38
rs1049080114 Chr.3: 129316261-129316261 on Build GRCh38
rs1048807914 Chr.3: 129316383-129316383 on Build GRCh38
rs865853416 Chr.3: 129316324-129316324 on Build GRCh38
rs369318786 Chr.3: 129316329-129316329 on Build GRCh38
rs1033627957 Chr.3: 129316257-129316257 on Build GRCh38
rs111249161 Chr.3: 129316248-129316248 on Build GRCh38
rs923236310 Chr.3: 129316234-129316234 on Build GRCh38

View Full Product Details