Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 139955618-139956117 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXorf66
Entrez Gene ID
347487
Gene Name
chromosome X open reading frame 66
Gene Aliases
SGPX
UniGene
Hs.121384
Gene Chromosome Location
Chr.X: 139955725-139965518 on Build GRCh38
Transcripts
NM_001013403.2

SNP Detail

SNP ID Chromosome Location
rs376640269 Chr.X: 139956030-139956030 on Build GRCh38
rs776433056 Chr.X: 139956072-139956072 on Build GRCh38
rs747755412 Chr.X: 139956056-139956056 on Build GRCh38
rs766223235 Chr.X: 139955701-139955701 on Build GRCh38
rs768355656 Chr.X: 139955704-139955704 on Build GRCh38
rs149552852 Chr.X: 139956001-139956001 on Build GRCh38
rs1020760618 Chr.X: 139955665-139955668 on Build GRCh38
rs760576216 Chr.X: 139955935-139955935 on Build GRCh38
rs773883172 Chr.X: 139955887-139955887 on Build GRCh38
rs922836766 Chr.X: 139955946-139955946 on Build GRCh38
rs202016289 Chr.X: 139956022-139956022 on Build GRCh38
rs748768957 Chr.X: 139956044-139956044 on Build GRCh38
rs773455019 Chr.X: 139955894-139955894 on Build GRCh38
rs930888606 Chr.X: 139956013-139956013 on Build GRCh38
rs1001263467 Chr.X: 139956062-139956062 on Build GRCh38
rs200431619 Chr.X: 139955958-139955958 on Build GRCh38
rs888235433 Chr.X: 139955651-139955651 on Build GRCh38
rs967792177 Chr.X: 139955764-139955764 on Build GRCh38
rs772971789 Chr.X: 139956064-139956064 on Build GRCh38
rs766261305 Chr.X: 139956071-139956071 on Build GRCh38
rs138947666 Chr.X: 139955925-139955925 on Build GRCh38
rs764629163 Chr.X: 139955767-139955767 on Build GRCh38
rs766839575 Chr.X: 139955995-139955995 on Build GRCh38
rs181545760 Chr.X: 139955856-139955856 on Build GRCh38
rs941141819 Chr.X: 139955970-139955970 on Build GRCh38
rs368846477 Chr.X: 139956091-139956091 on Build GRCh38
rs771755538 Chr.X: 139956057-139956057 on Build GRCh38
rs200754289 Chr.X: 139956051-139956051 on Build GRCh38
rs191143232 Chr.X: 139955757-139955757 on Build GRCh38
rs1001232417 Chr.X: 139955669-139955669 on Build GRCh38
rs368470145 Chr.X: 139956042-139956042 on Build GRCh38
rs973846005 Chr.X: 139955987-139955987 on Build GRCh38
rs770487786 Chr.X: 139955846-139955846 on Build GRCh38
rs751739004 Chr.X: 139955993-139955993 on Build GRCh38
rs767287883 Chr.X: 139955906-139955906 on Build GRCh38
rs764309945 Chr.X: 139955961-139955961 on Build GRCh38
rs937663859 Chr.X: 139956021-139956021 on Build GRCh38
rs768317765 Chr.X: 139955880-139955880 on Build GRCh38
rs186251954 Chr.X: 139956070-139956070 on Build GRCh38
rs149420594 Chr.X: 139955948-139955948 on Build GRCh38
rs1048092598 Chr.X: 139956043-139956043 on Build GRCh38
rs10587653 Chr.X: 139955666-139955669 on Build GRCh38
rs959847940 Chr.X: 139955680-139955680 on Build GRCh38
rs764066780 Chr.X: 139956088-139956088 on Build GRCh38
rs1034175254 Chr.X: 139955677-139955677 on Build GRCh38
rs746749646 Chr.X: 139955700-139955700 on Build GRCh38
rs371532440 Chr.X: 139956086-139956086 on Build GRCh38
rs760928454 Chr.X: 139956076-139956077 on Build GRCh38
rs201034623 Chr.X: 139955996-139955996 on Build GRCh38
rs758438020 Chr.X: 139955953-139955953 on Build GRCh38
rs766351254 Chr.X: 139955947-139955947 on Build GRCh38
rs373697036 Chr.X: 139955862-139955862 on Build GRCh38
rs761610936 Chr.X: 139955905-139955905 on Build GRCh38
rs776259885 Chr.X: 139955731-139955731 on Build GRCh38
rs568926391 Chr.X: 139955670-139955673 on Build GRCh38
rs868443257 Chr.X: 139955976-139955976 on Build GRCh38
rs762631672 Chr.X: 139956050-139956050 on Build GRCh38
rs1033463426 Chr.X: 139955746-139955746 on Build GRCh38

View Full Product Details