Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 132062204-132062752 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HHLA1
Entrez Gene ID
10086
Gene Name
HERV-H LTR-associating 1
Gene Aliases
PLA2L
UniGene
Hs.285026
Gene Chromosome Location
Chr.8: 132061486-132105265 on Build GRCh38
Transcripts
NM_001145095.1

SNP Detail

SNP ID Chromosome Location
rs916915747 Chr.8: 132062246-132062246 on Build GRCh38
rs115023526 Chr.8: 132062639-132062639 on Build GRCh38
rs1051115247 Chr.8: 132062728-132062728 on Build GRCh38
rs539525175 Chr.8: 132062342-132062342 on Build GRCh38
rs898283532 Chr.8: 132062278-132062278 on Build GRCh38
rs918165316 Chr.8: 132062726-132062726 on Build GRCh38
rs896751056 Chr.8: 132062503-132062504 on Build GRCh38
rs898232620 Chr.8: 132062544-132062544 on Build GRCh38
rs979614582 Chr.8: 132062463-132062463 on Build GRCh38
rs1003837497 Chr.8: 132062253-132062253 on Build GRCh38
rs537445453 Chr.8: 132062611-132062611 on Build GRCh38
rs1033218789 Chr.8: 132062577-132062577 on Build GRCh38
rs374568240 Chr.8: 132062448-132062448 on Build GRCh38
rs547074178 Chr.8: 132062240-132062240 on Build GRCh38
rs1039695326 Chr.8: 132062560-132062560 on Build GRCh38
rs1015262724 Chr.8: 132062269-132062269 on Build GRCh38
rs913986859 Chr.8: 132062361-132062361 on Build GRCh38
rs3802261 Chr.8: 132062580-132062580 on Build GRCh38
rs953972430 Chr.8: 132062316-132062316 on Build GRCh38
rs781638126 Chr.8: 132062683-132062683 on Build GRCh38
rs7839211 Chr.8: 132062331-132062331 on Build GRCh38
rs746391173 Chr.8: 132062712-132062712 on Build GRCh38
rs79450420 Chr.8: 132062509-132062509 on Build GRCh38
rs995692670 Chr.8: 132062547-132062547 on Build GRCh38
rs896133352 Chr.8: 132062238-132062238 on Build GRCh38
rs553620456 Chr.8: 132062662-132062662 on Build GRCh38
rs778892884 Chr.8: 132062500-132062500 on Build GRCh38
rs926731531 Chr.8: 132062674-132062674 on Build GRCh38
rs992319824 Chr.8: 132062715-132062715 on Build GRCh38
rs76608695 Chr.8: 132062230-132062230 on Build GRCh38
rs974356194 Chr.8: 132062692-132062692 on Build GRCh38
rs777342457 Chr.8: 132062645-132062645 on Build GRCh38
rs567927926 Chr.8: 132062646-132062646 on Build GRCh38
rs184490995 Chr.8: 132062668-132062668 on Build GRCh38
rs142763532 Chr.8: 132062654-132062654 on Build GRCh38
rs115677362 Chr.8: 132062304-132062304 on Build GRCh38
rs1028084763 Chr.8: 132062551-132062551 on Build GRCh38
rs946816698 Chr.8: 132062406-132062406 on Build GRCh38
rs182297821 Chr.8: 132062537-132062537 on Build GRCh38
rs965993849 Chr.8: 132062332-132062332 on Build GRCh38
rs995349670 Chr.8: 132062303-132062303 on Build GRCh38
rs770670882 Chr.8: 132062465-132062465 on Build GRCh38
rs3802262 Chr.8: 132062464-132062464 on Build GRCh38
rs918771301 Chr.8: 132062427-132062427 on Build GRCh38
rs747141680 Chr.8: 132062456-132062457 on Build GRCh38
rs75455660 Chr.8: 132062414-132062414 on Build GRCh38
rs988579648 Chr.8: 132062244-132062244 on Build GRCh38
rs1010393165 Chr.8: 132062605-132062605 on Build GRCh38
rs930195760 Chr.8: 132062717-132062717 on Build GRCh38
rs148237691 Chr.8: 132062386-132062386 on Build GRCh38
rs1000123705 Chr.8: 132062567-132062567 on Build GRCh38
rs771456848 Chr.8: 132062681-132062681 on Build GRCh38
rs190472018 Chr.8: 132062462-132062462 on Build GRCh38
rs941640229 Chr.8: 132062368-132062368 on Build GRCh38
rs900397140 Chr.8: 132062564-132062564 on Build GRCh38
rs1038916290 Chr.8: 132062411-132062411 on Build GRCh38

View Full Product Details