Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 2836560-2837060 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100507424
Entrez Gene ID
100507424
Gene Name
uncharacterized LOC100507424
Gene Aliases
-
UniGene
Hs.239
Gene Chromosome Location
Chr.12: 2836816-2859795 on Build GRCh38
Transcripts
NR_038933.1

SNP Detail

SNP ID Chromosome Location
rs930385260 Chr.12: 2836606-2836606 on Build GRCh38
rs544595375 Chr.12: 2836722-2836722 on Build GRCh38
rs1048837125 Chr.12: 2836611-2836611 on Build GRCh38
rs11062374 Chr.12: 2836768-2836768 on Build GRCh38
rs1013283822 Chr.12: 2837010-2837010 on Build GRCh38
rs943421311 Chr.12: 2836789-2836789 on Build GRCh38
rs906909552 Chr.12: 2836923-2836923 on Build GRCh38
rs149789945 Chr.12: 2837025-2837025 on Build GRCh38
rs144174822 Chr.12: 2836805-2836805 on Build GRCh38
rs564283715 Chr.12: 2836731-2836731 on Build GRCh38
rs993407926 Chr.12: 2836630-2836630 on Build GRCh38
rs1033862405 Chr.12: 2836673-2836673 on Build GRCh38
rs977645382 Chr.12: 2836766-2836767 on Build GRCh38
rs569973781 Chr.12: 2836680-2836680 on Build GRCh38
rs966356484 Chr.12: 2837031-2837031 on Build GRCh38
rs527584806 Chr.12: 2836877-2836877 on Build GRCh38
rs550562672 Chr.12: 2836969-2836969 on Build GRCh38
rs1006488367 Chr.12: 2836903-2836906 on Build GRCh38
rs770566298 Chr.12: 2836730-2836730 on Build GRCh38
rs887675217 Chr.12: 2836894-2836894 on Build GRCh38
rs892611553 Chr.12: 2836975-2836975 on Build GRCh38
rs1048117285 Chr.12: 2836875-2836875 on Build GRCh38
rs529432192 Chr.12: 2836784-2836784 on Build GRCh38
rs939578394 Chr.12: 2836965-2836965 on Build GRCh38
rs538111038 Chr.12: 2836870-2836870 on Build GRCh38
rs544394873 Chr.12: 2836654-2836654 on Build GRCh38
rs892671311 Chr.12: 2836671-2836671 on Build GRCh38
rs573051541 Chr.12: 2836648-2836648 on Build GRCh38
rs930521394 Chr.12: 2836770-2836770 on Build GRCh38
rs1045430404 Chr.12: 2836848-2836848 on Build GRCh38
rs573114010 Chr.12: 2837027-2837027 on Build GRCh38
rs552144468 Chr.12: 2836895-2836895 on Build GRCh38
rs1022774098 Chr.12: 2836997-2836997 on Build GRCh38
rs561952101 Chr.12: 2836670-2836670 on Build GRCh38
rs901843768 Chr.12: 2836810-2836810 on Build GRCh38
rs545542799 Chr.12: 2836612-2836612 on Build GRCh38
rs948435079 Chr.12: 2836613-2836613 on Build GRCh38
rs146253559 Chr.12: 2836749-2836749 on Build GRCh38
rs562691972 Chr.12: 2836757-2836757 on Build GRCh38
rs1033503584 Chr.12: 2836977-2836977 on Build GRCh38
rs1006021726 Chr.12: 2836669-2836669 on Build GRCh38
rs906795046 Chr.12: 2836621-2836621 on Build GRCh38
rs1011093412 Chr.12: 2836994-2836994 on Build GRCh38
rs4765999 Chr.12: 2836804-2836804 on Build GRCh38
rs150603259 Chr.12: 2837011-2837011 on Build GRCh38
rs984927339 Chr.12: 2836786-2836786 on Build GRCh38
rs953481211 Chr.12: 2836686-2836686 on Build GRCh38
rs915680534 Chr.12: 2836790-2836790 on Build GRCh38
rs986107798 Chr.12: 2836714-2836714 on Build GRCh38
rs1045316269 Chr.12: 2836620-2836620 on Build GRCh38
rs118184834 Chr.12: 2836647-2836647 on Build GRCh38
rs943370256 Chr.12: 2836599-2836599 on Build GRCh38
rs139698200 Chr.12: 2837015-2837015 on Build GRCh38
rs971659971 Chr.12: 2836763-2836763 on Build GRCh38
rs894920392 Chr.12: 2837006-2837006 on Build GRCh38
rs749045677 Chr.12: 2836682-2836682 on Build GRCh38
rs548578337 Chr.12: 2836769-2836769 on Build GRCh38
rs929254899 Chr.12: 2836863-2836863 on Build GRCh38

View Full Product Details