Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 67740292-67740795 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOL11
Entrez Gene ID
25926
Gene Name
nucleolar protein 11
Gene Aliases
-
UniGene
Hs.463936
Gene Chromosome Location
Chr.17: 67717833-67744208 on Build GRCh38
Transcripts
NM_001303272.1 NM_015462.4 NR_003706.2

Gene Symbol
SNORA38B
Entrez Gene ID
100124536
Gene Name
small nucleolar RNA, H/ACA box 38B
Gene Aliases
-
UniGene
Hs.693446
Gene Chromosome Location
Chr.17: 67740670-67740801 on Build GRCh38
Transcripts
NM_001303272.1 NM_015462.4 NR_003706.2

SNP Detail

SNP ID Chromosome Location
rs967813854 Chr.17: 67740622-67740623 on Build GRCh38
rs1023280355 Chr.17: 67740601-67740601 on Build GRCh38
rs369314445 Chr.17: 67740609-67740609 on Build GRCh38
rs372271564 Chr.17: 67740771-67740771 on Build GRCh38
rs557941055 Chr.17: 67740473-67740473 on Build GRCh38
rs914941336 Chr.17: 67740384-67740384 on Build GRCh38
rs1008158461 Chr.17: 67740334-67740334 on Build GRCh38
rs1011879149 Chr.17: 67740321-67740321 on Build GRCh38
rs894694444 Chr.17: 67740585-67740585 on Build GRCh38
rs963914742 Chr.17: 67740361-67740361 on Build GRCh38
rs998853187 Chr.17: 67740315-67740315 on Build GRCh38
rs916032861 Chr.17: 67740559-67740559 on Build GRCh38
rs561966590 Chr.17: 67740530-67740530 on Build GRCh38
rs1018500787 Chr.17: 67740348-67740348 on Build GRCh38
rs967761479 Chr.17: 67740338-67740338 on Build GRCh38
rs975406088 Chr.17: 67740715-67740715 on Build GRCh38
rs77124258 Chr.17: 67740333-67740333 on Build GRCh38
rs1039647758 Chr.17: 67740485-67740486 on Build GRCh38
rs947616284 Chr.17: 67740625-67740625 on Build GRCh38
rs150579812 Chr.17: 67740696-67740696 on Build GRCh38
rs142283623 Chr.17: 67740662-67740662 on Build GRCh38
rs956863366 Chr.17: 67740427-67740427 on Build GRCh38
rs1047137032 Chr.17: 67740760-67740760 on Build GRCh38
rs202021341 Chr.17: 67740610-67740610 on Build GRCh38
rs938911018 Chr.17: 67740549-67740549 on Build GRCh38
rs530305668 Chr.17: 67740672-67740672 on Build GRCh38
rs546023864 Chr.17: 67740455-67740455 on Build GRCh38
rs974194777 Chr.17: 67740362-67740362 on Build GRCh38
rs748951614 Chr.17: 67740639-67740642 on Build GRCh38
rs748598705 Chr.17: 67740587-67740587 on Build GRCh38
rs982467050 Chr.17: 67740485-67740485 on Build GRCh38
rs11439892 Chr.17: 67740609-67740610 on Build GRCh38
rs1043119952 Chr.17: 67740657-67740657 on Build GRCh38
rs931429049 Chr.17: 67740752-67740752 on Build GRCh38
rs13341463 Chr.17: 67740443-67740443 on Build GRCh38
rs397704864 Chr.17: 67740624-67740625 on Build GRCh38
rs540516277 Chr.17: 67740517-67740517 on Build GRCh38
rs140038057 Chr.17: 67740534-67740534 on Build GRCh38
rs562647805 Chr.17: 67740595-67740595 on Build GRCh38
rs749124818 Chr.17: 67740447-67740447 on Build GRCh38
rs942708585 Chr.17: 67740448-67740448 on Build GRCh38
rs919707136 Chr.17: 67740399-67740399 on Build GRCh38
rs745336909 Chr.17: 67740767-67740767 on Build GRCh38
rs963901213 Chr.17: 67740701-67740701 on Build GRCh38
rs573212400 Chr.17: 67740494-67740494 on Build GRCh38
rs987000266 Chr.17: 67740664-67740664 on Build GRCh38
rs887271910 Chr.17: 67740539-67740539 on Build GRCh38
rs919911086 Chr.17: 67740745-67740745 on Build GRCh38
rs1057379462 Chr.17: 67740580-67740580 on Build GRCh38
rs866946545 Chr.17: 67740550-67740550 on Build GRCh38
rs1030859495 Chr.17: 67740328-67740328 on Build GRCh38
rs573996832 Chr.17: 67740533-67740533 on Build GRCh38

View Full Product Details