Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 12967836-12968383 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100506474
Entrez Gene ID
100506474
Gene Name
uncharacterized LOC100506474
Gene Aliases
-
UniGene
Hs.242196
Gene Chromosome Location
Chr.2: 12966782-13007013 on Build GRCh38
Transcripts
NR_038434.1

SNP Detail

SNP ID Chromosome Location
rs374924594 Chr.2: 12967938-12967938 on Build GRCh38
rs190094929 Chr.2: 12968300-12968300 on Build GRCh38
rs982601938 Chr.2: 12968049-12968049 on Build GRCh38
rs538863261 Chr.2: 12968077-12968077 on Build GRCh38
rs369653367 Chr.2: 12968347-12968347 on Build GRCh38
rs533932925 Chr.2: 12967968-12967968 on Build GRCh38
rs947856623 Chr.2: 12968352-12968352 on Build GRCh38
rs915582487 Chr.2: 12968097-12968097 on Build GRCh38
rs752371239 Chr.2: 12968117-12968117 on Build GRCh38
rs1010984607 Chr.2: 12967942-12967942 on Build GRCh38
rs1017143400 Chr.2: 12968179-12968179 on Build GRCh38
rs899841392 Chr.2: 12968211-12968211 on Build GRCh38
rs571175710 Chr.2: 12967927-12967927 on Build GRCh38
rs976701731 Chr.2: 12967987-12967987 on Build GRCh38
rs548180465 Chr.2: 12967952-12967952 on Build GRCh38
rs964369288 Chr.2: 12967971-12967971 on Build GRCh38
rs539823894 Chr.2: 12968234-12968234 on Build GRCh38
rs991262221 Chr.2: 12968267-12968267 on Build GRCh38
rs924663161 Chr.2: 12967961-12967961 on Build GRCh38
rs566488160 Chr.2: 12967958-12967958 on Build GRCh38
rs13398738 Chr.2: 12968078-12968078 on Build GRCh38
rs951295498 Chr.2: 12968038-12968038 on Build GRCh38
rs1018289938 Chr.2: 12967949-12967949 on Build GRCh38
rs964533698 Chr.2: 12968193-12968193 on Build GRCh38
rs146994073 Chr.2: 12968162-12968162 on Build GRCh38
rs111809810 Chr.2: 12968085-12968085 on Build GRCh38
rs12994713 Chr.2: 12968125-12968125 on Build GRCh38
rs906216136 Chr.2: 12968120-12968120 on Build GRCh38
rs990178408 Chr.2: 12968282-12968282 on Build GRCh38
rs138124863 Chr.2: 12968278-12968278 on Build GRCh38
rs542700585 Chr.2: 12968107-12968107 on Build GRCh38
rs949717163 Chr.2: 12968103-12968103 on Build GRCh38
rs563796713 Chr.2: 12968034-12968034 on Build GRCh38
rs998630437 Chr.2: 12968228-12968228 on Build GRCh38
rs955410993 Chr.2: 12968242-12968242 on Build GRCh38
rs72769634 Chr.2: 12968328-12968328 on Build GRCh38
rs909704331 Chr.2: 12968053-12968053 on Build GRCh38
rs565604554 Chr.2: 12968260-12968260 on Build GRCh38
rs893951963 Chr.2: 12968160-12968160 on Build GRCh38
rs922643774 Chr.2: 12967879-12967879 on Build GRCh38
rs928241158 Chr.2: 12968109-12968109 on Build GRCh38
rs764908335 Chr.2: 12968174-12968174 on Build GRCh38
rs951327763 Chr.2: 12968241-12968241 on Build GRCh38
rs1040155455 Chr.2: 12968175-12968175 on Build GRCh38
rs750089016 Chr.2: 12968246-12968246 on Build GRCh38
rs1053585538 Chr.2: 12968022-12968022 on Build GRCh38
rs571431667 Chr.2: 12968031-12968031 on Build GRCh38
rs1016741373 Chr.2: 12968259-12968259 on Build GRCh38
rs186824716 Chr.2: 12968030-12968030 on Build GRCh38
rs1030025622 Chr.2: 12968003-12968003 on Build GRCh38
rs935551952 Chr.2: 12968118-12968118 on Build GRCh38
rs1031174015 Chr.2: 12967928-12967928 on Build GRCh38

View Full Product Details