Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42867034-42867583 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AOC4P
Entrez Gene ID
90586
Gene Name
amine oxidase, copper containing 4, pseudogene
Gene Aliases
AOC4, UPAT
UniGene
Hs.367781
Gene Chromosome Location
Chr.17: 42867145-42869217 on Build GRCh38
Transcripts
NR_002773.1

SNP Detail

SNP ID Chromosome Location
rs544918973 Chr.17: 42867100-42867100 on Build GRCh38
rs140682150 Chr.17: 42867077-42867077 on Build GRCh38
rs767207226 Chr.17: 42867342-42867342 on Build GRCh38
rs527358581 Chr.17: 42867260-42867260 on Build GRCh38
rs756342456 Chr.17: 42867550-42867550 on Build GRCh38
rs747124204 Chr.17: 42867381-42867381 on Build GRCh38
rs757345772 Chr.17: 42867476-42867476 on Build GRCh38
rs749193874 Chr.17: 42867403-42867403 on Build GRCh38
rs769686722 Chr.17: 42867394-42867394 on Build GRCh38
rs760271653 Chr.17: 42867519-42867519 on Build GRCh38
rs767591348 Chr.17: 42867546-42867546 on Build GRCh38
rs573280960 Chr.17: 42867060-42867060 on Build GRCh38
rs750548735 Chr.17: 42867548-42867548 on Build GRCh38
rs766276427 Chr.17: 42867479-42867479 on Build GRCh38
rs765640961 Chr.17: 42867358-42867358 on Build GRCh38
rs771543365 Chr.17: 42867422-42867422 on Build GRCh38
rs760361497 Chr.17: 42867435-42867435 on Build GRCh38
rs764162252 Chr.17: 42867320-42867320 on Build GRCh38
rs1047679495 Chr.17: 42867343-42867343 on Build GRCh38
rs775156050 Chr.17: 42867314-42867314 on Build GRCh38
rs770584278 Chr.17: 42867521-42867521 on Build GRCh38
rs755324177 Chr.17: 42867349-42867349 on Build GRCh38
rs980669112 Chr.17: 42867075-42867075 on Build GRCh38
rs775462818 Chr.17: 42867400-42867400 on Build GRCh38
rs780422474 Chr.17: 42867551-42867551 on Build GRCh38
rs773672296 Chr.17: 42867417-42867417 on Build GRCh38
rs774319172 Chr.17: 42867326-42867326 on Build GRCh38
rs150122707 Chr.17: 42867329-42867329 on Build GRCh38
rs1024728730 Chr.17: 42867059-42867059 on Build GRCh38
rs772900821 Chr.17: 42867430-42867430 on Build GRCh38
rs988189084 Chr.17: 42867139-42867139 on Build GRCh38
rs943517304 Chr.17: 42867261-42867261 on Build GRCh38
rs73983764 Chr.17: 42867056-42867056 on Build GRCh38
rs577715805 Chr.17: 42867171-42867171 on Build GRCh38
rs752070681 Chr.17: 42867473-42867473 on Build GRCh38
rs571165748 Chr.17: 42867464-42867464 on Build GRCh38
rs370183495 Chr.17: 42867195-42867195 on Build GRCh38
rs774147982 Chr.17: 42867558-42867559 on Build GRCh38
rs771305037 Chr.17: 42867240-42867240 on Build GRCh38
rs969184592 Chr.17: 42867072-42867072 on Build GRCh38
rs563148672 Chr.17: 42867108-42867108 on Build GRCh38
rs530217650 Chr.17: 42867481-42867481 on Build GRCh38
rs111992231 Chr.17: 42867375-42867375 on Build GRCh38
rs555909270 Chr.17: 42867082-42867082 on Build GRCh38
rs561507579 Chr.17: 42867346-42867346 on Build GRCh38
rs560762415 Chr.17: 42867278-42867278 on Build GRCh38
rs771578591 Chr.17: 42867405-42867405 on Build GRCh38
rs548939350 Chr.17: 42867263-42867263 on Build GRCh38
rs775764108 Chr.17: 42867524-42867524 on Build GRCh38
rs781471083 Chr.17: 42867392-42867392 on Build GRCh38
rs574422330 Chr.17: 42867092-42867092 on Build GRCh38
rs750638819 Chr.17: 42867482-42867482 on Build GRCh38
rs763393501 Chr.17: 42867466-42867466 on Build GRCh38
rs933525953 Chr.17: 42867192-42867192 on Build GRCh38
rs753173022 Chr.17: 42867367-42867367 on Build GRCh38
rs757353645 Chr.17: 42867383-42867383 on Build GRCh38
rs899214366 Chr.17: 42867324-42867324 on Build GRCh38
rs534287244 Chr.17: 42867526-42867526 on Build GRCh38
rs752081319 Chr.17: 42867538-42867538 on Build GRCh38
rs544979351 Chr.17: 42867186-42867186 on Build GRCh38
rs749104332 Chr.17: 42867493-42867493 on Build GRCh38
rs985814191 Chr.17: 42867446-42867446 on Build GRCh38
rs897631492 Chr.17: 42867202-42867205 on Build GRCh38
rs551751961 Chr.17: 42867486-42867486 on Build GRCh38
rs986210076 Chr.17: 42867151-42867151 on Build GRCh38
rs771581129 Chr.17: 42867507-42867507 on Build GRCh38
rs762507089 Chr.17: 42867317-42867317 on Build GRCh38
rs747373753 Chr.17: 42867230-42867230 on Build GRCh38
rs764422797 Chr.17: 42867531-42867531 on Build GRCh38
rs750112380 Chr.17: 42867345-42867345 on Build GRCh38
rs748108756 Chr.17: 42867134-42867134 on Build GRCh38
rs758934917 Chr.17: 42867369-42867369 on Build GRCh38
rs765547018 Chr.17: 42867461-42867461 on Build GRCh38
rs938512523 Chr.17: 42867208-42867208 on Build GRCh38
rs779941371 Chr.17: 42867492-42867492 on Build GRCh38
rs549665500 Chr.17: 42867404-42867404 on Build GRCh38
rs748087203 Chr.17: 42867503-42867503 on Build GRCh38
rs560035175 Chr.17: 42867217-42867217 on Build GRCh38
rs746147604 Chr.17: 42867393-42867393 on Build GRCh38
rs963086739 Chr.17: 42867469-42867469 on Build GRCh38
rs758383948 Chr.17: 42867145-42867145 on Build GRCh38
rs28691051 Chr.17: 42867544-42867544 on Build GRCh38
rs764665755 Chr.17: 42867468-42867468 on Build GRCh38
rs1036182826 Chr.17: 42867196-42867196 on Build GRCh38
rs28661937 Chr.17: 42867508-42867508 on Build GRCh38

View Full Product Details