Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 138838252-138838641 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECT2L
Entrez Gene ID
345930
Gene Name
epithelial cell transforming 2 like
Gene Aliases
ARHGEF32, C6orf91, FBXO49, LFDH, dJ509I19.2, dJ509I19.3, dJ509I19.5
UniGene
Hs.660603
Gene Chromosome Location
Chr.6: 138796107-138904070 on Build GRCh38
Transcripts
NM_001077706.2 NM_001195037.2 XM_006715472.3 XM_011535795.2 XM_011535797.2 XM_017010828.1 XM_017010829.1 XM_017010830.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1440817 c.257C>A p.S86Y Chr.6: 138838429-138838429 on Build GRCh38
4423139 c.281G>C p.S94T Chr.6: 138838453-138838453 on Build GRCh38
6876655 c.249C>T p.R83R Chr.6: 138838421-138838421 on Build GRCh38
6105521 c.241T>A p.L81I Chr.6: 138838413-138838413 on Build GRCh38
1204904 c.289G>A p.D97N Chr.6: 138838461-138838461 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747575140 Chr.6: 138838382-138838382 on Build GRCh38
rs754252750 Chr.6: 138838556-138838556 on Build GRCh38
rs768864069 Chr.6: 138838386-138838386 on Build GRCh38
rs371746776 Chr.6: 138838322-138838322 on Build GRCh38
rs751411325 Chr.6: 138838337-138838340 on Build GRCh38
rs755559534 Chr.6: 138838491-138838491 on Build GRCh38
rs971492945 Chr.6: 138838516-138838516 on Build GRCh38
rs1055108083 Chr.6: 138838286-138838286 on Build GRCh38
rs889950942 Chr.6: 138838574-138838574 on Build GRCh38
rs377144964 Chr.6: 138838559-138838559 on Build GRCh38
rs968041170 Chr.6: 138838333-138838333 on Build GRCh38
rs1022589061 Chr.6: 138838292-138838292 on Build GRCh38
rs762882490 Chr.6: 138838420-138838420 on Build GRCh38
rs759556864 Chr.6: 138838436-138838436 on Build GRCh38
rs762324898 Chr.6: 138838417-138838417 on Build GRCh38
rs367563166 Chr.6: 138838320-138838320 on Build GRCh38
rs752362246 Chr.6: 138838355-138838355 on Build GRCh38
rs116211453 Chr.6: 138838444-138838444 on Build GRCh38
rs199633256 Chr.6: 138838472-138838472 on Build GRCh38
rs757233322 Chr.6: 138838339-138838342 on Build GRCh38
rs772205101 Chr.6: 138838541-138838541 on Build GRCh38
rs748878656 Chr.6: 138838363-138838363 on Build GRCh38
rs376114360 Chr.6: 138838375-138838375 on Build GRCh38
rs762943354 Chr.6: 138838303-138838303 on Build GRCh38
rs777922360 Chr.6: 138838507-138838507 on Build GRCh38
rs543235657 Chr.6: 138838380-138838380 on Build GRCh38
rs777020972 Chr.6: 138838395-138838395 on Build GRCh38
rs933088230 Chr.6: 138838500-138838500 on Build GRCh38
rs754677600 Chr.6: 138838324-138838324 on Build GRCh38
rs756799483 Chr.6: 138838374-138838374 on Build GRCh38
rs936174623 Chr.6: 138838544-138838544 on Build GRCh38
rs748442558 Chr.6: 138838496-138838496 on Build GRCh38
rs373973518 Chr.6: 138838528-138838528 on Build GRCh38
rs764613811 Chr.6: 138838522-138838522 on Build GRCh38
rs1006578703 Chr.6: 138838411-138838411 on Build GRCh38
rs760493089 Chr.6: 138838550-138838550 on Build GRCh38
rs201725092 Chr.6: 138838495-138838495 on Build GRCh38
rs376602257 Chr.6: 138838482-138838482 on Build GRCh38
rs775575461 Chr.6: 138838549-138838549 on Build GRCh38
rs199661194 Chr.6: 138838419-138838419 on Build GRCh38
rs766159948 Chr.6: 138838306-138838306 on Build GRCh38
rs1007157184 Chr.6: 138838281-138838281 on Build GRCh38
rs756674110 Chr.6: 138838473-138838473 on Build GRCh38
rs913492781 Chr.6: 138838589-138838589 on Build GRCh38
rs748348446 Chr.6: 138838401-138838401 on Build GRCh38
rs990295223 Chr.6: 138838576-138838576 on Build GRCh38
rs150345992 Chr.6: 138838360-138838360 on Build GRCh38
rs766501979 Chr.6: 138838425-138838425 on Build GRCh38
rs749553023 Chr.6: 138838508-138838508 on Build GRCh38
rs1050742412 Chr.6: 138838592-138838592 on Build GRCh38
rs978143563 Chr.6: 138838337-138838338 on Build GRCh38
rs945705288 Chr.6: 138838321-138838321 on Build GRCh38
rs753393006 Chr.6: 138838566-138838566 on Build GRCh38
rs780504259 Chr.6: 138838523-138838523 on Build GRCh38
rs562150109 Chr.6: 138838457-138838457 on Build GRCh38
rs909156777 Chr.6: 138838466-138838466 on Build GRCh38
rs760403373 Chr.6: 138838456-138838456 on Build GRCh38
rs186827164 Chr.6: 138838591-138838591 on Build GRCh38
rs775866342 Chr.6: 138838525-138838525 on Build GRCh38
rs777412855 Chr.6: 138838362-138838362 on Build GRCh38
rs183610834 Chr.6: 138838396-138838396 on Build GRCh38
rs372385966 Chr.6: 138838433-138838433 on Build GRCh38
rs770157237 Chr.6: 138838502-138838502 on Build GRCh38
rs532802319 Chr.6: 138838511-138838511 on Build GRCh38
rs375451846 Chr.6: 138838440-138838440 on Build GRCh38
rs767373049 Chr.6: 138838352-138838352 on Build GRCh38

View Full Product Details