Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 46235217-46235478 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MCM3AP
Entrez Gene ID
8888
Gene Name
minichromosome maintenance complex component 3 associated protein
Gene Aliases
GANP, MAP80, SAC3
UniGene
Hs.389037
Gene Chromosome Location
Chr.21: 46235125-46286274 on Build GRCh38
Transcripts
NM_003906.4 XM_005261203.4 XM_005261204.4 XM_005261205.3 NR_002776.4 NR_110565.1 NR_110566.1 NR_110567.1

Gene Symbol
MCM3AP-AS1
Entrez Gene ID
114044
Gene Name
MCM3AP antisense RNA 1
Gene Aliases
C21orf85, MCM3AP-AS, MCM3APAS, MCM3APASB, NCRNA00031
UniGene
Hs.655095
Gene Chromosome Location
Chr.21: 46229231-46251701 on Build GRCh38
Transcripts
NM_003906.4 XM_005261203.4 XM_005261204.4 XM_005261205.3 NR_002776.4 NR_110565.1 NR_110566.1 NR_110567.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6867814 c.5932G>A p.V1978M Chr.21: 46235279-46235279 on Build GRCh38
6094810 c.5879C>T p.S1960L Chr.21: 46235332-46235332 on Build GRCh38
1682077 c.5845C>T p.R1949* Chr.21: 46235366-46235366 on Build GRCh38
6709832 c.5884G>T p.E1962* Chr.21: 46235327-46235327 on Build GRCh38
1414592 c.5803C>T p.Q1935* Chr.21: 46235408-46235408 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs562593941 Chr.21: 46235384-46235384 on Build GRCh38
rs745324820 Chr.21: 46235258-46235258 on Build GRCh38
rs528321869 Chr.21: 46235312-46235312 on Build GRCh38
rs199830023 Chr.21: 46235440-46235440 on Build GRCh38
rs373865756 Chr.21: 46235432-46235432 on Build GRCh38
rs141757607 Chr.21: 46235388-46235388 on Build GRCh38
rs763969969 Chr.21: 46235293-46235293 on Build GRCh38
rs577520453 Chr.21: 46235379-46235379 on Build GRCh38
rs771749901 Chr.21: 46235452-46235452 on Build GRCh38
rs141098252 Chr.21: 46235380-46235380 on Build GRCh38
rs924304133 Chr.21: 46235339-46235339 on Build GRCh38
rs780913938 Chr.21: 46235405-46235405 on Build GRCh38
rs891668251 Chr.21: 46235390-46235390 on Build GRCh38
rs763049470 Chr.21: 46235274-46235274 on Build GRCh38
rs559155265 Chr.21: 46235338-46235338 on Build GRCh38
rs201045045 Chr.21: 46235302-46235302 on Build GRCh38
rs780672309 Chr.21: 46235361-46235361 on Build GRCh38
rs374178711 Chr.21: 46235365-46235365 on Build GRCh38
rs765377965 Chr.21: 46235317-46235318 on Build GRCh38
rs756921773 Chr.21: 46235349-46235349 on Build GRCh38
rs756080303 Chr.21: 46235318-46235318 on Build GRCh38
rs17183403 Chr.21: 46235389-46235389 on Build GRCh38
rs755718483 Chr.21: 46235416-46235416 on Build GRCh38
rs554702135 Chr.21: 46235256-46235256 on Build GRCh38
rs766396220 Chr.21: 46235313-46235313 on Build GRCh38
rs745392476 Chr.21: 46235366-46235366 on Build GRCh38
rs574611806 Chr.21: 46235394-46235394 on Build GRCh38
rs765679894 Chr.21: 46235393-46235393 on Build GRCh38
rs754285154 Chr.21: 46235330-46235330 on Build GRCh38
rs984428261 Chr.21: 46235355-46235355 on Build GRCh38
rs761660668 Chr.21: 46235381-46235381 on Build GRCh38
rs112785693 Chr.21: 46235362-46235362 on Build GRCh38
rs746709172 Chr.21: 46235347-46235347 on Build GRCh38
rs750547964 Chr.21: 46235315-46235315 on Build GRCh38
rs371635864 Chr.21: 46235418-46235418 on Build GRCh38
rs749288099 Chr.21: 46235423-46235423 on Build GRCh38
rs766362107 Chr.21: 46235386-46235386 on Build GRCh38
rs766921331 Chr.21: 46235329-46235329 on Build GRCh38
rs960017364 Chr.21: 46235429-46235429 on Build GRCh38
rs775558146 Chr.21: 46235370-46235370 on Build GRCh38
rs541892506 Chr.21: 46235399-46235399 on Build GRCh38
rs747969243 Chr.21: 46235436-46235436 on Build GRCh38
rs762822817 Chr.21: 46235292-46235292 on Build GRCh38
rs200214494 Chr.21: 46235244-46235244 on Build GRCh38
rs146999256 Chr.21: 46235283-46235283 on Build GRCh38
rs767783662 Chr.21: 46235383-46235383 on Build GRCh38
rs779215140 Chr.21: 46235345-46235345 on Build GRCh38
rs201946782 Chr.21: 46235413-46235413 on Build GRCh38
rs138113935 Chr.21: 46235369-46235369 on Build GRCh38

View Full Product Details