Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 88057531-88057804 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPACA1
Entrez Gene ID
81833
Gene Name
sperm acrosome associated 1
Gene Aliases
SAMP32
UniGene
Hs.161241
Gene Chromosome Location
Chr.6: 88046907-88066832 on Build GRCh38
Transcripts
NM_030960.2 XM_011536160.2 XM_017011335.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1292377 c.331C>T p.R111W Chr.6: 88057677-88057677 on Build GRCh38
1697623 c.364G>A p.G122S Chr.6: 88057710-88057710 on Build GRCh38
129369 c.319A>C p.K107Q Chr.6: 88057665-88057665 on Build GRCh38
7166547 c.362G>A p.C121Y Chr.6: 88057708-88057708 on Build GRCh38
5763230 c.352C>A p.P118T Chr.6: 88057698-88057698 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776492466 Chr.6: 88057728-88057728 on Build GRCh38
rs987521169 Chr.6: 88057696-88057696 on Build GRCh38
rs750028950 Chr.6: 88057646-88057646 on Build GRCh38
rs770914140 Chr.6: 88057615-88057615 on Build GRCh38
rs145923815 Chr.6: 88057682-88057682 on Build GRCh38
rs895628380 Chr.6: 88057558-88057558 on Build GRCh38
rs1013544111 Chr.6: 88057579-88057579 on Build GRCh38
rs756425291 Chr.6: 88057691-88057691 on Build GRCh38
rs779127049 Chr.6: 88057718-88057718 on Build GRCh38
rs201142491 Chr.6: 88057742-88057742 on Build GRCh38
rs777804035 Chr.6: 88057594-88057594 on Build GRCh38
rs770203131 Chr.6: 88057590-88057590 on Build GRCh38
rs765093783 Chr.6: 88057645-88057645 on Build GRCh38
rs759062780 Chr.6: 88057747-88057747 on Build GRCh38
rs745906077 Chr.6: 88057721-88057721 on Build GRCh38
rs922971157 Chr.6: 88057688-88057688 on Build GRCh38
rs910636662 Chr.6: 88057714-88057714 on Build GRCh38
rs779671948 Chr.6: 88057566-88057566 on Build GRCh38
rs373479619 Chr.6: 88057744-88057744 on Build GRCh38
rs556349892 Chr.6: 88057595-88057595 on Build GRCh38
rs771231302 Chr.6: 88057712-88057712 on Build GRCh38
rs781770276 Chr.6: 88057580-88057580 on Build GRCh38
rs756688735 Chr.6: 88057762-88057762 on Build GRCh38
rs142673280 Chr.6: 88057616-88057616 on Build GRCh38
rs572905991 Chr.6: 88057743-88057743 on Build GRCh38
rs761868325 Chr.6: 88057729-88057729 on Build GRCh38
rs370510634 Chr.6: 88057683-88057683 on Build GRCh38
rs768527680 Chr.6: 88057624-88057624 on Build GRCh38
rs778269594 Chr.6: 88057698-88057698 on Build GRCh38
rs760580332 Chr.6: 88057619-88057619 on Build GRCh38
rs765785735 Chr.6: 88057676-88057676 on Build GRCh38
rs762732429 Chr.6: 88057663-88057663 on Build GRCh38
rs752082389 Chr.6: 88057571-88057571 on Build GRCh38
rs954844868 Chr.6: 88057693-88057693 on Build GRCh38
rs557931381 Chr.6: 88057704-88057704 on Build GRCh38
rs1033040477 Chr.6: 88057639-88057639 on Build GRCh38
rs987054701 Chr.6: 88057769-88057769 on Build GRCh38
rs767196661 Chr.6: 88057749-88057749 on Build GRCh38
rs969590084 Chr.6: 88057611-88057611 on Build GRCh38
rs748484266 Chr.6: 88057588-88057588 on Build GRCh38
rs761528597 Chr.6: 88057634-88057634 on Build GRCh38
rs200011030 Chr.6: 88057746-88057746 on Build GRCh38
rs943400926 Chr.6: 88057734-88057734 on Build GRCh38
rs1021094748 Chr.6: 88057692-88057692 on Build GRCh38
rs769471113 Chr.6: 88057730-88057730 on Build GRCh38
rs757787364 Chr.6: 88057564-88057564 on Build GRCh38
rs772140087 Chr.6: 88057723-88057723 on Build GRCh38
rs776577127 Chr.6: 88057633-88057633 on Build GRCh38
rs369745181 Chr.6: 88057678-88057678 on Build GRCh38
rs755594093 Chr.6: 88057574-88057574 on Build GRCh38
rs751066460 Chr.6: 88057677-88057677 on Build GRCh38
rs753255602 Chr.6: 88057752-88057752 on Build GRCh38

View Full Product Details