Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 46919134-46919405 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP35
Entrez Gene ID
2909
Gene Name
Rho GTPase activating protein 35
Gene Aliases
GRF-1, GRLF1, P190-A, P190A, p190ARhoGAP, p190RhoGAP
UniGene
Hs.509447
Gene Chromosome Location
Chr.19: 46861002-47005077 on Build GRCh38
Transcripts
NM_004491.4 XM_011526873.2 XM_017026714.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3105783 c.609C>T p.D203D Chr.19: 46919284-46919284 on Build GRCh38
998534 c.595_598delCTGA p.T200fs*18 Chr.19: 46919270-46919273 on Build GRCh38
4656906 c.579G>T p.K193N Chr.19: 46919254-46919254 on Build GRCh38
1394824 c.706G>A p.V236M Chr.19: 46919381-46919381 on Build GRCh38
5856239 c.650C>T p.A217V Chr.19: 46919325-46919325 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770319490 Chr.19: 46919165-46919165 on Build GRCh38
rs752976959 Chr.19: 46919284-46919284 on Build GRCh38
rs759686905 Chr.19: 46919266-46919266 on Build GRCh38
rs369569752 Chr.19: 46919380-46919380 on Build GRCh38
rs921963889 Chr.19: 46919272-46919272 on Build GRCh38
rs775346521 Chr.19: 46919207-46919207 on Build GRCh38
rs200079500 Chr.19: 46919220-46919220 on Build GRCh38
rs189377450 Chr.19: 46919230-46919230 on Build GRCh38
rs549628211 Chr.19: 46919373-46919373 on Build GRCh38
rs376317356 Chr.19: 46919365-46919365 on Build GRCh38
rs367587187 Chr.19: 46919200-46919200 on Build GRCh38
rs201226801 Chr.19: 46919223-46919223 on Build GRCh38
rs766321071 Chr.19: 46919232-46919232 on Build GRCh38
rs760530208 Chr.19: 46919209-46919209 on Build GRCh38
rs778151791 Chr.19: 46919341-46919341 on Build GRCh38
rs1053684230 Chr.19: 46919259-46919259 on Build GRCh38
rs971340341 Chr.19: 46919202-46919202 on Build GRCh38
rs751670981 Chr.19: 46919251-46919251 on Build GRCh38
rs771867647 Chr.19: 46919203-46919203 on Build GRCh38
rs754262695 Chr.19: 46919344-46919344 on Build GRCh38
rs745390841 Chr.19: 46919374-46919374 on Build GRCh38
rs767723487 Chr.19: 46919269-46919269 on Build GRCh38
rs200894233 Chr.19: 46919179-46919179 on Build GRCh38
rs371996167 Chr.19: 46919315-46919315 on Build GRCh38
rs1040337182 Chr.19: 46919197-46919197 on Build GRCh38
rs756448377 Chr.19: 46919340-46919340 on Build GRCh38

View Full Product Details