Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 76271555-76271796 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM47E
Entrez Gene ID
100129583
Gene Name
family with sequence similarity 47 member E
Gene Aliases
-
UniGene
Hs.720509
Gene Chromosome Location
Chr.4: 76214040-76283783 on Build GRCh38
Transcripts
NM_001136570.2 NM_001242936.1 NM_001242939.1

Gene Symbol
FAM47E-STBD1
Entrez Gene ID
100631383
Gene Name
FAM47E-STBD1 readthrough
Gene Aliases
-
UniGene
Hs.720509
Gene Chromosome Location
Chr.4: 76251700-76311130 on Build GRCh38
Transcripts
NM_001136570.2 NM_001242936.1 NM_001242939.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5425368 c.523G>A p.E175K Chr.4: 76271715-76271715 on Build GRCh38
5401572 c.390C>T p.I130I Chr.4: 76271582-76271582 on Build GRCh38
1431076 c.755C>T p.T252M Chr.4: 76271653-76271653 on Build GRCh38
1057652 c.397G>T p.E133* Chr.4: 76271589-76271589 on Build GRCh38
2832123 c.728C>T p.T243I Chr.4: 76271626-76271626 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs975752078 Chr.4: 76271600-76271600 on Build GRCh38
rs948886965 Chr.4: 76271697-76271697 on Build GRCh38
rs963871498 Chr.4: 76271582-76271582 on Build GRCh38
rs960123621 Chr.4: 76271683-76271683 on Build GRCh38
rs916083372 Chr.4: 76271689-76271689 on Build GRCh38
rs534203291 Chr.4: 76271764-76271764 on Build GRCh38
rs977254577 Chr.4: 76271660-76271660 on Build GRCh38
rs754205276 Chr.4: 76271688-76271688 on Build GRCh38
rs574751211 Chr.4: 76271723-76271723 on Build GRCh38
rs755809691 Chr.4: 76271693-76271693 on Build GRCh38
rs1019872926 Chr.4: 76271581-76271581 on Build GRCh38
rs1039731373 Chr.4: 76271740-76271740 on Build GRCh38
rs977392038 Chr.4: 76271655-76271655 on Build GRCh38
rs774617220 Chr.4: 76271604-76271604 on Build GRCh38
rs772084828 Chr.4: 76271619-76271619 on Build GRCh38
rs769457260 Chr.4: 76271646-76271646 on Build GRCh38
rs3733251 Chr.4: 76271685-76271685 on Build GRCh38
rs924496883 Chr.4: 76271667-76271667 on Build GRCh38
rs375151279 Chr.4: 76271637-76271637 on Build GRCh38
rs768804874 Chr.4: 76271596-76271596 on Build GRCh38
rs191481797 Chr.4: 76271639-76271639 on Build GRCh38
rs949830253 Chr.4: 76271654-76271654 on Build GRCh38
rs202038650 Chr.4: 76271678-76271678 on Build GRCh38
rs371976589 Chr.4: 76271583-76271583 on Build GRCh38
rs761862662 Chr.4: 76271613-76271613 on Build GRCh38
rs3733250 Chr.4: 76271715-76271715 on Build GRCh38
rs1021511555 Chr.4: 76271609-76271609 on Build GRCh38
rs893820799 Chr.4: 76271711-76271711 on Build GRCh38
rs901269374 Chr.4: 76271775-76271775 on Build GRCh38
rs867217004 Chr.4: 76271653-76271653 on Build GRCh38

View Full Product Details