Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 174522384-174522623 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HPGD
Entrez Gene ID
3248
Gene Name
hydroxyprostaglandin dehydrogenase 15-(NAD)
Gene Aliases
15-PGDH, PGDH, PGDH1, PHOAR1, SDR36C1
UniGene
Hs.596913
Gene Chromosome Location
Chr.4: 174490177-174522898 on Build GRCh38
Transcripts
NM_000860.5 NM_001145816.2 NM_001256301.1 NM_001256305.1 NM_001256306.1 NM_001256307.1 XM_011531907.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6848790 c.20T>G p.V7G Chr.4: 174522432-174522432 on Build GRCh38
6405353 c.23C>T p.A8V Chr.4: 174522429-174522429 on Build GRCh38
5060252 c.44A>G p.Q15R Chr.4: 174522408-174522408 on Build GRCh38
244949 c.60C>G p.A20A Chr.4: 174522392-174522392 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs886059256 Chr.4: 174522574-174522574 on Build GRCh38
rs901973274 Chr.4: 174522533-174522533 on Build GRCh38
rs991380566 Chr.4: 174522565-174522565 on Build GRCh38
rs558988612 Chr.4: 174522537-174522537 on Build GRCh38
rs1054368444 Chr.4: 174522504-174522504 on Build GRCh38
rs577045722 Chr.4: 174522451-174522451 on Build GRCh38
rs748329924 Chr.4: 174522478-174522478 on Build GRCh38
rs557149774 Chr.4: 174522405-174522405 on Build GRCh38
rs936056403 Chr.4: 174522491-174522491 on Build GRCh38
rs1005244591 Chr.4: 174522557-174522557 on Build GRCh38
rs1040540365 Chr.4: 174522522-174522522 on Build GRCh38
rs1052813362 Chr.4: 174522531-174522531 on Build GRCh38
rs773544789 Chr.4: 174522496-174522496 on Build GRCh38
rs938395873 Chr.4: 174522525-174522526 on Build GRCh38
rs1025424253 Chr.4: 174522545-174522545 on Build GRCh38
rs748049663 Chr.4: 174522590-174522590 on Build GRCh38
rs770044314 Chr.4: 174522494-174522494 on Build GRCh38
rs891596704 Chr.4: 174522535-174522535 on Build GRCh38
rs1001305547 Chr.4: 174522556-174522556 on Build GRCh38
rs776797460 Chr.4: 174522473-174522473 on Build GRCh38
rs768565455 Chr.4: 174522471-174522471 on Build GRCh38
rs958623993 Chr.4: 174522564-174522564 on Build GRCh38
rs777667720 Chr.4: 174522446-174522446 on Build GRCh38
rs917058294 Chr.4: 174522415-174522415 on Build GRCh38
rs917072096 Chr.4: 174522569-174522569 on Build GRCh38
rs971289358 Chr.4: 174522574-174522574 on Build GRCh38
rs868773592 Chr.4: 174522438-174522438 on Build GRCh38
rs763367344 Chr.4: 174522500-174522500 on Build GRCh38
rs1016596370 Chr.4: 174522559-174522559 on Build GRCh38
rs1044075961 Chr.4: 174522521-174522521 on Build GRCh38
rs986963526 Chr.4: 174522429-174522429 on Build GRCh38
rs961575704 Chr.4: 174522490-174522490 on Build GRCh38
rs889396738 Chr.4: 174522507-174522507 on Build GRCh38
rs778786044 Chr.4: 174522461-174522461 on Build GRCh38
rs45472095 Chr.4: 174522518-174522518 on Build GRCh38
rs745825366 Chr.4: 174522463-174522463 on Build GRCh38
rs755848982 Chr.4: 174522423-174522423 on Build GRCh38
rs1043346213 Chr.4: 174522544-174522544 on Build GRCh38
rs951200798 Chr.4: 174522553-174522553 on Build GRCh38

View Full Product Details