Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 154709486-154709759 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNJ3
Entrez Gene ID
3760
Gene Name
potassium voltage-gated channel subfamily J member 3
Gene Aliases
GIRK1, KGA, KIR3.1
UniGene
Hs.591606
Gene Chromosome Location
Chr.2: 154698581-154858352 on Build GRCh38
Transcripts
NM_001260508.1 NM_001260509.1 NM_002239.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3961323 c.856C>A p.R286R Chr.2: 154709756-154709756 on Build GRCh38
148958 c.703-8T>A p.? Chr.2: 154709595-154709595 on Build GRCh38
3568109 c.729C>T p.F243F Chr.2: 154709629-154709629 on Build GRCh38
1400230 c.790C>A p.L264I Chr.2: 154709690-154709690 on Build GRCh38
3299486 c.801C>T p.P267P Chr.2: 154709701-154709701 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201650481 Chr.2: 154709723-154709723 on Build GRCh38
rs559654341 Chr.2: 154709592-154709592 on Build GRCh38
rs866946714 Chr.2: 154709633-154709633 on Build GRCh38
rs1011981924 Chr.2: 154709559-154709559 on Build GRCh38
rs903749753 Chr.2: 154709591-154709591 on Build GRCh38
rs984689076 Chr.2: 154709692-154709692 on Build GRCh38
rs773276399 Chr.2: 154709571-154709571 on Build GRCh38
rs557478922 Chr.2: 154709534-154709534 on Build GRCh38
rs764893987 Chr.2: 154709704-154709704 on Build GRCh38
rs775178008 Chr.2: 154709594-154709594 on Build GRCh38
rs758872840 Chr.2: 154709711-154709711 on Build GRCh38
rs745623519 Chr.2: 154709676-154709677 on Build GRCh38
rs116584454 Chr.2: 154709540-154709540 on Build GRCh38
rs752403034 Chr.2: 154709671-154709671 on Build GRCh38
rs999374411 Chr.2: 154709677-154709677 on Build GRCh38
rs766872166 Chr.2: 154709716-154709716 on Build GRCh38
rs749931705 Chr.2: 154709705-154709705 on Build GRCh38
rs919965807 Chr.2: 154709575-154709575 on Build GRCh38
rs769799515 Chr.2: 154709563-154709563 on Build GRCh38
rs776353517 Chr.2: 154709668-154709668 on Build GRCh38
rs991982138 Chr.2: 154709568-154709568 on Build GRCh38
rs940535938 Chr.2: 154709736-154709736 on Build GRCh38
rs761550314 Chr.2: 154709695-154709695 on Build GRCh38
rs760468149 Chr.2: 154709599-154709599 on Build GRCh38
rs748436406 Chr.2: 154709727-154709727 on Build GRCh38
rs748241833 Chr.2: 154709562-154709562 on Build GRCh38
rs868672401 Chr.2: 154709654-154709654 on Build GRCh38
rs550218820 Chr.2: 154709583-154709583 on Build GRCh38
rs759441007 Chr.2: 154709578-154709578 on Build GRCh38
rs763849608 Chr.2: 154709600-154709600 on Build GRCh38
rs199712102 Chr.2: 154709561-154709561 on Build GRCh38
rs267598931 Chr.2: 154709722-154709722 on Build GRCh38

View Full Product Details