Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50625016-50625289 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARSA
Entrez Gene ID
410
Gene Name
arylsulfatase A
Gene Aliases
MLD
UniGene
Hs.731715
Gene Chromosome Location
Chr.22: 50622754-50628173 on Build GRCh38
Transcripts
NM_000487.5 NM_001085425.2 NM_001085426.2 NM_001085427.2 NM_001085428.2 XM_011530690.2 XM_011530691.2 XM_017028800.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3555837 c.1428C>T p.P476P Chr.22: 50625247-50625247 on Build GRCh38
3992257 c.1486A>C p.T496P Chr.22: 50625189-50625189 on Build GRCh38
3555836 c.1512C>T p.C504C Chr.22: 50625163-50625163 on Build GRCh38
4582525 c.1408G>A p.A470T Chr.22: 50625267-50625267 on Build GRCh38
3405761 c.1506C>T p.C502C Chr.22: 50625169-50625169 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148352371 Chr.22: 50625228-50625228 on Build GRCh38
rs146160737 Chr.22: 50625042-50625042 on Build GRCh38
rs764345345 Chr.22: 50625159-50625159 on Build GRCh38
rs1017776709 Chr.22: 50625252-50625252 on Build GRCh38
rs767496539 Chr.22: 50625202-50625202 on Build GRCh38
rs1005640475 Chr.22: 50625101-50625101 on Build GRCh38
rs985877264 Chr.22: 50625240-50625240 on Build GRCh38
rs759357711 Chr.22: 50625229-50625229 on Build GRCh38
rs199476385 Chr.22: 50625263-50625263 on Build GRCh38
rs978574508 Chr.22: 50625260-50625260 on Build GRCh38
rs759058860 Chr.22: 50625131-50625131 on Build GRCh38
rs201085386 Chr.22: 50625188-50625188 on Build GRCh38
rs199476388 Chr.22: 50625204-50625204 on Build GRCh38
rs775857149 Chr.22: 50625136-50625136 on Build GRCh38
rs755974448 Chr.22: 50625207-50625207 on Build GRCh38
rs144323513 Chr.22: 50625195-50625195 on Build GRCh38
rs777458560 Chr.22: 50625219-50625219 on Build GRCh38
rs942357377 Chr.22: 50625052-50625052 on Build GRCh38
rs903106180 Chr.22: 50625220-50625220 on Build GRCh38
rs770665890 Chr.22: 50625122-50625122 on Build GRCh38
rs377187248 Chr.22: 50625130-50625130 on Build GRCh38
rs941712158 Chr.22: 50625055-50625055 on Build GRCh38
rs74315482 Chr.22: 50625213-50625213 on Build GRCh38
rs762111673 Chr.22: 50625265-50625265 on Build GRCh38
rs762056118 Chr.22: 50625175-50625175 on Build GRCh38
rs1038594113 Chr.22: 50625064-50625064 on Build GRCh38
rs761159278 Chr.22: 50625151-50625151 on Build GRCh38
rs772710219 Chr.22: 50625135-50625135 on Build GRCh38
rs769169905 Chr.22: 50625241-50625241 on Build GRCh38
rs777528466 Chr.22: 50625094-50625094 on Build GRCh38
rs377717242 Chr.22: 50625231-50625231 on Build GRCh38
rs1047241241 Chr.22: 50625123-50625123 on Build GRCh38
rs535795733 Chr.22: 50625084-50625084 on Build GRCh38
rs6151429 Chr.22: 50625049-50625049 on Build GRCh38
rs776884597 Chr.22: 50625253-50625253 on Build GRCh38
rs201846218 Chr.22: 50625143-50625143 on Build GRCh38
rs762361933 Chr.22: 50625040-50625040 on Build GRCh38
rs747443475 Chr.22: 50625234-50625234 on Build GRCh38
rs764962721 Chr.22: 50625268-50625268 on Build GRCh38
rs80338823 Chr.22: 50625258-50625268 on Build GRCh38
rs981169628 Chr.22: 50625128-50625128 on Build GRCh38
rs748857964 Chr.22: 50625108-50625108 on Build GRCh38
rs998027946 Chr.22: 50625081-50625081 on Build GRCh38
rs749061034 Chr.22: 50625225-50625225 on Build GRCh38
rs6151428 Chr.22: 50625182-50625182 on Build GRCh38
rs765452751 Chr.22: 50625267-50625267 on Build GRCh38
rs794727904 Chr.22: 50625257-50625267 on Build GRCh38
rs776116011 Chr.22: 50625233-50625233 on Build GRCh38
rs878992768 Chr.22: 50625183-50625183 on Build GRCh38
rs745438306 Chr.22: 50625230-50625230 on Build GRCh38
rs774153480 Chr.22: 50625182-50625183 on Build GRCh38
rs776953235 Chr.22: 50625168-50625168 on Build GRCh38
rs745450676 Chr.22: 50625205-50625205 on Build GRCh38

View Full Product Details