Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 8870337-8870598 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENO1
Entrez Gene ID
2023
Gene Name
enolase 1
Gene Aliases
ENO1L1, HEL-S-17, MPB1, NNE, PPH
UniGene
Hs.517145
Gene Chromosome Location
Chr.1: 8861000-8879092 on Build GRCh38
Transcripts
NM_001201483.1 NM_001428.3 XM_006710433.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1667119 c.221C>T p.A74V Chr.1: 8870471-8870471 on Build GRCh38
2258629 c.209A>G p.N70S Chr.1: 8870483-8870483 on Build GRCh38
7127378 c.196G>A p.V66I Chr.1: 8870496-8870496 on Build GRCh38
6432628 c.187T>C p.S63P Chr.1: 8870505-8870505 on Build GRCh38
4937378 c.240G>A p.K80K Chr.1: 8870452-8870452 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs56193077 Chr.1: 8870547-8870547 on Build GRCh38
rs766980625 Chr.1: 8870419-8870419 on Build GRCh38
rs746112581 Chr.1: 8870455-8870455 on Build GRCh38
rs1037336712 Chr.1: 8870563-8870563 on Build GRCh38
rs914039866 Chr.1: 8870475-8870475 on Build GRCh38
rs111311515 Chr.1: 8870450-8870450 on Build GRCh38
rs772803420 Chr.1: 8870524-8870524 on Build GRCh38
rs201513751 Chr.1: 8870488-8870488 on Build GRCh38
rs28999073 Chr.1: 8870517-8870517 on Build GRCh38
rs778235569 Chr.1: 8870451-8870451 on Build GRCh38
rs1002135002 Chr.1: 8870445-8870445 on Build GRCh38
rs778554927 Chr.1: 8870430-8870430 on Build GRCh38
rs752691268 Chr.1: 8870557-8870557 on Build GRCh38
rs774096595 Chr.1: 8870391-8870391 on Build GRCh38
rs747469478 Chr.1: 8870517-8870517 on Build GRCh38
rs201867667 Chr.1: 8870483-8870483 on Build GRCh38
rs767224734 Chr.1: 8870414-8870414 on Build GRCh38
rs762099234 Chr.1: 8870513-8870513 on Build GRCh38
rs372176939 Chr.1: 8870405-8870405 on Build GRCh38
rs146619828 Chr.1: 8870460-8870460 on Build GRCh38
rs781503761 Chr.1: 8870454-8870454 on Build GRCh38
rs769721191 Chr.1: 8870458-8870458 on Build GRCh38
rs938548623 Chr.1: 8870403-8870403 on Build GRCh38
rs191061261 Chr.1: 8870418-8870418 on Build GRCh38
rs541167224 Chr.1: 8870406-8870406 on Build GRCh38
rs28999075 Chr.1: 8870461-8870461 on Build GRCh38
rs759006817 Chr.1: 8870422-8870423 on Build GRCh38
rs753440595 Chr.1: 8870544-8870544 on Build GRCh38
rs765196468 Chr.1: 8870548-8870548 on Build GRCh38
rs767883418 Chr.1: 8870521-8870521 on Build GRCh38
rs1047586836 Chr.1: 8870373-8870373 on Build GRCh38
rs993337672 Chr.1: 8870363-8870363 on Build GRCh38
rs758216269 Chr.1: 8870562-8870562 on Build GRCh38
rs920544513 Chr.1: 8870570-8870570 on Build GRCh38
rs556393236 Chr.1: 8870561-8870561 on Build GRCh38
rs879912370 Chr.1: 8870470-8870470 on Build GRCh38
rs368662699 Chr.1: 8870423-8870423 on Build GRCh38
rs771992513 Chr.1: 8870437-8870437 on Build GRCh38
rs28999074 Chr.1: 8870491-8870491 on Build GRCh38
rs1053009798 Chr.1: 8870421-8870421 on Build GRCh38
rs868296090 Chr.1: 8870568-8870568 on Build GRCh38
rs754986062 Chr.1: 8870425-8870425 on Build GRCh38
rs369657785 Chr.1: 8870471-8870471 on Build GRCh38
rs760409104 Chr.1: 8870537-8870537 on Build GRCh38
rs200277735 Chr.1: 8870435-8870435 on Build GRCh38
rs558578550 Chr.1: 8870380-8870380 on Build GRCh38
rs377244479 Chr.1: 8870415-8870415 on Build GRCh38
rs113642590 Chr.1: 8870512-8870512 on Build GRCh38
rs765777536 Chr.1: 8870541-8870541 on Build GRCh38
rs929205770 Chr.1: 8870571-8870571 on Build GRCh38
rs910280066 Chr.1: 8870556-8870556 on Build GRCh38
rs906885070 Chr.1: 8870384-8870384 on Build GRCh38
rs146938354 Chr.1: 8870469-8870469 on Build GRCh38

View Full Product Details