Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 39293266-39293531 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTAGE5
Entrez Gene ID
4253
Gene Name
CTAGE family member 5, ER export factor
Gene Aliases
MEA6, MGEA, MGEA11, MGEA6
UniGene
Hs.509200
Gene Chromosome Location
Chr.14: 39265264-39351193 on Build GRCh38
Transcripts
NM_001247988.1 NM_001247989.1 NM_001247990.1 NM_005930.3 NM_203354.2 NM_203355.2 NM_203356.2 XM_005267648.4 XM_006720148.2 XM_011536775.2 XM_011536778.2 XM_011536784.2 XM_011536785.2 XM_017021314.1 XM_017021315.1 XM_017021316.1 XM_017021317.1 XM_017021318.1 XM_017021319.1 XM_017021320.1 XM_017021321.1 XM_017021322.1 XM_017021323.1 XM_017021324.1 XM_017021325.1 XM_017021326.1 XM_017021327.1 XM_017021328.1 XM_017021329.1 XM_017021330.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7082073 c.427G>A p.D143N Chr.14: 39293313-39293313 on Build GRCh38
6768786 c.354C>A p.T118T Chr.14: 39293276-39293276 on Build GRCh38
6263320 c.471C>T p.S157S Chr.14: 39293357-39293357 on Build GRCh38
6263321 c.435C>T p.S145S Chr.14: 39293357-39293357 on Build GRCh38
6637116 c.495+8delT p.? Chr.14: 39293389-39293389 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs184293867 Chr.14: 39293408-39293408 on Build GRCh38
rs975675731 Chr.14: 39293452-39293452 on Build GRCh38
rs369726723 Chr.14: 39293343-39293343 on Build GRCh38
rs776959976 Chr.14: 39293288-39293288 on Build GRCh38
rs780088436 Chr.14: 39293390-39293390 on Build GRCh38
rs776176063 Chr.14: 39293475-39293475 on Build GRCh38
rs964338110 Chr.14: 39293447-39293447 on Build GRCh38
rs752533771 Chr.14: 39293415-39293418 on Build GRCh38
rs987161395 Chr.14: 39293436-39293436 on Build GRCh38
rs1042831737 Chr.14: 39293437-39293437 on Build GRCh38
rs914787012 Chr.14: 39293461-39293461 on Build GRCh38
rs567277819 Chr.14: 39293332-39293332 on Build GRCh38
rs752546706 Chr.14: 39293365-39293365 on Build GRCh38
rs536341881 Chr.14: 39293370-39293370 on Build GRCh38
rs1052596762 Chr.14: 39293454-39293454 on Build GRCh38
rs114538941 Chr.14: 39293496-39293496 on Build GRCh38
rs563260826 Chr.14: 39293473-39293473 on Build GRCh38
rs772434558 Chr.14: 39293383-39293383 on Build GRCh38
rs764708874 Chr.14: 39293293-39293293 on Build GRCh38
rs767617793 Chr.14: 39293410-39293415 on Build GRCh38
rs544758892 Chr.14: 39293423-39293423 on Build GRCh38
rs73277460 Chr.14: 39293371-39293371 on Build GRCh38
rs899040045 Chr.14: 39293375-39293375 on Build GRCh38
rs747428298 Chr.14: 39293395-39293395 on Build GRCh38
rs768867924 Chr.14: 39293404-39293404 on Build GRCh38
rs778777792 Chr.14: 39293410-39293410 on Build GRCh38
rs765952244 Chr.14: 39293313-39293313 on Build GRCh38
rs578164342 Chr.14: 39293444-39293444 on Build GRCh38
rs751384300 Chr.14: 39293314-39293314 on Build GRCh38
rs375487092 Chr.14: 39293291-39293291 on Build GRCh38
rs558093586 Chr.14: 39293428-39293428 on Build GRCh38
rs757469583 Chr.14: 39293368-39293368 on Build GRCh38
rs754681787 Chr.14: 39293328-39293328 on Build GRCh38
rs750137503 Chr.14: 39293299-39293299 on Build GRCh38

View Full Product Details