Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 36053506-36053735 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SETD4
Entrez Gene ID
54093
Gene Name
SET domain containing 4
Gene Aliases
C21orf18, C21orf27
UniGene
Hs.606200
Gene Chromosome Location
Chr.21: 36034541-36060818 on Build GRCh38
Transcripts
NM_001007259.2 NM_001007261.2 NM_001286752.1 NM_017438.4 NR_040087.2 XM_011529636.1 XM_011529637.1 XM_011529638.2 XM_011529639.1 XM_011529640.2 XM_011529642.1 XM_011529643.1 XM_011529644.1 XM_017028403.1 XM_017028404.1 XM_017028405.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1413976 c.172A>G p.T58A Chr.21: 36053618-36053618 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs762361 Chr.21: 36053673-36053673 on Build GRCh38
rs762252160 Chr.21: 36053550-36053550 on Build GRCh38
rs752372909 Chr.21: 36053635-36053635 on Build GRCh38
rs912551868 Chr.21: 36053684-36053684 on Build GRCh38
rs768227880 Chr.21: 36053637-36053637 on Build GRCh38
rs762927622 Chr.21: 36053581-36053581 on Build GRCh38
rs754922548 Chr.21: 36053616-36053616 on Build GRCh38
rs778223967 Chr.21: 36053651-36053651 on Build GRCh38
rs777200248 Chr.21: 36053659-36053659 on Build GRCh38
rs750723502 Chr.21: 36053555-36053555 on Build GRCh38
rs965403102 Chr.21: 36053688-36053688 on Build GRCh38
rs979586975 Chr.21: 36053599-36053599 on Build GRCh38
rs770250415 Chr.21: 36053663-36053663 on Build GRCh38
rs777143977 Chr.21: 36053639-36053640 on Build GRCh38
rs769094061 Chr.21: 36053658-36053658 on Build GRCh38
rs575631068 Chr.21: 36053584-36053584 on Build GRCh38
rs1036139210 Chr.21: 36053691-36053691 on Build GRCh38
rs200711955 Chr.21: 36053551-36053551 on Build GRCh38
rs539859716 Chr.21: 36053622-36053622 on Build GRCh38
rs758512991 Chr.21: 36053680-36053680 on Build GRCh38
rs750472232 Chr.21: 36053641-36053642 on Build GRCh38
rs374443603 Chr.21: 36053669-36053669 on Build GRCh38
rs76388036 Chr.21: 36053700-36053700 on Build GRCh38
rs748457007 Chr.21: 36053662-36053662 on Build GRCh38
rs751509627 Chr.21: 36053588-36053588 on Build GRCh38
rs762360 Chr.21: 36053657-36053657 on Build GRCh38
rs373526431 Chr.21: 36053701-36053701 on Build GRCh38
rs749022139 Chr.21: 36053644-36053644 on Build GRCh38
rs915174762 Chr.21: 36053667-36053667 on Build GRCh38
rs367831332 Chr.21: 36053653-36053653 on Build GRCh38

View Full Product Details