Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 88059358-88059631 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPACA1
Entrez Gene ID
81833
Gene Name
sperm acrosome associated 1
Gene Aliases
SAMP32
UniGene
Hs.161241
Gene Chromosome Location
Chr.6: 88046907-88066832 on Build GRCh38
Transcripts
NM_030960.2 XM_011536160.2 XM_017011335.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2156304 c.520A>C p.K174Q Chr.6: 88059498-88059498 on Build GRCh38
4758578 c.518G>A p.R173H Chr.6: 88059496-88059496 on Build GRCh38
451895 c.559G>T p.D187Y Chr.6: 88059537-88059537 on Build GRCh38
297479 c.543C>T p.F181F Chr.6: 88059521-88059521 on Build GRCh38
1082389 c.475-1G>T p.? Chr.6: 88059452-88059452 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201338817 Chr.6: 88059521-88059521 on Build GRCh38
rs746640806 Chr.6: 88059592-88059592 on Build GRCh38
rs145520071 Chr.6: 88059509-88059509 on Build GRCh38
rs570946743 Chr.6: 88059393-88059394 on Build GRCh38
rs762285405 Chr.6: 88059516-88059516 on Build GRCh38
rs942414880 Chr.6: 88059435-88059435 on Build GRCh38
rs747591947 Chr.6: 88059603-88059603 on Build GRCh38
rs781199770 Chr.6: 88059474-88059474 on Build GRCh38
rs752779127 Chr.6: 88059571-88059571 on Build GRCh38
rs757316591 Chr.6: 88059495-88059495 on Build GRCh38
rs547193034 Chr.6: 88059559-88059559 on Build GRCh38
rs140977022 Chr.6: 88059529-88059529 on Build GRCh38
rs778923812 Chr.6: 88059577-88059577 on Build GRCh38
rs117078336 Chr.6: 88059581-88059581 on Build GRCh38
rs766521657 Chr.6: 88059448-88059448 on Build GRCh38
rs762371349 Chr.6: 88059410-88059410 on Build GRCh38
rs919288565 Chr.6: 88059589-88059589 on Build GRCh38
rs868835549 Chr.6: 88059450-88059450 on Build GRCh38
rs767616206 Chr.6: 88059562-88059562 on Build GRCh38
rs144924901 Chr.6: 88059548-88059548 on Build GRCh38
rs201534695 Chr.6: 88059496-88059496 on Build GRCh38
rs751805552 Chr.6: 88059545-88059545 on Build GRCh38
rs199682806 Chr.6: 88059502-88059502 on Build GRCh38
rs746695214 Chr.6: 88059511-88059511 on Build GRCh38
rs765555063 Chr.6: 88059419-88059419 on Build GRCh38
rs760894581 Chr.6: 88059433-88059434 on Build GRCh38
rs750835283 Chr.6: 88059425-88059425 on Build GRCh38
rs1033741140 Chr.6: 88059563-88059563 on Build GRCh38
rs768242414 Chr.6: 88059595-88059595 on Build GRCh38
rs61739900 Chr.6: 88059576-88059576 on Build GRCh38
rs1045886088 Chr.6: 88059523-88059523 on Build GRCh38
rs1009859130 Chr.6: 88059505-88059505 on Build GRCh38
rs763340467 Chr.6: 88059441-88059441 on Build GRCh38
rs768207176 Chr.6: 88059512-88059512 on Build GRCh38
rs750397283 Chr.6: 88059580-88059580 on Build GRCh38
rs753779949 Chr.6: 88059479-88059479 on Build GRCh38
rs200679572 Chr.6: 88059426-88059426 on Build GRCh38
rs751745168 Chr.6: 88059459-88059459 on Build GRCh38
rs779778772 Chr.6: 88059591-88059591 on Build GRCh38
rs975471254 Chr.6: 88059452-88059452 on Build GRCh38
rs61739907 Chr.6: 88059514-88059514 on Build GRCh38
rs371980319 Chr.6: 88059522-88059522 on Build GRCh38
rs199658934 Chr.6: 88059501-88059501 on Build GRCh38
rs967151019 Chr.6: 88059404-88059404 on Build GRCh38
rs369933345 Chr.6: 88059600-88059600 on Build GRCh38
rs755074009 Chr.6: 88059464-88059464 on Build GRCh38

View Full Product Details