Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 12089013-12089286 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIVEP1
Entrez Gene ID
3096
Gene Name
human immunodeficiency virus type I enhancer binding protein 1
Gene Aliases
CIRIP, CRYBP1, GAAP, MBP-1, PRDII-BF1, Schnurri-1, ZAS1, ZNF40, ZNF40A
UniGene
Hs.567284
Gene Chromosome Location
Chr.6: 12002096-12212034 on Build GRCh38
Transcripts
NM_002114.3 XM_011514546.2 XM_011514547.2 XM_011514548.2 XM_011514549.2 XM_011514550.2 XM_011514551.2 XM_011514552.2 XM_011514553.1 XM_011514555.2 XM_017010800.1 XM_017010801.1 XM_017010802.1 XM_017010803.1 XM_017010804.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6680410 c.86_87insA p.E32fs*8 Chr.6: 12089229-12089230 on Build GRCh38
4880720 c.41-7T>C p.? Chr.6: 12089177-12089177 on Build GRCh38
4853857 c.64G>C p.E22Q Chr.6: 12089207-12089207 on Build GRCh38
1634462 c.51A>G p.E17E Chr.6: 12089194-12089194 on Build GRCh38
3171000 c.87delA p.E32fs*17 Chr.6: 12089230-12089230 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777036722 Chr.6: 12089212-12089212 on Build GRCh38
rs902278468 Chr.6: 12089051-12089051 on Build GRCh38
rs946368054 Chr.6: 12089045-12089045 on Build GRCh38
rs747423574 Chr.6: 12089096-12089096 on Build GRCh38
rs962303559 Chr.6: 12089220-12089220 on Build GRCh38
rs779050520 Chr.6: 12089163-12089163 on Build GRCh38
rs999256247 Chr.6: 12089087-12089087 on Build GRCh38
rs1043389718 Chr.6: 12089050-12089050 on Build GRCh38
rs768615854 Chr.6: 12089157-12089157 on Build GRCh38
rs375502738 Chr.6: 12089186-12089186 on Build GRCh38
rs868077048 Chr.6: 12089089-12089089 on Build GRCh38
rs758872462 Chr.6: 12089172-12089172 on Build GRCh38
rs1028719189 Chr.6: 12089049-12089049 on Build GRCh38
rs954350225 Chr.6: 12089075-12089075 on Build GRCh38
rs772292927 Chr.6: 12089166-12089166 on Build GRCh38
rs749076708 Chr.6: 12089153-12089153 on Build GRCh38
rs779196618 Chr.6: 12089156-12089156 on Build GRCh38
rs765772941 Chr.6: 12089245-12089245 on Build GRCh38
rs771099895 Chr.6: 12089203-12089203 on Build GRCh38
rs779925732 Chr.6: 12089133-12089133 on Build GRCh38
rs759880305 Chr.6: 12089218-12089218 on Build GRCh38
rs1012458875 Chr.6: 12089134-12089135 on Build GRCh38
rs775984489 Chr.6: 12089250-12089250 on Build GRCh38
rs773520691 Chr.6: 12089169-12089169 on Build GRCh38
rs907781414 Chr.6: 12089144-12089144 on Build GRCh38
rs748683721 Chr.6: 12089172-12089173 on Build GRCh38
rs560180115 Chr.6: 12089219-12089219 on Build GRCh38
rs1040044984 Chr.6: 12089120-12089120 on Build GRCh38
rs747287534 Chr.6: 12089172-12089172 on Build GRCh38

View Full Product Details