Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 18249554-18249781 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEK
Entrez Gene ID
7913
Gene Name
DEK proto-oncogene
Gene Aliases
D6S231E
UniGene
Hs.484813
Gene Chromosome Location
Chr.6: 18224169-18264568 on Build GRCh38
Transcripts
NM_001134709.1 NM_003472.3 XM_017011273.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6642223 c.745A>G p.K249E Chr.6: 18249668-18249668 on Build GRCh38
4918059 c.726G>A p.E242E Chr.6: 18249687-18249687 on Build GRCh38
5884340 c.670G>A p.E224K Chr.6: 18249743-18249743 on Build GRCh38
1076394 c.756A>C p.E252D Chr.6: 18249657-18249657 on Build GRCh38
3777351 c.742G>C p.D248H Chr.6: 18249671-18249671 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1021251992 Chr.6: 18249687-18249687 on Build GRCh38
rs767101165 Chr.6: 18249733-18249733 on Build GRCh38
rs553832727 Chr.6: 18249582-18249582 on Build GRCh38
rs771093832 Chr.6: 18249700-18249700 on Build GRCh38
rs748009155 Chr.6: 18249649-18249649 on Build GRCh38
rs543058394 Chr.6: 18249640-18249640 on Build GRCh38
rs756795891 Chr.6: 18249612-18249612 on Build GRCh38
rs748859851 Chr.6: 18249666-18249668 on Build GRCh38
rs368379351 Chr.6: 18249734-18249734 on Build GRCh38
rs775732626 Chr.6: 18249676-18249676 on Build GRCh38
rs769379509 Chr.6: 18249606-18249607 on Build GRCh38
rs763357931 Chr.6: 18249731-18249731 on Build GRCh38
rs910022553 Chr.6: 18249665-18249665 on Build GRCh38
rs377602386 Chr.6: 18249611-18249611 on Build GRCh38
rs747299568 Chr.6: 18249697-18249697 on Build GRCh38
rs575836736 Chr.6: 18249627-18249627 on Build GRCh38
rs766934022 Chr.6: 18249614-18249614 on Build GRCh38
rs926824535 Chr.6: 18249746-18249746 on Build GRCh38
rs1039914491 Chr.6: 18249694-18249697 on Build GRCh38
rs186273654 Chr.6: 18249623-18249623 on Build GRCh38
rs140697635 Chr.6: 18249706-18249706 on Build GRCh38
rs760192042 Chr.6: 18249750-18249750 on Build GRCh38
rs773146547 Chr.6: 18249668-18249668 on Build GRCh38
rs766042113 Chr.6: 18249757-18249757 on Build GRCh38
rs754715706 Chr.6: 18249642-18249642 on Build GRCh38
rs942981432 Chr.6: 18249681-18249681 on Build GRCh38
rs770218067 Chr.6: 18249708-18249708 on Build GRCh38
rs779863414 Chr.6: 18249635-18249635 on Build GRCh38
rs994009625 Chr.6: 18249752-18249752 on Build GRCh38
rs958050966 Chr.6: 18249648-18249648 on Build GRCh38
rs754768955 Chr.6: 18249758-18249758 on Build GRCh38
rs772257561 Chr.6: 18249655-18249655 on Build GRCh38
rs765871877 Chr.6: 18249605-18249605 on Build GRCh38
rs200071802 Chr.6: 18249645-18249645 on Build GRCh38
rs776948529 Chr.6: 18249701-18249701 on Build GRCh38
rs200257894 Chr.6: 18249726-18249726 on Build GRCh38
rs776207453 Chr.6: 18249723-18249723 on Build GRCh38
rs775257092 Chr.6: 18249661-18249664 on Build GRCh38

View Full Product Details