Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 17803885-17804144 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF16
Entrez Gene ID
54876
Gene Name
DDB1 and CUL4 associated factor 16
Gene Aliases
C4orf30
UniGene
Hs.614787
Gene Chromosome Location
Chr.4: 17798236-17810761 on Build GRCh38
Transcripts
NM_017741.3 XM_005248169.3 XM_005248171.3 XM_006713966.3 XM_017008327.1 XM_017008328.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
460646 c.58G>T p.E20* Chr.4: 17804084-17804084 on Build GRCh38
6640474 c.230A>G p.H77R Chr.4: 17803912-17803912 on Build GRCh38
6529192 c.114A>G p.E38E Chr.4: 17804028-17804028 on Build GRCh38
334583 c.244G>T p.D82Y Chr.4: 17803898-17803898 on Build GRCh38
1729995 c.126G>T p.M42I Chr.4: 17804016-17804016 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs34085539 Chr.4: 17804008-17804008 on Build GRCh38
rs774129412 Chr.4: 17804116-17804116 on Build GRCh38
rs757220719 Chr.4: 17803931-17803931 on Build GRCh38
rs766032980 Chr.4: 17804053-17804053 on Build GRCh38
rs866757126 Chr.4: 17803924-17803924 on Build GRCh38
rs773687815 Chr.4: 17804010-17804010 on Build GRCh38
rs762154142 Chr.4: 17803920-17803920 on Build GRCh38
rs184417488 Chr.4: 17804078-17804078 on Build GRCh38
rs781370127 Chr.4: 17804108-17804108 on Build GRCh38
rs774045276 Chr.4: 17804033-17804033 on Build GRCh38
rs762591657 Chr.4: 17803907-17803907 on Build GRCh38
rs567145276 Chr.4: 17804003-17804003 on Build GRCh38
rs940479405 Chr.4: 17804111-17804111 on Build GRCh38
rs1001118601 Chr.4: 17803911-17803911 on Build GRCh38
rs1008620639 Chr.4: 17804007-17804007 on Build GRCh38
rs368200267 Chr.4: 17804085-17804085 on Build GRCh38
rs748689641 Chr.4: 17804004-17804004 on Build GRCh38
rs749400593 Chr.4: 17804002-17804002 on Build GRCh38
rs191876701 Chr.4: 17804018-17804018 on Build GRCh38
rs775214207 Chr.4: 17804019-17804019 on Build GRCh38
rs758461267 Chr.4: 17804081-17804081 on Build GRCh38
rs375394077 Chr.4: 17804021-17804021 on Build GRCh38
rs909034958 Chr.4: 17804084-17804084 on Build GRCh38
rs201315646 Chr.4: 17804061-17804061 on Build GRCh38
rs200735316 Chr.4: 17804048-17804048 on Build GRCh38
rs755844762 Chr.4: 17803999-17803999 on Build GRCh38
rs957557863 Chr.4: 17803949-17803949 on Build GRCh38
rs373206825 Chr.4: 17803915-17803915 on Build GRCh38
rs377283826 Chr.4: 17803994-17803994 on Build GRCh38
rs201013608 Chr.4: 17804079-17804079 on Build GRCh38
rs746211863 Chr.4: 17804113-17804113 on Build GRCh38
rs142767113 Chr.4: 17804035-17804035 on Build GRCh38
rs768242452 Chr.4: 17804114-17804114 on Build GRCh38
rs1036216432 Chr.4: 17804113-17804114 on Build GRCh38
rs771148926 Chr.4: 17803908-17803908 on Build GRCh38
rs755119876 Chr.4: 17804058-17804058 on Build GRCh38
rs754115187 Chr.4: 17804054-17804054 on Build GRCh38
rs138808306 Chr.4: 17804094-17804094 on Build GRCh38
rs779079995 Chr.4: 17804077-17804077 on Build GRCh38
rs146048575 Chr.4: 17804038-17804038 on Build GRCh38
rs774993985 Chr.4: 17803905-17803905 on Build GRCh38
rs372001633 Chr.4: 17804051-17804051 on Build GRCh38
rs767615779 Chr.4: 17804046-17804046 on Build GRCh38
rs374317092 Chr.4: 17804052-17804052 on Build GRCh38
rs989008425 Chr.4: 17803957-17803957 on Build GRCh38
rs747410481 Chr.4: 17804088-17804088 on Build GRCh38
rs777222946 Chr.4: 17803971-17803971 on Build GRCh38
rs751562954 Chr.4: 17803917-17803917 on Build GRCh38
rs780388370 Chr.4: 17804001-17804001 on Build GRCh38
rs199782348 Chr.4: 17804030-17804030 on Build GRCh38

View Full Product Details