Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 30285287-30285560 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BLMH
Entrez Gene ID
642
Gene Name
bleomycin hydrolase
Gene Aliases
BH, BMH
UniGene
Hs.371914
Gene Chromosome Location
Chr.17: 30248195-30292166 on Build GRCh38
Transcripts
NM_000386.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
69827 c.593G>C p.S198T Chr.17: 30285440-30285440 on Build GRCh38
4555093 c.640G>T p.E214* Chr.17: 30285393-30285393 on Build GRCh38
3755374 c.603C>G p.T201T Chr.17: 30285430-30285430 on Build GRCh38
2802271 c.610G>A p.E204K Chr.17: 30285423-30285423 on Build GRCh38
371571 c.577C>T p.R193W Chr.17: 30285456-30285456 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758647060 Chr.17: 30285465-30285465 on Build GRCh38
rs771653622 Chr.17: 30285519-30285519 on Build GRCh38
rs752875189 Chr.17: 30285399-30285399 on Build GRCh38
rs148543508 Chr.17: 30285452-30285452 on Build GRCh38
rs780354426 Chr.17: 30285507-30285507 on Build GRCh38
rs753603009 Chr.17: 30285372-30285372 on Build GRCh38
rs760621364 Chr.17: 30285356-30285356 on Build GRCh38
rs761350888 Chr.17: 30285421-30285421 on Build GRCh38
rs765597662 Chr.17: 30285456-30285456 on Build GRCh38
rs925251273 Chr.17: 30285419-30285419 on Build GRCh38
rs751783856 Chr.17: 30285341-30285342 on Build GRCh38
rs766336534 Chr.17: 30285447-30285447 on Build GRCh38
rs756469451 Chr.17: 30285506-30285506 on Build GRCh38
rs755222331 Chr.17: 30285376-30285376 on Build GRCh38
rs768248635 Chr.17: 30285418-30285418 on Build GRCh38
rs778016475 Chr.17: 30285469-30285469 on Build GRCh38
rs771600141 Chr.17: 30285425-30285425 on Build GRCh38
rs767490479 Chr.17: 30285341-30285341 on Build GRCh38
rs746264097 Chr.17: 30285505-30285505 on Build GRCh38
rs757767115 Chr.17: 30285487-30285487 on Build GRCh38
rs199684026 Chr.17: 30285528-30285528 on Build GRCh38
rs751796919 Chr.17: 30285433-30285433 on Build GRCh38
rs747820562 Chr.17: 30285518-30285518 on Build GRCh38
rs149410629 Chr.17: 30285402-30285402 on Build GRCh38
rs772776758 Chr.17: 30285525-30285525 on Build GRCh38
rs767219842 Chr.17: 30285362-30285363 on Build GRCh38
rs755221304 Chr.17: 30285354-30285355 on Build GRCh38
rs1006705389 Chr.17: 30285500-30285500 on Build GRCh38
rs552488995 Chr.17: 30285390-30285390 on Build GRCh38
rs781713523 Chr.17: 30285501-30285501 on Build GRCh38
rs367890382 Chr.17: 30285484-30285484 on Build GRCh38
rs777962429 Chr.17: 30285406-30285406 on Build GRCh38
rs7342921 Chr.17: 30285430-30285430 on Build GRCh38
rs771415262 Chr.17: 30285414-30285414 on Build GRCh38
rs753634567 Chr.17: 30285451-30285451 on Build GRCh38
rs752430988 Chr.17: 30285421-30285422 on Build GRCh38
rs753002041 Chr.17: 30285462-30285462 on Build GRCh38
rs374487604 Chr.17: 30285527-30285527 on Build GRCh38
rs190498822 Chr.17: 30285415-30285415 on Build GRCh38
rs185643713 Chr.17: 30285315-30285315 on Build GRCh38
rs758540736 Chr.17: 30285416-30285416 on Build GRCh38
rs368757390 Chr.17: 30285357-30285357 on Build GRCh38
rs760542660 Chr.17: 30285431-30285431 on Build GRCh38
rs747545839 Chr.17: 30285408-30285408 on Build GRCh38
rs936615994 Chr.17: 30285429-30285429 on Build GRCh38
rs750327577 Chr.17: 30285345-30285345 on Build GRCh38

View Full Product Details