Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 27382333-27382592 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD164L2
Entrez Gene ID
388611
Gene Name
CD164 molecule like 2
Gene Aliases
EAPG6122, UNQ6122
UniGene
Hs.664836
Gene Chromosome Location
Chr.1: 27379100-27383333 on Build GRCh38
Transcripts
NM_207397.3 XM_005245868.3 XM_011541441.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4854969 c.232G>C p.E78Q Chr.1: 27382524-27382524 on Build GRCh38
4847030 c.219C>T p.S73S Chr.1: 27382537-27382537 on Build GRCh38
7360201 c.241C>T p.R81W Chr.1: 27382515-27382515 on Build GRCh38
4905470 c.199G>A p.D67N Chr.1: 27382557-27382557 on Build GRCh38
6623484 c.225C>T p.C75C Chr.1: 27382531-27382531 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2474297 Chr.1: 27382554-27382554 on Build GRCh38
rs774604501 Chr.1: 27382440-27382440 on Build GRCh38
rs892139472 Chr.1: 27382544-27382544 on Build GRCh38
rs749630215 Chr.1: 27382514-27382514 on Build GRCh38
rs2504779 Chr.1: 27382530-27382530 on Build GRCh38
rs924855853 Chr.1: 27382427-27382427 on Build GRCh38
rs199814220 Chr.1: 27382506-27382506 on Build GRCh38
rs201003060 Chr.1: 27382528-27382528 on Build GRCh38
rs771324495 Chr.1: 27382515-27382515 on Build GRCh38
rs372070371 Chr.1: 27382419-27382419 on Build GRCh38
rs779916718 Chr.1: 27382405-27382405 on Build GRCh38
rs781135338 Chr.1: 27382442-27382442 on Build GRCh38
rs761749706 Chr.1: 27382468-27382468 on Build GRCh38
rs545591475 Chr.1: 27382563-27382563 on Build GRCh38
rs749720301 Chr.1: 27382438-27382438 on Build GRCh38
rs140375229 Chr.1: 27382503-27382503 on Build GRCh38
rs773326480 Chr.1: 27382532-27382532 on Build GRCh38
rs770217281 Chr.1: 27382420-27382420 on Build GRCh38
rs980967918 Chr.1: 27382400-27382400 on Build GRCh38
rs750224271 Chr.1: 27382475-27382475 on Build GRCh38
rs778203250 Chr.1: 27382511-27382511 on Build GRCh38
rs374864144 Chr.1: 27382549-27382549 on Build GRCh38
rs758320628 Chr.1: 27382477-27382477 on Build GRCh38
rs765030275 Chr.1: 27382369-27382369 on Build GRCh38
rs775570390 Chr.1: 27382523-27382523 on Build GRCh38
rs369404538 Chr.1: 27382408-27382408 on Build GRCh38
rs376739270 Chr.1: 27382421-27382421 on Build GRCh38
rs752238611 Chr.1: 27382550-27382550 on Build GRCh38
rs745597977 Chr.1: 27382486-27382486 on Build GRCh38
rs775454054 Chr.1: 27382465-27382465 on Build GRCh38
rs866498408 Chr.1: 27382393-27382393 on Build GRCh38
rs115765119 Chr.1: 27382371-27382371 on Build GRCh38
rs139730633 Chr.1: 27382567-27382567 on Build GRCh38
rs767591758 Chr.1: 27382456-27382456 on Build GRCh38
rs369848375 Chr.1: 27382439-27382439 on Build GRCh38
rs553077725 Chr.1: 27382545-27382545 on Build GRCh38
rs746040059 Chr.1: 27382516-27382516 on Build GRCh38
rs966337205 Chr.1: 27382395-27382395 on Build GRCh38
rs766102186 Chr.1: 27382479-27382479 on Build GRCh38
rs535850283 Chr.1: 27382543-27382543 on Build GRCh38
rs759199032 Chr.1: 27382547-27382547 on Build GRCh38
rs758054516 Chr.1: 27382376-27382376 on Build GRCh38
rs754579947 Chr.1: 27382416-27382416 on Build GRCh38
rs765088953 Chr.1: 27382470-27382470 on Build GRCh38
rs752154370 Chr.1: 27382504-27382504 on Build GRCh38
rs375908957 Chr.1: 27382555-27382555 on Build GRCh38
rs373655798 Chr.1: 27382539-27382539 on Build GRCh38
rs772193693 Chr.1: 27382517-27382517 on Build GRCh38
rs376208723 Chr.1: 27382484-27382484 on Build GRCh38
rs558677751 Chr.1: 27382566-27382566 on Build GRCh38
rs201417742 Chr.1: 27382446-27382446 on Build GRCh38
rs754702685 Chr.1: 27382494-27382494 on Build GRCh38
rs753852121 Chr.1: 27382363-27382363 on Build GRCh38

View Full Product Details