Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 118096588-118096835 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INSIG2
Entrez Gene ID
51141
Gene Name
insulin induced gene 2
Gene Aliases
INSIG-2
UniGene
Hs.7089
Gene Chromosome Location
Chr.2: 118088418-118110031 on Build GRCh38
Transcripts
NM_001321329.1 NM_001321330.1 NM_001321331.1 NM_001321332.1 NM_001321333.1 NM_016133.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4084419 c.187A>T p.I63F Chr.2: 118096743-118096743 on Build GRCh38
3961039 c.127G>T p.A43S Chr.2: 118096683-118096683 on Build GRCh38
6482774 c.95G>T p.G32V Chr.2: 118096651-118096651 on Build GRCh38
3836674 c.177A>C p.P59P Chr.2: 118096733-118096733 on Build GRCh38
6542969 c.148C>T p.Q50* Chr.2: 118096704-118096704 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs922095354 Chr.2: 118096627-118096627 on Build GRCh38
rs756111660 Chr.2: 118096688-118096688 on Build GRCh38
rs781017559 Chr.2: 118096656-118096656 on Build GRCh38
rs940346154 Chr.2: 118096775-118096775 on Build GRCh38
rs773442222 Chr.2: 118096769-118096769 on Build GRCh38
rs764572574 Chr.2: 118096777-118096777 on Build GRCh38
rs376950162 Chr.2: 118096724-118096724 on Build GRCh38
rs765415653 Chr.2: 118096674-118096674 on Build GRCh38
rs568055123 Chr.2: 118096786-118096786 on Build GRCh38
rs138954785 Chr.2: 118096727-118096727 on Build GRCh38
rs762335199 Chr.2: 118096703-118096704 on Build GRCh38
rs375348923 Chr.2: 118096622-118096622 on Build GRCh38
rs751179994 Chr.2: 118096796-118096796 on Build GRCh38
rs765960423 Chr.2: 118096615-118096615 on Build GRCh38
rs76370593 Chr.2: 118096800-118096800 on Build GRCh38
rs745585272 Chr.2: 118096792-118096792 on Build GRCh38
rs746239528 Chr.2: 118096734-118096734 on Build GRCh38
rs375157361 Chr.2: 118096793-118096793 on Build GRCh38
rs748170040 Chr.2: 118096669-118096669 on Build GRCh38
rs150499979 Chr.2: 118096707-118096707 on Build GRCh38
rs754766313 Chr.2: 118096631-118096631 on Build GRCh38
rs776841805 Chr.2: 118096700-118096701 on Build GRCh38
rs761312280 Chr.2: 118096752-118096752 on Build GRCh38
rs749320625 Chr.2: 118096723-118096723 on Build GRCh38
rs539714248 Chr.2: 118096742-118096742 on Build GRCh38
rs772723612 Chr.2: 118096790-118096790 on Build GRCh38
rs770287324 Chr.2: 118096705-118096706 on Build GRCh38
rs932227381 Chr.2: 118096721-118096721 on Build GRCh38
rs890738296 Chr.2: 118096745-118096745 on Build GRCh38
rs775687117 Chr.2: 118096749-118096749 on Build GRCh38
rs978916683 Chr.2: 118096624-118096624 on Build GRCh38

View Full Product Details