Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 41016013-41016260 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLFNL1
Entrez Gene ID
200172
Gene Name
schlafen like 1
Gene Aliases
-
UniGene
Hs.194609
Gene Chromosome Location
Chr.1: 41015597-41023444 on Build GRCh38
Transcripts
NM_001168247.2 NM_001300859.1 NM_144990.3 XM_005270597.2 XM_005270598.2 XM_005270599.2 XM_006710432.3 XM_011540943.1 XM_011540945.1 XM_011540947.1 XM_011540948.1 XM_011540949.1 XM_011540950.1 XM_011540951.1 XM_011540952.2 XM_011540953.2 XM_011540954.1 XM_017000573.1 XM_017000574.1 NR_037868.1

Gene Symbol
SLFNL1-AS1
Entrez Gene ID
100507178
Gene Name
SLFNL1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.660056
Gene Chromosome Location
Chr.1: 41014590-41043890 on Build GRCh38
Transcripts
NM_001168247.2 NM_001300859.1 NM_144990.3 XM_005270597.2 XM_005270598.2 XM_005270599.2 XM_006710432.3 XM_011540943.1 XM_011540945.1 XM_011540947.1 XM_011540948.1 XM_011540949.1 XM_011540950.1 XM_011540951.1 XM_011540952.2 XM_011540953.2 XM_011540954.1 XM_017000573.1 XM_017000574.1 NR_037868.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
909517 c.1191C>T p.H397H Chr.1: 41016139-41016139 on Build GRCh38
7160622 c.1140G>A p.M380I Chr.1: 41016190-41016190 on Build GRCh38
110432 c.1158G>A p.E386E Chr.1: 41016172-41016172 on Build GRCh38
4522311 c.1156G>A p.E386K Chr.1: 41016174-41016174 on Build GRCh38
7080389 c.1198G>C p.V400L Chr.1: 41016132-41016132 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746828416 Chr.1: 41016219-41016219 on Build GRCh38
rs760632146 Chr.1: 41016119-41016119 on Build GRCh38
rs898846613 Chr.1: 41016168-41016168 on Build GRCh38
rs751918350 Chr.1: 41016078-41016078 on Build GRCh38
rs200327092 Chr.1: 41016066-41016066 on Build GRCh38
rs761397562 Chr.1: 41016239-41016239 on Build GRCh38
rs112540189 Chr.1: 41016064-41016064 on Build GRCh38
rs200527951 Chr.1: 41016139-41016139 on Build GRCh38
rs75898383 Chr.1: 41016042-41016042 on Build GRCh38
rs751814836 Chr.1: 41016141-41016149 on Build GRCh38
rs558283454 Chr.1: 41016192-41016194 on Build GRCh38
rs756564867 Chr.1: 41016183-41016183 on Build GRCh38
rs1018979651 Chr.1: 41016199-41016199 on Build GRCh38
rs769258280 Chr.1: 41016240-41016240 on Build GRCh38
rs779660426 Chr.1: 41016217-41016217 on Build GRCh38
rs779190149 Chr.1: 41016208-41016208 on Build GRCh38
rs142166637 Chr.1: 41016172-41016172 on Build GRCh38
rs764917731 Chr.1: 41016144-41016144 on Build GRCh38
rs775359624 Chr.1: 41016116-41016116 on Build GRCh38
rs201233485 Chr.1: 41016138-41016138 on Build GRCh38
rs779045649 Chr.1: 41016099-41016099 on Build GRCh38
rs766629180 Chr.1: 41016140-41016151 on Build GRCh38
rs768733429 Chr.1: 41016124-41016124 on Build GRCh38
rs755097422 Chr.1: 41016169-41016171 on Build GRCh38
rs768673867 Chr.1: 41016185-41016185 on Build GRCh38
rs1013087236 Chr.1: 41016141-41016141 on Build GRCh38
rs376355039 Chr.1: 41016236-41016236 on Build GRCh38
rs756653141 Chr.1: 41016069-41016070 on Build GRCh38
rs755349248 Chr.1: 41016079-41016079 on Build GRCh38
rs915229511 Chr.1: 41016055-41016055 on Build GRCh38
rs768680414 Chr.1: 41016226-41016226 on Build GRCh38
rs745660099 Chr.1: 41016103-41016103 on Build GRCh38
rs760945226 Chr.1: 41016082-41016082 on Build GRCh38
rs777772666 Chr.1: 41016085-41016085 on Build GRCh38
rs771103533 Chr.1: 41016096-41016096 on Build GRCh38
rs546646656 Chr.1: 41016083-41016083 on Build GRCh38
rs757383404 Chr.1: 41016204-41016204 on Build GRCh38
rs750104018 Chr.1: 41016152-41016152 on Build GRCh38
rs749393274 Chr.1: 41016092-41016092 on Build GRCh38
rs201980070 Chr.1: 41016184-41016184 on Build GRCh38
rs768402041 Chr.1: 41016212-41016212 on Build GRCh38
rs781717526 Chr.1: 41016081-41016081 on Build GRCh38
rs748420105 Chr.1: 41016227-41016228 on Build GRCh38
rs374134776 Chr.1: 41016077-41016077 on Build GRCh38
rs372833325 Chr.1: 41016214-41016214 on Build GRCh38
rs753408231 Chr.1: 41016071-41016071 on Build GRCh38
rs764856759 Chr.1: 41016036-41016037 on Build GRCh38
rs926667523 Chr.1: 41016104-41016104 on Build GRCh38
rs1044028780 Chr.1: 41016142-41016142 on Build GRCh38
rs140785040 Chr.1: 41016162-41016162 on Build GRCh38
rs763630418 Chr.1: 41016068-41016068 on Build GRCh38
rs758519191 Chr.1: 41016116-41016117 on Build GRCh38
rs369206165 Chr.1: 41016133-41016133 on Build GRCh38
rs771827727 Chr.1: 41016105-41016105 on Build GRCh38
rs775346090 Chr.1: 41016058-41016058 on Build GRCh38
rs767712399 Chr.1: 41016171-41016171 on Build GRCh38

View Full Product Details