Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 45069613-45069886 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCM2
Entrez Gene ID
83605
Gene Name
CCM2 scaffolding protein
Gene Aliases
C7orf22, OSM, PP10187
UniGene
Hs.148272
Gene Chromosome Location
Chr.7: 44999746-45076470 on Build GRCh38
Transcripts
NM_001029835.2 NM_001167934.1 NM_001167935.1 NM_031443.3 NR_030770.1 XM_006715785.3 XM_006715786.3 XM_011515561.2 XM_011515562.1 XM_011515563.2 XM_011515564.2 XM_017012671.1 XM_017012672.1 XM_017012673.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3263244 c.675C>T p.V225V Chr.7: 45069828-45069828 on Build GRCh38
290043 c.697delC p.L233fs*2 Chr.7: 45069850-45069850 on Build GRCh38
7300208 c.681G>A p.A227A Chr.7: 45069834-45069834 on Build GRCh38
1090031 c.680C>T p.A227V Chr.7: 45069833-45069833 on Build GRCh38
6622882 c.723T>C p.V241V Chr.7: 45069876-45069876 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs539468913 Chr.7: 45069800-45069800 on Build GRCh38
rs902403681 Chr.7: 45069819-45069819 on Build GRCh38
rs982697196 Chr.7: 45069675-45069675 on Build GRCh38
rs190206358 Chr.7: 45069669-45069669 on Build GRCh38
rs759257759 Chr.7: 45069822-45069822 on Build GRCh38
rs377330230 Chr.7: 45069811-45069811 on Build GRCh38
rs372454105 Chr.7: 45069681-45069681 on Build GRCh38
rs772744146 Chr.7: 45069779-45069779 on Build GRCh38
rs764635326 Chr.7: 45069776-45069776 on Build GRCh38
rs947328509 Chr.7: 45069650-45069650 on Build GRCh38
rs913146648 Chr.7: 45069649-45069649 on Build GRCh38
rs909875982 Chr.7: 45069751-45069751 on Build GRCh38
rs766074926 Chr.7: 45069805-45069805 on Build GRCh38
rs921724161 Chr.7: 45069666-45069666 on Build GRCh38
rs918808809 Chr.7: 45069678-45069678 on Build GRCh38
rs970913159 Chr.7: 45069651-45069651 on Build GRCh38
rs150076154 Chr.7: 45069852-45069852 on Build GRCh38
rs112514422 Chr.7: 45069648-45069648 on Build GRCh38
rs756126156 Chr.7: 45069829-45069829 on Build GRCh38
rs978711914 Chr.7: 45069658-45069658 on Build GRCh38
rs1039757556 Chr.7: 45069780-45069780 on Build GRCh38
rs1023429314 Chr.7: 45069648-45069648 on Build GRCh38
rs777710102 Chr.7: 45069833-45069833 on Build GRCh38
rs568877034 Chr.7: 45069723-45069723 on Build GRCh38
rs774877844 Chr.7: 45069729-45069729 on Build GRCh38
rs933132482 Chr.7: 45069806-45069806 on Build GRCh38
rs1036476643 Chr.7: 45069855-45069855 on Build GRCh38
rs922710733 Chr.7: 45069801-45069801 on Build GRCh38
rs1051483056 Chr.7: 45069766-45069766 on Build GRCh38
rs181474100 Chr.7: 45069828-45069828 on Build GRCh38
rs754030131 Chr.7: 45069850-45069850 on Build GRCh38
rs984060809 Chr.7: 45069737-45069737 on Build GRCh38

View Full Product Details