Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 43648026-43648263 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CATSPER2
Entrez Gene ID
117155
Gene Name
cation channel sperm associated 2
Gene Aliases
-
UniGene
Hs.662284
Gene Chromosome Location
Chr.15: 43630562-43648845 on Build GRCh38
Transcripts
NM_001282309.1 NM_001282310.1 NM_054020.3 NM_172095.2 NR_110319.1 XM_017021904.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
433930 c.27G>A p.Q9Q Chr.15: 43648035-43648035 on Build GRCh38
3501511 c.35T>C p.L12P Chr.15: 43648027-43648027 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs909694245 Chr.15: 43648233-43648233 on Build GRCh38
rs775681786 Chr.15: 43648180-43648180 on Build GRCh38
rs747909781 Chr.15: 43648113-43648113 on Build GRCh38
rs753160194 Chr.15: 43648063-43648063 on Build GRCh38
rs7162321 Chr.15: 43648159-43648159 on Build GRCh38
rs146757686 Chr.15: 43648055-43648055 on Build GRCh38
rs764314779 Chr.15: 43648212-43648212 on Build GRCh38
rs146995350 Chr.15: 43648102-43648104 on Build GRCh38
rs542864714 Chr.15: 43648196-43648196 on Build GRCh38
rs982671886 Chr.15: 43648220-43648220 on Build GRCh38
rs1000388056 Chr.15: 43648136-43648136 on Build GRCh38
rs543310402 Chr.15: 43648206-43648206 on Build GRCh38
rs1034334068 Chr.15: 43648145-43648145 on Build GRCh38
rs574106065 Chr.15: 43648188-43648188 on Build GRCh38
rs761351527 Chr.15: 43648053-43648053 on Build GRCh38
rs780837499 Chr.15: 43648111-43648111 on Build GRCh38
rs757281550 Chr.15: 43648090-43648090 on Build GRCh38
rs958672054 Chr.15: 43648156-43648156 on Build GRCh38
rs752069856 Chr.15: 43648058-43648058 on Build GRCh38
rs768687783 Chr.15: 43648110-43648110 on Build GRCh38
rs1024355371 Chr.15: 43648194-43648196 on Build GRCh38
rs1050802817 Chr.15: 43648062-43648062 on Build GRCh38
rs778996275 Chr.15: 43648105-43648105 on Build GRCh38
rs202093954 Chr.15: 43648072-43648072 on Build GRCh38
rs746919652 Chr.15: 43648106-43648106 on Build GRCh38
rs760126816 Chr.15: 43648061-43648061 on Build GRCh38
rs904805113 Chr.15: 43648132-43648132 on Build GRCh38
rs766008928 Chr.15: 43648056-43648056 on Build GRCh38
rs138765145 Chr.15: 43648066-43648066 on Build GRCh38

View Full Product Details