Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1966334-1966565 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP74
Entrez Gene ID
85452
Gene Name
cilia and flagella associated protein 74
Gene Aliases
C1orf222, KIAA1751
UniGene
Hs.232092
Gene Chromosome Location
Chr.1: 1921951-2003837 on Build GRCh38
Transcripts
NM_001304360.1 XM_011542333.2 XM_011542335.1 XM_011542336.2 XM_017002641.1 XM_017002642.1 XM_017002643.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6237919 c.1303G>T p.E435* Chr.1: 1966469-1966469 on Build GRCh38
901261 c.1393C>T p.P465S Chr.1: 1966379-1966379 on Build GRCh38
4885591 c.1269C>T p.P423P Chr.1: 1966503-1966503 on Build GRCh38
1255751 c.1346C>T p.T449M Chr.1: 1966426-1966426 on Build GRCh38
5950534 c.1247A>G p.D416G Chr.1: 1966525-1966525 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373629182 Chr.1: 1966502-1966502 on Build GRCh38
rs747878199 Chr.1: 1966501-1966501 on Build GRCh38
rs749252978 Chr.1: 1966492-1966492 on Build GRCh38
rs778815142 Chr.1: 1966499-1966499 on Build GRCh38
rs754334604 Chr.1: 1966368-1966368 on Build GRCh38
rs60018876 Chr.1: 1966487-1966487 on Build GRCh38
rs773974855 Chr.1: 1966425-1966425 on Build GRCh38
rs61734009 Chr.1: 1966521-1966521 on Build GRCh38
rs761563997 Chr.1: 1966426-1966426 on Build GRCh38
rs754420662 Chr.1: 1966451-1966451 on Build GRCh38
rs765674924 Chr.1: 1966462-1966462 on Build GRCh38
rs769789385 Chr.1: 1966402-1966402 on Build GRCh38
rs760270293 Chr.1: 1966449-1966449 on Build GRCh38
rs776250661 Chr.1: 1966510-1966510 on Build GRCh38
rs755410642 Chr.1: 1966371-1966371 on Build GRCh38
rs370517519 Chr.1: 1966479-1966479 on Build GRCh38
rs772756874 Chr.1: 1966503-1966503 on Build GRCh38
rs999796766 Chr.1: 1966369-1966369 on Build GRCh38
rs201162759 Chr.1: 1966534-1966534 on Build GRCh38
rs766992282 Chr.1: 1966354-1966354 on Build GRCh38
rs1022532102 Chr.1: 1966529-1966529 on Build GRCh38
rs772833233 Chr.1: 1966448-1966448 on Build GRCh38
rs770600149 Chr.1: 1966507-1966507 on Build GRCh38
rs954778503 Chr.1: 1966498-1966498 on Build GRCh38
rs202215201 Chr.1: 1966383-1966383 on Build GRCh38
rs779537087 Chr.1: 1966383-1966386 on Build GRCh38
rs35213058 Chr.1: 1966458-1966459 on Build GRCh38
rs763230262 Chr.1: 1966526-1966526 on Build GRCh38
rs765648888 Chr.1: 1966515-1966515 on Build GRCh38
rs553969437 Chr.1: 1966488-1966488 on Build GRCh38
rs144739795 Chr.1: 1966393-1966393 on Build GRCh38
rs760358107 Chr.1: 1966505-1966505 on Build GRCh38
rs751736348 Chr.1: 1966480-1966480 on Build GRCh38
rs777958583 Chr.1: 1966388-1966388 on Build GRCh38
rs145774922 Chr.1: 1966478-1966478 on Build GRCh38
rs760172100 Chr.1: 1966511-1966511 on Build GRCh38
rs779971499 Chr.1: 1966404-1966404 on Build GRCh38
rs376236547 Chr.1: 1966413-1966413 on Build GRCh38
rs771598585 Chr.1: 1966431-1966431 on Build GRCh38
rs41310347 Chr.1: 1966455-1966455 on Build GRCh38
rs371732380 Chr.1: 1966528-1966528 on Build GRCh38
rs368393908 Chr.1: 1966545-1966545 on Build GRCh38
rs758519763 Chr.1: 1966385-1966385 on Build GRCh38
rs752984029 Chr.1: 1966465-1966465 on Build GRCh38
rs749164767 Chr.1: 1966412-1966412 on Build GRCh38
rs376720703 Chr.1: 1966535-1966535 on Build GRCh38
rs544429524 Chr.1: 1966530-1966530 on Build GRCh38
rs768546253 Chr.1: 1966493-1966493 on Build GRCh38
rs533765020 Chr.1: 1966483-1966483 on Build GRCh38
rs958069592 Chr.1: 1966401-1966401 on Build GRCh38
rs61734007 Chr.1: 1966450-1966450 on Build GRCh38

View Full Product Details