Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81084763-81084992 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAIAP2
Entrez Gene ID
10458
Gene Name
BAI1 associated protein 2
Gene Aliases
BAP2, FLAF3, IRSP53
UniGene
Hs.128316
Gene Chromosome Location
Chr.17: 81035147-81117432 on Build GRCh38
Transcripts
NM_001144888.1 NM_006340.2 NM_017450.2 NM_017451.2 XM_005256943.1 XM_005256944.1 XM_005256945.1 XM_005256948.3 XM_006721635.1 XM_006721636.1 XM_006721637.2 XM_011524193.1 XM_011524194.1 XM_017024017.1 XM_017024018.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3523470 c.228C>T p.L76L Chr.17: 81084842-81084842 on Build GRCh38
5796255 c.222C>T p.D74D Chr.17: 81084836-81084836 on Build GRCh38
4665117 c.274G>T p.E92* Chr.17: 81084888-81084888 on Build GRCh38
6295931 c.279+5G>T p.? Chr.17: 81084898-81084898 on Build GRCh38
5796256 c.222C>T p.D74D Chr.17: 81084836-81084836 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs890142261 Chr.17: 81084787-81084787 on Build GRCh38
rs369625815 Chr.17: 81084826-81084826 on Build GRCh38
rs765624125 Chr.17: 81084920-81084920 on Build GRCh38
rs958763714 Chr.17: 81084876-81084876 on Build GRCh38
rs546067385 Chr.17: 81084849-81084849 on Build GRCh38
rs774959989 Chr.17: 81084916-81084916 on Build GRCh38
rs750807245 Chr.17: 81084921-81084921 on Build GRCh38
rs760386011 Chr.17: 81084939-81084939 on Build GRCh38
rs777643110 Chr.17: 81084892-81084892 on Build GRCh38
rs963657676 Chr.17: 81084966-81084966 on Build GRCh38
rs769593965 Chr.17: 81084855-81084855 on Build GRCh38
rs751902118 Chr.17: 81084931-81084931 on Build GRCh38
rs769441194 Chr.17: 81084931-81084931 on Build GRCh38
rs900046945 Chr.17: 81084845-81084845 on Build GRCh38
rs549634906 Chr.17: 81084965-81084965 on Build GRCh38
rs765683160 Chr.17: 81084797-81084797 on Build GRCh38
rs372033045 Chr.17: 81084836-81084836 on Build GRCh38
rs766840877 Chr.17: 81084925-81084925 on Build GRCh38
rs372465591 Chr.17: 81084937-81084937 on Build GRCh38
rs756105844 Chr.17: 81084940-81084940 on Build GRCh38
rs758680350 Chr.17: 81084806-81084806 on Build GRCh38
rs531317082 Chr.17: 81084951-81084951 on Build GRCh38
rs369917301 Chr.17: 81084910-81084910 on Build GRCh38
rs755171519 Chr.17: 81084828-81084828 on Build GRCh38
rs991859286 Chr.17: 81084960-81084960 on Build GRCh38
rs780076647 Chr.17: 81084823-81084823 on Build GRCh38
rs762405590 Chr.17: 81084917-81084917 on Build GRCh38
rs754400575 Chr.17: 81084783-81084783 on Build GRCh38
rs747967420 Chr.17: 81084922-81084923 on Build GRCh38
rs764597188 Chr.17: 81084782-81084782 on Build GRCh38
rs375538586 Chr.17: 81084804-81084804 on Build GRCh38
rs763173308 Chr.17: 81084923-81084923 on Build GRCh38
rs762277193 Chr.17: 81084786-81084786 on Build GRCh38
rs960043853 Chr.17: 81084952-81084952 on Build GRCh38
rs748994634 Chr.17: 81084906-81084906 on Build GRCh38
rs771610575 Chr.17: 81084911-81084911 on Build GRCh38
rs376653089 Chr.17: 81084907-81084907 on Build GRCh38
rs774950154 Chr.17: 81084936-81084936 on Build GRCh38

View Full Product Details